Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80433973-80434417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE16
Entrez Gene ID
9765
Gene Name
zinc finger FYVE-type containing 16
Gene Aliases
PPP1R69
UniGene
Hs.482660
Gene Chromosome Location
Chr.5: 80407615-80479869 on Build GRCh38
Transcripts
NM_001105251.2 NM_001284236.1 NM_001284237.1 NM_014733.4 XM_005248632.4 XM_011543753.2 XM_017010091.1 XM_017010092.1 XM_017010095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618302 c.45C>T p.L15L Chr.5: 80434192-80434192 on Build GRCh38
7141589 c.52G>T p.D18Y Chr.5: 80434199-80434199 on Build GRCh38
278215 c.63G>T p.Q21H Chr.5: 80434210-80434210 on Build GRCh38
1070647 c.29G>A p.S10N Chr.5: 80434176-80434176 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371014147 Chr.5: 80434129-80434129 on Build GRCh38
rs1042472978 Chr.5: 80434067-80434067 on Build GRCh38
rs573856632 Chr.5: 80434051-80434051 on Build GRCh38
rs866748288 Chr.5: 80434154-80434154 on Build GRCh38
rs1057458324 Chr.5: 80434388-80434388 on Build GRCh38
rs771140703 Chr.5: 80434155-80434155 on Build GRCh38
rs376635939 Chr.5: 80434266-80434266 on Build GRCh38
rs544655261 Chr.5: 80434255-80434255 on Build GRCh38
rs1054848951 Chr.5: 80434344-80434344 on Build GRCh38
rs371332632 Chr.5: 80434264-80434264 on Build GRCh38
rs770378410 Chr.5: 80434205-80434205 on Build GRCh38
rs754305562 Chr.5: 80434100-80434100 on Build GRCh38
rs995452891 Chr.5: 80434023-80434023 on Build GRCh38
rs1016039885 Chr.5: 80434206-80434206 on Build GRCh38
rs1004272923 Chr.5: 80434347-80434347 on Build GRCh38
rs375888570 Chr.5: 80434127-80434127 on Build GRCh38
rs377381919 Chr.5: 80434098-80434098 on Build GRCh38
rs754147688 Chr.5: 80434327-80434327 on Build GRCh38
rs758132748 Chr.5: 80434255-80434255 on Build GRCh38
rs375038540 Chr.5: 80434243-80434243 on Build GRCh38
rs751738899 Chr.5: 80434231-80434231 on Build GRCh38
rs368613691 Chr.5: 80434241-80434241 on Build GRCh38
rs896349963 Chr.5: 80434394-80434394 on Build GRCh38
rs529588810 Chr.5: 80434326-80434326 on Build GRCh38
rs368684694 Chr.5: 80434128-80434128 on Build GRCh38
rs374342948 Chr.5: 80434209-80434209 on Build GRCh38
rs866872285 Chr.5: 80434014-80434014 on Build GRCh38
rs903562590 Chr.5: 80434290-80434290 on Build GRCh38
rs531190738 Chr.5: 80434145-80434145 on Build GRCh38
rs1023961805 Chr.5: 80434114-80434114 on Build GRCh38
rs748038650 Chr.5: 80434261-80434261 on Build GRCh38
rs552922248 Chr.5: 80434141-80434141 on Build GRCh38
rs974167148 Chr.5: 80434232-80434232 on Build GRCh38
rs749257174 Chr.5: 80434138-80434138 on Build GRCh38
rs894908466 Chr.5: 80434383-80434383 on Build GRCh38
rs753111682 Chr.5: 80434113-80434113 on Build GRCh38
rs753199674 Chr.5: 80434223-80434223 on Build GRCh38
rs903593332 Chr.5: 80434070-80434070 on Build GRCh38
rs1009214027 Chr.5: 80434041-80434041 on Build GRCh38
rs779856208 Chr.5: 80434269-80434273 on Build GRCh38
rs774397481 Chr.5: 80434143-80434143 on Build GRCh38
rs776158955 Chr.5: 80434207-80434207 on Build GRCh38
rs765612471 Chr.5: 80434101-80434101 on Build GRCh38
rs763369159 Chr.5: 80434229-80434229 on Build GRCh38
rs371588030 Chr.5: 80434242-80434242 on Build GRCh38
rs1000653493 Chr.5: 80434093-80434093 on Build GRCh38
rs2098691 Chr.5: 80434158-80434158 on Build GRCh38
rs111815748 Chr.5: 80434316-80434316 on Build GRCh38
rs887076067 Chr.5: 80434332-80434332 on Build GRCh38
rs759024800 Chr.5: 80434208-80434208 on Build GRCh38
rs150835848 Chr.5: 80434259-80434259 on Build GRCh38
rs931352319 Chr.5: 80434299-80434299 on Build GRCh38
rs982782803 Chr.5: 80434170-80434170 on Build GRCh38
rs887189100 Chr.5: 80434049-80434049 on Build GRCh38
rs760308625 Chr.5: 80434194-80434194 on Build GRCh38
rs962760296 Chr.5: 80434213-80434213 on Build GRCh38
rs370449302 Chr.5: 80434117-80434117 on Build GRCh38
rs533318667 Chr.5: 80434265-80434265 on Build GRCh38
rs1027060286 Chr.5: 80434045-80434045 on Build GRCh38
rs764465026 Chr.5: 80434230-80434230 on Build GRCh38
rs543982090 Chr.5: 80434281-80434281 on Build GRCh38
rs139164403 Chr.5: 80434159-80434159 on Build GRCh38
rs764855948 Chr.5: 80434287-80434287 on Build GRCh38
rs781269944 Chr.5: 80434137-80434137 on Build GRCh38
rs188072477 Chr.5: 80434033-80434033 on Build GRCh38
rs868466047 Chr.5: 80434048-80434048 on Build GRCh38
rs879149774 Chr.5: 80434212-80434212 on Build GRCh38
rs1042050582 Chr.5: 80434288-80434288 on Build GRCh38
rs1004213675 Chr.5: 80434068-80434068 on Build GRCh38
rs765854783 Chr.5: 80434099-80434099 on Build GRCh38
rs372448223 Chr.5: 80434165-80434165 on Build GRCh38

View Full Product Details