Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80665137-80665686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
UniGene
Hs.280987
Gene Chromosome Location
Chr.5: 80654648-80876815 on Build GRCh38
Transcripts
NM_002439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6475123 c.540A>G p.A180A Chr.5: 80665324-80665324 on Build GRCh38
6470539 c.382A>G p.R128G Chr.5: 80665166-80665166 on Build GRCh38
5715883 c.440C>T p.P147L Chr.5: 80665224-80665224 on Build GRCh38
3429714 c.339G>T p.K113N Chr.5: 80665150-80665150 on Build GRCh38
6475119 c.562C>A p.R188S Chr.5: 80665346-80665346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569679162 Chr.5: 80665318-80665318 on Build GRCh38
rs748721462 Chr.5: 80665214-80665214 on Build GRCh38
rs750311213 Chr.5: 80665301-80665301 on Build GRCh38
rs779648151 Chr.5: 80665306-80665306 on Build GRCh38
rs370894909 Chr.5: 80665193-80665193 on Build GRCh38
rs572193350 Chr.5: 80665347-80665347 on Build GRCh38
rs374128221 Chr.5: 80665170-80665170 on Build GRCh38
rs776922419 Chr.5: 80665258-80665258 on Build GRCh38
rs374133543 Chr.5: 80665358-80665358 on Build GRCh38
rs897270856 Chr.5: 80665378-80665378 on Build GRCh38
rs6151614 Chr.5: 80665516-80665516 on Build GRCh38
rs1033973823 Chr.5: 80665430-80665430 on Build GRCh38
rs143733332 Chr.5: 80665205-80665205 on Build GRCh38
rs928035794 Chr.5: 80665496-80665496 on Build GRCh38
rs757991599 Chr.5: 80665406-80665406 on Build GRCh38
rs966254697 Chr.5: 80665215-80665215 on Build GRCh38
rs977366961 Chr.5: 80665223-80665223 on Build GRCh38
rs771370664 Chr.5: 80665244-80665244 on Build GRCh38
rs773751711 Chr.5: 80665228-80665228 on Build GRCh38
rs144012714 Chr.5: 80665338-80665338 on Build GRCh38
rs888626337 Chr.5: 80665355-80665355 on Build GRCh38
rs775653573 Chr.5: 80665333-80665333 on Build GRCh38
rs765418313 Chr.5: 80665268-80665268 on Build GRCh38
rs71603559 Chr.5: 80665469-80665470 on Build GRCh38
rs757177212 Chr.5: 80665164-80665164 on Build GRCh38
rs924517772 Chr.5: 80665230-80665230 on Build GRCh38
rs778153497 Chr.5: 80665622-80665622 on Build GRCh38
rs55750137 Chr.5: 80665626-80665626 on Build GRCh38
rs541207361 Chr.5: 80665407-80665407 on Build GRCh38
rs1824838 Chr.5: 80665493-80665493 on Build GRCh38
rs6151613 Chr.5: 80665470-80665470 on Build GRCh38
rs941577931 Chr.5: 80665570-80665570 on Build GRCh38
rs372828270 Chr.5: 80665421-80665421 on Build GRCh38
rs957233749 Chr.5: 80665260-80665260 on Build GRCh38
rs913692200 Chr.5: 80665221-80665221 on Build GRCh38
rs1015764579 Chr.5: 80665377-80665377 on Build GRCh38
rs1824837 Chr.5: 80665422-80665422 on Build GRCh38
rs6151612 Chr.5: 80665469-80665469 on Build GRCh38
rs778461068 Chr.5: 80665396-80665396 on Build GRCh38
rs778443399 Chr.5: 80665314-80665314 on Build GRCh38
rs1024325700 Chr.5: 80665419-80665419 on Build GRCh38
rs752700097 Chr.5: 80665275-80665275 on Build GRCh38
rs6151616 Chr.5: 80665651-80665651 on Build GRCh38
rs750366331 Chr.5: 80665341-80665341 on Build GRCh38
rs41562817 Chr.5: 80665593-80665593 on Build GRCh38
rs989487112 Chr.5: 80665263-80665263 on Build GRCh38
rs932272400 Chr.5: 80665504-80665504 on Build GRCh38
rs976443124 Chr.5: 80665486-80665486 on Build GRCh38
rs957285874 Chr.5: 80665450-80665450 on Build GRCh38
rs746425286 Chr.5: 80665337-80665337 on Build GRCh38
rs754571381 Chr.5: 80665309-80665309 on Build GRCh38
rs919399229 Chr.5: 80665569-80665569 on Build GRCh38
rs1031525397 Chr.5: 80665650-80665650 on Build GRCh38
rs138381683 Chr.5: 80665211-80665211 on Build GRCh38
rs762556920 Chr.5: 80665404-80665404 on Build GRCh38
rs755863687 Chr.5: 80665203-80665203 on Build GRCh38
rs140749169 Chr.5: 80665324-80665324 on Build GRCh38
rs953165915 Chr.5: 80665192-80665192 on Build GRCh38
rs767478058 Chr.5: 80665296-80665296 on Build GRCh38
rs375095092 Chr.5: 80665222-80665222 on Build GRCh38
rs773797934 Chr.5: 80665481-80665481 on Build GRCh38
rs1804661 Chr.5: 80665165-80665165 on Build GRCh38
rs1824839 Chr.5: 80665543-80665543 on Build GRCh38
rs371881555 Chr.5: 80665374-80665374 on Build GRCh38
rs769878887 Chr.5: 80665329-80665329 on Build GRCh38
rs753594929 Chr.5: 80665308-80665308 on Build GRCh38
rs969779948 Chr.5: 80665484-80665484 on Build GRCh38
rs771284065 Chr.5: 80665218-80665218 on Build GRCh38
rs745553087 Chr.5: 80665246-80665247 on Build GRCh38
rs901711895 Chr.5: 80665367-80665367 on Build GRCh38
rs761147959 Chr.5: 80665242-80665242 on Build GRCh38
rs772683764 Chr.5: 80665226-80665226 on Build GRCh38
rs748835474 Chr.5: 80665210-80665210 on Build GRCh38
rs957977998 Chr.5: 80665431-80665431 on Build GRCh38
rs570665839 Chr.5: 80665635-80665635 on Build GRCh38
rs753715694 Chr.5: 80665608-80665608 on Build GRCh38
rs768134044 Chr.5: 80665279-80665279 on Build GRCh38
rs771327705 Chr.5: 80665320-80665320 on Build GRCh38
rs998574337 Chr.5: 80665640-80665640 on Build GRCh38
rs759608517 Chr.5: 80665265-80665265 on Build GRCh38
rs372073889 Chr.5: 80665313-80665313 on Build GRCh38
rs755916911 Chr.5: 80665305-80665305 on Build GRCh38
rs6151615 Chr.5: 80665613-80665613 on Build GRCh38
rs747583051 Chr.5: 80665408-80665408 on Build GRCh38
rs147289289 Chr.5: 80665346-80665346 on Build GRCh38
rs41553917 Chr.5: 80665571-80665571 on Build GRCh38
rs867080501 Chr.5: 80665266-80665266 on Build GRCh38
rs368468467 Chr.5: 80665312-80665312 on Build GRCh38
rs760604405 Chr.5: 80665342-80665342 on Build GRCh38

View Full Product Details