Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81312779-81313282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
UniGene
Hs.591756
Gene Chromosome Location
Chr.5: 81309045-81394174 on Build GRCh38
Transcripts
XM_017009045.1 XM_017009046.1 XM_017009047.1 XM_017009048.1 NM_001131035.1 NM_001131036.1 NM_032280.2 XM_005248616.3

Gene Symbol
ZCCHC9
Entrez Gene ID
84240
Gene Name
zinc finger CCHC-type containing 9
Gene Aliases
PPP1R41
UniGene
Hs.15536
Gene Chromosome Location
Chr.5: 81301583-81313147 on Build GRCh38
Transcripts
XM_017009045.1 XM_017009046.1 XM_017009047.1 XM_017009048.1 NM_001131035.1 NM_001131036.1 NM_032280.2 XM_005248616.3

SNP Detail

SNP ID Chromosome Location
rs1003186465 Chr.5: 81313139-81313139 on Build GRCh38
rs574231814 Chr.5: 81312818-81312818 on Build GRCh38
rs959294241 Chr.5: 81312856-81312856 on Build GRCh38
rs1017735216 Chr.5: 81312943-81312943 on Build GRCh38
rs899323649 Chr.5: 81313068-81313068 on Build GRCh38
rs978864244 Chr.5: 81312879-81312879 on Build GRCh38
rs114326982 Chr.5: 81313113-81313113 on Build GRCh38
rs1034906108 Chr.5: 81312842-81312842 on Build GRCh38
rs888078246 Chr.5: 81313247-81313247 on Build GRCh38
rs181432063 Chr.5: 81313098-81313098 on Build GRCh38
rs550676274 Chr.5: 81313112-81313112 on Build GRCh38
rs533391896 Chr.5: 81312953-81312953 on Build GRCh38
rs988839812 Chr.5: 81313027-81313027 on Build GRCh38
rs146249074 Chr.5: 81312814-81312814 on Build GRCh38
rs912124075 Chr.5: 81312993-81312993 on Build GRCh38
rs16878596 Chr.5: 81313014-81313014 on Build GRCh38
rs947462773 Chr.5: 81313083-81313083 on Build GRCh38
rs1039146777 Chr.5: 81313032-81313034 on Build GRCh38
rs897084905 Chr.5: 81313184-81313184 on Build GRCh38
rs1057355708 Chr.5: 81313160-81313160 on Build GRCh38
rs980218900 Chr.5: 81313093-81313093 on Build GRCh38
rs967886778 Chr.5: 81313228-81313228 on Build GRCh38
rs938519239 Chr.5: 81313143-81313143 on Build GRCh38
rs74333593 Chr.5: 81313217-81313217 on Build GRCh38
rs990613250 Chr.5: 81312862-81312862 on Build GRCh38
rs756698001 Chr.5: 81312886-81312886 on Build GRCh38
rs976218790 Chr.5: 81312959-81312959 on Build GRCh38
rs375630574 Chr.5: 81313157-81313157 on Build GRCh38
rs886217691 Chr.5: 81313122-81313123 on Build GRCh38
rs1034686370 Chr.5: 81313159-81313159 on Build GRCh38
rs924711826 Chr.5: 81312929-81312929 on Build GRCh38
rs559512543 Chr.5: 81312881-81312881 on Build GRCh38
rs113758347 Chr.5: 81313079-81313079 on Build GRCh38
rs915086905 Chr.5: 81312869-81312869 on Build GRCh38
rs768452697 Chr.5: 81313136-81313139 on Build GRCh38
rs186099753 Chr.5: 81312877-81312877 on Build GRCh38
rs565157474 Chr.5: 81313071-81313071 on Build GRCh38
rs747254161 Chr.5: 81313223-81313223 on Build GRCh38
rs758615777 Chr.5: 81312990-81312990 on Build GRCh38
rs1048061169 Chr.5: 81313225-81313225 on Build GRCh38
rs528572970 Chr.5: 81312962-81312962 on Build GRCh38

View Full Product Details