Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 94153750-94154290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0825
Entrez Gene ID
285600
Gene Name
KIAA0825
Gene Aliases
C5orf36
UniGene
Hs.425123
Gene Chromosome Location
Chr.5: 94150851-94618604 on Build GRCh38
Transcripts
NM_001145678.1 XM_006714588.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159631 c.3767A>G p.H1256R Chr.5: 94154068-94154068 on Build GRCh38
1071260 c.3800C>A p.S1267Y Chr.5: 94154035-94154035 on Build GRCh38
6323892 c.3713G>A p.W1238* Chr.5: 94154122-94154122 on Build GRCh38
5733295 c.3806C>T p.S1269L Chr.5: 94154029-94154029 on Build GRCh38
3141259 c.3742G>A p.E1248K Chr.5: 94154093-94154093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777435686 Chr.5: 94154032-94154032 on Build GRCh38
rs370604950 Chr.5: 94153786-94153786 on Build GRCh38
rs536591394 Chr.5: 94153829-94153829 on Build GRCh38
rs919551260 Chr.5: 94154075-94154075 on Build GRCh38
rs576527157 Chr.5: 94154257-94154257 on Build GRCh38
rs972907750 Chr.5: 94153802-94153802 on Build GRCh38
rs62367163 Chr.5: 94153906-94153906 on Build GRCh38
rs912287284 Chr.5: 94154116-94154116 on Build GRCh38
rs189655650 Chr.5: 94154258-94154258 on Build GRCh38
rs552595797 Chr.5: 94153812-94153812 on Build GRCh38
rs570116230 Chr.5: 94153778-94153778 on Build GRCh38
rs879500658 Chr.5: 94154195-94154195 on Build GRCh38
rs373813457 Chr.5: 94153996-94153996 on Build GRCh38
rs913072575 Chr.5: 94153943-94153943 on Build GRCh38
rs150081194 Chr.5: 94153792-94153792 on Build GRCh38
rs993914669 Chr.5: 94154177-94154177 on Build GRCh38
rs992716967 Chr.5: 94153980-94153980 on Build GRCh38
rs574951491 Chr.5: 94154021-94154021 on Build GRCh38
rs773456189 Chr.5: 94154143-94154143 on Build GRCh38
rs889999748 Chr.5: 94154117-94154117 on Build GRCh38
rs960933199 Chr.5: 94153968-94153968 on Build GRCh38
rs566409812 Chr.5: 94153844-94153844 on Build GRCh38
rs919590967 Chr.5: 94153834-94153834 on Build GRCh38
rs1019752610 Chr.5: 94154146-94154146 on Build GRCh38
rs537612345 Chr.5: 94154086-94154086 on Build GRCh38
rs780374581 Chr.5: 94153925-94153925 on Build GRCh38
rs201666365 Chr.5: 94154068-94154068 on Build GRCh38
rs573275691 Chr.5: 94153929-94153929 on Build GRCh38
rs936710682 Chr.5: 94154043-94154043 on Build GRCh38
rs899763845 Chr.5: 94154247-94154247 on Build GRCh38
rs867019172 Chr.5: 94154040-94154040 on Build GRCh38
rs776946092 Chr.5: 94154179-94154179 on Build GRCh38
rs199863477 Chr.5: 94154113-94154113 on Build GRCh38
rs982588678 Chr.5: 94154099-94154099 on Build GRCh38
rs1042893506 Chr.5: 94154010-94154010 on Build GRCh38
rs952349176 Chr.5: 94154194-94154194 on Build GRCh38
rs980012947 Chr.5: 94153851-94153851 on Build GRCh38
rs945450232 Chr.5: 94153960-94153960 on Build GRCh38
rs769648904 Chr.5: 94154072-94154072 on Build GRCh38
rs761410453 Chr.5: 94154201-94154201 on Build GRCh38
rs772191114 Chr.5: 94154098-94154098 on Build GRCh38
rs199584476 Chr.5: 94154041-94154041 on Build GRCh38
rs979730362 Chr.5: 94154237-94154237 on Build GRCh38
rs748174533 Chr.5: 94154087-94154087 on Build GRCh38
rs556641522 Chr.5: 94154138-94154138 on Build GRCh38
rs916847905 Chr.5: 94153992-94153992 on Build GRCh38
rs769210988 Chr.5: 94154093-94154093 on Build GRCh38
rs769148272 Chr.5: 94154239-94154239 on Build GRCh38
rs760973667 Chr.5: 94154159-94154159 on Build GRCh38
rs927228756 Chr.5: 94153864-94153864 on Build GRCh38
rs113082362 Chr.5: 94153931-94153931 on Build GRCh38
rs376383241 Chr.5: 94154069-94154069 on Build GRCh38
rs776804130 Chr.5: 94154173-94154173 on Build GRCh38
rs771317388 Chr.5: 94154169-94154169 on Build GRCh38
rs897345107 Chr.5: 94154176-94154176 on Build GRCh38
rs185185520 Chr.5: 94153913-94153913 on Build GRCh38
rs745688608 Chr.5: 94154071-94154071 on Build GRCh38

View Full Product Details