Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 94391442-94391982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0825
Entrez Gene ID
285600
Gene Name
KIAA0825
Gene Aliases
C5orf36
UniGene
Hs.425123
Gene Chromosome Location
Chr.5: 94150851-94618604 on Build GRCh38
Transcripts
NM_001145678.1 XM_006714588.2 XM_011543324.2 XM_011543325.2 XM_011543327.2 XM_011543328.2 XM_011543329.2 XM_011543330.2 XM_011543331.1 XM_011543332.1 XM_011543333.2 XM_011543334.2 XM_011543335.2 XM_017009373.1 XM_017009374.1 XM_017009375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006078 c.3315G>C p.E1105D Chr.5: 94391676-94391676 on Build GRCh38
3141270 c.3349C>G p.L1117V Chr.5: 94391642-94391642 on Build GRCh38
3357922 c.3378G>A p.T1126T Chr.5: 94391613-94391613 on Build GRCh38
1071263 c.3334G>A p.E1112K Chr.5: 94391657-94391657 on Build GRCh38
1487043 c.3338_3340delGAG p.G1113delG Chr.5: 94391651-94391653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75845455 Chr.5: 94391513-94391513 on Build GRCh38
rs886921096 Chr.5: 94391844-94391844 on Build GRCh38
rs186808582 Chr.5: 94391875-94391875 on Build GRCh38
rs975427890 Chr.5: 94391572-94391572 on Build GRCh38
rs866738196 Chr.5: 94391472-94391472 on Build GRCh38
rs1019223330 Chr.5: 94391921-94391921 on Build GRCh38
rs200795815 Chr.5: 94391516-94391516 on Build GRCh38
rs746157612 Chr.5: 94391610-94391610 on Build GRCh38
rs767092689 Chr.5: 94391680-94391680 on Build GRCh38
rs779618038 Chr.5: 94391514-94391515 on Build GRCh38
rs924746903 Chr.5: 94391643-94391643 on Build GRCh38
rs1043305120 Chr.5: 94391634-94391634 on Build GRCh38
rs753765837 Chr.5: 94391497-94391497 on Build GRCh38
rs1039346733 Chr.5: 94391769-94391769 on Build GRCh38
rs751063650 Chr.5: 94391938-94391938 on Build GRCh38
rs970931815 Chr.5: 94391654-94391654 on Build GRCh38
rs867415084 Chr.5: 94391644-94391644 on Build GRCh38
rs574293739 Chr.5: 94391525-94391525 on Build GRCh38
rs1006848726 Chr.5: 94391481-94391481 on Build GRCh38
rs75413171 Chr.5: 94391539-94391539 on Build GRCh38
rs527480661 Chr.5: 94391842-94391842 on Build GRCh38
rs758037063 Chr.5: 94391750-94391750 on Build GRCh38
rs566961516 Chr.5: 94391816-94391816 on Build GRCh38
rs769000684 Chr.5: 94391667-94391667 on Build GRCh38
rs765753863 Chr.5: 94391613-94391613 on Build GRCh38
rs375980456 Chr.5: 94391805-94391805 on Build GRCh38
rs562568543 Chr.5: 94391612-94391612 on Build GRCh38
rs112295170 Chr.5: 94391498-94391498 on Build GRCh38
rs554442302 Chr.5: 94391524-94391524 on Build GRCh38
rs578131882 Chr.5: 94391480-94391480 on Build GRCh38
rs753614711 Chr.5: 94391804-94391804 on Build GRCh38
rs994727743 Chr.5: 94391819-94391819 on Build GRCh38
rs1023565318 Chr.5: 94391942-94391942 on Build GRCh38
rs950651198 Chr.5: 94391831-94391831 on Build GRCh38
rs1005005442 Chr.5: 94391845-94391845 on Build GRCh38
rs1044048096 Chr.5: 94391768-94391768 on Build GRCh38
rs1016752210 Chr.5: 94391907-94391907 on Build GRCh38
rs560192446 Chr.5: 94391711-94391711 on Build GRCh38
rs916834905 Chr.5: 94391653-94391653 on Build GRCh38
rs369988324 Chr.5: 94391614-94391614 on Build GRCh38
rs935119954 Chr.5: 94391671-94391671 on Build GRCh38
rs114483743 Chr.5: 94391733-94391733 on Build GRCh38
rs754818484 Chr.5: 94391506-94391506 on Build GRCh38
rs747020377 Chr.5: 94391515-94391515 on Build GRCh38
rs766037879 Chr.5: 94391720-94391720 on Build GRCh38
rs983928919 Chr.5: 94391548-94391548 on Build GRCh38
rs963459676 Chr.5: 94391928-94391928 on Build GRCh38
rs998073020 Chr.5: 94391817-94391817 on Build GRCh38
rs991063911 Chr.5: 94391617-94391617 on Build GRCh38
rs1046959981 Chr.5: 94391826-94391826 on Build GRCh38
rs753568429 Chr.5: 94391724-94391724 on Build GRCh38
rs1013108524 Chr.5: 94391747-94391747 on Build GRCh38
rs561223193 Chr.5: 94391922-94391922 on Build GRCh38
rs944154220 Chr.5: 94391567-94391567 on Build GRCh38
rs1016287753 Chr.5: 94391553-94391553 on Build GRCh38
rs750070509 Chr.5: 94391700-94391700 on Build GRCh38
rs373480280 Chr.5: 94391620-94391620 on Build GRCh38
rs772565831 Chr.5: 94391668-94391669 on Build GRCh38
rs377748930 Chr.5: 94391668-94391668 on Build GRCh38

View Full Product Details