Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 110738916-110739414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A46
Entrez Gene ID
91137
Gene Name
solute carrier family 25 member 46
Gene Aliases
HMSN6B
UniGene
Hs.75639
Gene Chromosome Location
Chr.5: 110738136-110765161 on Build GRCh38
Transcripts
NM_001303249.1 NM_001303250.1 NM_138773.2 XM_006714670.3 XM_011543552.2 XM_011543555.2 XM_011543557.2 XM_011543559.2 XM_011543561.2 XM_011543563.2 XM_011543564.2 XM_011543565.2 XM_011543566.2 XM_011543567.2 XM_017009705.1 XM_017009706.1 XM_017009707.1

Gene Symbol
TMEM232
Entrez Gene ID
642987
Gene Name
transmembrane protein 232
Gene Aliases
-
UniGene
Hs.729154
Gene Chromosome Location
Chr.5: 110387255-110738936 on Build GRCh38
Transcripts
NM_001303249.1 NM_001303250.1 NM_138773.2 XM_006714670.3 XM_011543552.2 XM_011543555.2 XM_011543557.2 XM_011543559.2 XM_011543561.2 XM_011543563.2 XM_011543564.2 XM_011543565.2 XM_011543566.2 XM_011543567.2 XM_017009705.1 XM_017009706.1 XM_017009707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669358 c.147C>T p.I49I Chr.5: 110739266-110739266 on Build GRCh38
7315655 c.28G>C p.D10H Chr.5: 110739147-110739147 on Build GRCh38
5502991 c.67G>T p.G23C Chr.5: 110739186-110739186 on Build GRCh38
7390297 c.28G>A p.D10N Chr.5: 110739147-110739147 on Build GRCh38
1059448 c.216G>A p.P72P Chr.5: 110739335-110739335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149199100 Chr.5: 110739062-110739062 on Build GRCh38
rs536654643 Chr.5: 110738939-110738939 on Build GRCh38
rs373558070 Chr.5: 110739031-110739031 on Build GRCh38
rs773343482 Chr.5: 110739358-110739358 on Build GRCh38
rs527254972 Chr.5: 110739040-110739040 on Build GRCh38
rs910693612 Chr.5: 110739127-110739127 on Build GRCh38
rs145119144 Chr.5: 110739207-110739207 on Build GRCh38
rs752451949 Chr.5: 110739257-110739257 on Build GRCh38
rs758140632 Chr.5: 110739315-110739315 on Build GRCh38
rs1006234534 Chr.5: 110738952-110738952 on Build GRCh38
rs1035573966 Chr.5: 110739016-110739016 on Build GRCh38
rs760129269 Chr.5: 110739387-110739387 on Build GRCh38
rs369936836 Chr.5: 110739162-110739162 on Build GRCh38
rs767703868 Chr.5: 110739215-110739215 on Build GRCh38
rs868536854 Chr.5: 110739259-110739259 on Build GRCh38
rs532755024 Chr.5: 110739211-110739211 on Build GRCh38
rs576455970 Chr.5: 110738957-110738957 on Build GRCh38
rs748373062 Chr.5: 110739282-110739282 on Build GRCh38
rs145421520 Chr.5: 110739266-110739266 on Build GRCh38
rs868425088 Chr.5: 110738946-110738946 on Build GRCh38
rs960197684 Chr.5: 110739142-110739142 on Build GRCh38
rs919490546 Chr.5: 110739261-110739261 on Build GRCh38
rs892374338 Chr.5: 110739366-110739366 on Build GRCh38
rs754258035 Chr.5: 110739234-110739234 on Build GRCh38
rs577796065 Chr.5: 110738996-110738996 on Build GRCh38
rs759961883 Chr.5: 110739298-110739298 on Build GRCh38
rs759814821 Chr.5: 110739066-110739066 on Build GRCh38
rs904962071 Chr.5: 110739328-110739328 on Build GRCh38
rs747086911 Chr.5: 110739360-110739360 on Build GRCh38
rs754702528 Chr.5: 110739115-110739115 on Build GRCh38
rs956676307 Chr.5: 110739094-110739094 on Build GRCh38
rs888191453 Chr.5: 110739068-110739068 on Build GRCh38
rs1041040640 Chr.5: 110739061-110739061 on Build GRCh38
rs373730487 Chr.5: 110739037-110739037 on Build GRCh38
rs761232447 Chr.5: 110739317-110739317 on Build GRCh38
rs988132072 Chr.5: 110739104-110739104 on Build GRCh38
rs924331595 Chr.5: 110739192-110739192 on Build GRCh38
rs531287378 Chr.5: 110739083-110739083 on Build GRCh38
rs993458791 Chr.5: 110739128-110739128 on Build GRCh38
rs781167879 Chr.5: 110739333-110739333 on Build GRCh38
rs778727882 Chr.5: 110739057-110739057 on Build GRCh38
rs751659870 Chr.5: 110739071-110739071 on Build GRCh38
rs200566665 Chr.5: 110739354-110739354 on Build GRCh38
rs147648476 Chr.5: 110739267-110739267 on Build GRCh38
rs542221845 Chr.5: 110739049-110739049 on Build GRCh38
rs868222085 Chr.5: 110739318-110739318 on Build GRCh38
rs543936823 Chr.5: 110738978-110738978 on Build GRCh38
rs377179458 Chr.5: 110739091-110739091 on Build GRCh38
rs191332512 Chr.5: 110739171-110739171 on Build GRCh38
rs1022875973 Chr.5: 110739343-110739343 on Build GRCh38
rs748319527 Chr.5: 110739355-110739355 on Build GRCh38
rs745863416 Chr.5: 110739337-110739337 on Build GRCh38
rs934318056 Chr.5: 110739067-110739067 on Build GRCh38
rs747634003 Chr.5: 110739141-110739141 on Build GRCh38
rs929613387 Chr.5: 110739063-110739063 on Build GRCh38
rs369381954 Chr.5: 110739250-110739250 on Build GRCh38
rs745632467 Chr.5: 110739271-110739271 on Build GRCh38
rs140606969 Chr.5: 110739287-110739287 on Build GRCh38
rs769538816 Chr.5: 110739021-110739021 on Build GRCh38
rs753376360 Chr.5: 110739114-110739114 on Build GRCh38
rs769626359 Chr.5: 110739275-110739275 on Build GRCh38
rs1022269942 Chr.5: 110739389-110739389 on Build GRCh38
rs770228533 Chr.5: 110739392-110739392 on Build GRCh38
rs867367132 Chr.5: 110738970-110738970 on Build GRCh38
rs889175123 Chr.5: 110738951-110738951 on Build GRCh38
rs778529430 Chr.5: 110739132-110739132 on Build GRCh38
rs768895732 Chr.5: 110739206-110739206 on Build GRCh38
rs946817134 Chr.5: 110739027-110739027 on Build GRCh38
rs1057519416 Chr.5: 110739285-110739286 on Build GRCh38
rs536991478 Chr.5: 110739270-110739270 on Build GRCh38
rs755840361 Chr.5: 110739338-110739338 on Build GRCh38
rs777324317 Chr.5: 110739296-110739296 on Build GRCh38
rs748696438 Chr.5: 110739074-110739074 on Build GRCh38
rs1022344910 Chr.5: 110739106-110739106 on Build GRCh38
rs867453277 Chr.5: 110738998-110738998 on Build GRCh38
rs761045034 Chr.5: 110739288-110739288 on Build GRCh38
rs560033851 Chr.5: 110739038-110739038 on Build GRCh38
rs777269362 Chr.5: 110739384-110739384 on Build GRCh38
rs770235761 Chr.5: 110739081-110739081 on Build GRCh38
rs771987009 Chr.5: 110739285-110739285 on Build GRCh38
rs867841573 Chr.5: 110739277-110739277 on Build GRCh38
rs763466158 Chr.5: 110739322-110739322 on Build GRCh38
rs750996882 Chr.5: 110739078-110739078 on Build GRCh38
rs562528566 Chr.5: 110738990-110738990 on Build GRCh38
rs897837735 Chr.5: 110739119-110739119 on Build GRCh38
rs772367949 Chr.5: 110739357-110739357 on Build GRCh38
rs554993391 Chr.5: 110739305-110739305 on Build GRCh38
rs761259380 Chr.5: 110739225-110739225 on Build GRCh38
rs889277279 Chr.5: 110739098-110739098 on Build GRCh38
rs978518059 Chr.5: 110739180-110739180 on Build GRCh38
rs868711975 Chr.5: 110739208-110739208 on Build GRCh38
rs766876006 Chr.5: 110739292-110739292 on Build GRCh38
rs865849109 Chr.5: 110739274-110739274 on Build GRCh38
rs370525588 Chr.5: 110739019-110739019 on Build GRCh38
rs757981265 Chr.5: 110739260-110739260 on Build GRCh38
rs17132319 Chr.5: 110738976-110738976 on Build GRCh38
rs1051001816 Chr.5: 110739095-110739095 on Build GRCh38
rs933521608 Chr.5: 110739092-110739092 on Build GRCh38
rs867789049 Chr.5: 110739198-110739198 on Build GRCh38
rs955762085 Chr.5: 110739380-110739380 on Build GRCh38
rs13186465 Chr.5: 110739043-110739043 on Build GRCh38
rs763111033 Chr.5: 110739173-110739173 on Build GRCh38
rs773525488 Chr.5: 110739217-110739217 on Build GRCh38
rs377196064 Chr.5: 110739323-110739323 on Build GRCh38
rs762266931 Chr.5: 110739072-110739072 on Build GRCh38
rs1037125606 Chr.5: 110739087-110739088 on Build GRCh38
rs372780604 Chr.5: 110739279-110739279 on Build GRCh38
rs968966762 Chr.5: 110739349-110739349 on Build GRCh38
rs186912891 Chr.5: 110738943-110738943 on Build GRCh38
rs770969480 Chr.5: 110739370-110739370 on Build GRCh38
rs867218123 Chr.5: 110739334-110739334 on Build GRCh38
rs770562376 Chr.5: 110739165-110739165 on Build GRCh38
rs771761288 Chr.5: 110739161-110739161 on Build GRCh38
rs976459195 Chr.5: 110739383-110739383 on Build GRCh38
rs139629514 Chr.5: 110739263-110739263 on Build GRCh38
rs764957322 Chr.5: 110739256-110739256 on Build GRCh38
rs982442142 Chr.5: 110738958-110738958 on Build GRCh38
rs756759699 Chr.5: 110739332-110739332 on Build GRCh38
rs372382932 Chr.5: 110739163-110739163 on Build GRCh38
rs755431213 Chr.5: 110739255-110739255 on Build GRCh38

View Full Product Details