Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132753179-132753681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNI2
Entrez Gene ID
645121
Gene Name
cyclin I family member 2
Gene Aliases
-
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132747402-132754403 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132750817-132778216 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

SNP Detail

SNP ID Chromosome Location
rs184411228 Chr.5: 132753535-132753535 on Build GRCh38
rs888892336 Chr.5: 132753280-132753280 on Build GRCh38
rs922977881 Chr.5: 132753224-132753224 on Build GRCh38
rs544202961 Chr.5: 132753283-132753283 on Build GRCh38
rs1004392058 Chr.5: 132753230-132753230 on Build GRCh38
rs933040360 Chr.5: 132753236-132753236 on Build GRCh38
rs192784762 Chr.5: 132753534-132753534 on Build GRCh38
rs1033290081 Chr.5: 132753436-132753436 on Build GRCh38
rs907184157 Chr.5: 132753366-132753366 on Build GRCh38
rs985792148 Chr.5: 132753527-132753527 on Build GRCh38
rs76476505 Chr.5: 132753287-132753287 on Build GRCh38
rs978695785 Chr.5: 132753376-132753376 on Build GRCh38
rs976785882 Chr.5: 132753511-132753511 on Build GRCh38
rs747717922 Chr.5: 132753441-132753441 on Build GRCh38
rs187532554 Chr.5: 132753476-132753476 on Build GRCh38
rs1026036528 Chr.5: 132753310-132753310 on Build GRCh38
rs1048620517 Chr.5: 132753541-132753541 on Build GRCh38
rs57012085 Chr.5: 132753508-132753508 on Build GRCh38
rs906477093 Chr.5: 132753621-132753621 on Build GRCh38
rs1046299478 Chr.5: 132753604-132753604 on Build GRCh38
rs1002903795 Chr.5: 132753371-132753371 on Build GRCh38
rs755142933 Chr.5: 132753546-132753546 on Build GRCh38
rs1002872515 Chr.5: 132753628-132753628 on Build GRCh38
rs56998927 Chr.5: 132753263-132753263 on Build GRCh38
rs977721870 Chr.5: 132753481-132753481 on Build GRCh38
rs1015361299 Chr.5: 132753246-132753246 on Build GRCh38
rs894531087 Chr.5: 132753655-132753655 on Build GRCh38
rs1022547914 Chr.5: 132753295-132753295 on Build GRCh38
rs1037129366 Chr.5: 132753565-132753565 on Build GRCh38
rs993435069 Chr.5: 132753309-132753309 on Build GRCh38
rs934817431 Chr.5: 132753466-132753466 on Build GRCh38
rs969989085 Chr.5: 132753355-132753355 on Build GRCh38
rs530150177 Chr.5: 132753363-132753363 on Build GRCh38
rs756159119 Chr.5: 132753646-132753649 on Build GRCh38
rs1044077486 Chr.5: 132753630-132753630 on Build GRCh38
rs992614934 Chr.5: 132753523-132753523 on Build GRCh38
rs910100645 Chr.5: 132753544-132753544 on Build GRCh38
rs73787059 Chr.5: 132753584-132753584 on Build GRCh38
rs1034426564 Chr.5: 132753411-132753411 on Build GRCh38
rs748112454 Chr.5: 132753564-132753564 on Build GRCh38
rs185878684 Chr.5: 132753260-132753260 on Build GRCh38
rs923540815 Chr.5: 132753506-132753506 on Build GRCh38
rs1034356147 Chr.5: 132753636-132753636 on Build GRCh38
rs1037463053 Chr.5: 132753550-132753550 on Build GRCh38
rs968007614 Chr.5: 132753478-132753478 on Build GRCh38
rs990280896 Chr.5: 132753444-132753444 on Build GRCh38
rs941857635 Chr.5: 132753282-132753282 on Build GRCh38
rs778950355 Chr.5: 132753552-132753552 on Build GRCh38

View Full Product Details