Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 136056334-136056848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFBI
Entrez Gene ID
7045
Gene Name
transforming growth factor beta induced
Gene Aliases
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
UniGene
Hs.369397
Gene Chromosome Location
Chr.5: 136028895-136063818 on Build GRCh38
Transcripts
NM_000358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
303372 c.1598G>T p.R533L Chr.5: 136056715-136056715 on Build GRCh38
4895992 c.1665G>A p.R555R Chr.5: 136056782-136056782 on Build GRCh38
5402147 c.1633G>A p.E545K Chr.5: 136056750-136056750 on Build GRCh38
5402145 c.1625C>T p.P542L Chr.5: 136056742-136056742 on Build GRCh38
222470 c.1585G>A p.E529K Chr.5: 136056702-136056702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772680357 Chr.5: 136056651-136056651 on Build GRCh38
rs4669 Chr.5: 136056737-136056737 on Build GRCh38
rs760908139 Chr.5: 136056800-136056800 on Build GRCh38
rs541615265 Chr.5: 136056408-136056408 on Build GRCh38
rs770235053 Chr.5: 136056653-136056653 on Build GRCh38
rs747785814 Chr.5: 136056642-136056642 on Build GRCh38
rs201518300 Chr.5: 136056817-136056817 on Build GRCh38
rs746890042 Chr.5: 136056815-136056815 on Build GRCh38
rs764361610 Chr.5: 136056683-136056683 on Build GRCh38
rs767742299 Chr.5: 136056703-136056703 on Build GRCh38
rs2074810 Chr.5: 136056413-136056413 on Build GRCh38
rs746455784 Chr.5: 136056750-136056750 on Build GRCh38
rs906782180 Chr.5: 136056359-136056359 on Build GRCh38
rs777288957 Chr.5: 136056748-136056748 on Build GRCh38
rs771620738 Chr.5: 136056647-136056647 on Build GRCh38
rs991002137 Chr.5: 136056773-136056773 on Build GRCh38
rs1010679779 Chr.5: 136056550-136056550 on Build GRCh38
rs762155321 Chr.5: 136056785-136056785 on Build GRCh38
rs267607109 Chr.5: 136056754-136056754 on Build GRCh38
rs774822557 Chr.5: 136056774-136056774 on Build GRCh38
rs554888573 Chr.5: 136056794-136056794 on Build GRCh38
rs200202768 Chr.5: 136056810-136056810 on Build GRCh38
rs775730175 Chr.5: 136056658-136056658 on Build GRCh38
rs1018425770 Chr.5: 136056601-136056601 on Build GRCh38
rs200219644 Chr.5: 136056793-136056793 on Build GRCh38
rs747839006 Chr.5: 136056739-136056739 on Build GRCh38
rs121909208 Chr.5: 136056780-136056780 on Build GRCh38
rs991426617 Chr.5: 136056602-136056602 on Build GRCh38
rs796996840 Chr.5: 136056640-136056640 on Build GRCh38
rs1050842080 Chr.5: 136056697-136056697 on Build GRCh38
rs916824993 Chr.5: 136056605-136056605 on Build GRCh38
rs530375415 Chr.5: 136056478-136056478 on Build GRCh38
rs548453576 Chr.5: 136056610-136056610 on Build GRCh38
rs1031183804 Chr.5: 136056505-136056505 on Build GRCh38
rs534215738 Chr.5: 136056512-136056512 on Build GRCh38
rs771674035 Chr.5: 136056747-136056747 on Build GRCh38
rs530953796 Chr.5: 136056700-136056700 on Build GRCh38
rs121909214 Chr.5: 136056736-136056736 on Build GRCh38
rs565005341 Chr.5: 136056392-136056392 on Build GRCh38
rs982631735 Chr.5: 136056726-136056726 on Build GRCh38
rs770204995 Chr.5: 136056755-136056755 on Build GRCh38
rs185285049 Chr.5: 136056396-136056396 on Build GRCh38
rs778550933 Chr.5: 136056738-136056738 on Build GRCh38
rs372357214 Chr.5: 136056804-136056804 on Build GRCh38
rs570927940 Chr.5: 136056708-136056708 on Build GRCh38
rs1007153647 Chr.5: 136056375-136056375 on Build GRCh38
rs778698555 Chr.5: 136056641-136056641 on Build GRCh38
rs267607110 Chr.5: 136056769-136056769 on Build GRCh38
rs769108952 Chr.5: 136056772-136056772 on Build GRCh38
rs960889420 Chr.5: 136056593-136056593 on Build GRCh38
rs769946729 Chr.5: 136056666-136056666 on Build GRCh38
rs776665133 Chr.5: 136056575-136056575 on Build GRCh38
rs942535467 Chr.5: 136056704-136056704 on Build GRCh38
rs1018171635 Chr.5: 136056383-136056383 on Build GRCh38
rs763310312 Chr.5: 136056669-136056669 on Build GRCh38
rs906498847 Chr.5: 136056767-136056767 on Build GRCh38
rs552261238 Chr.5: 136056701-136056701 on Build GRCh38
rs950696749 Chr.5: 136056475-136056475 on Build GRCh38
rs923574898 Chr.5: 136056659-136056659 on Build GRCh38
rs760069147 Chr.5: 136056652-136056652 on Build GRCh38
rs371923032 Chr.5: 136056715-136056715 on Build GRCh38
rs61160827 Chr.5: 136056635-136056635 on Build GRCh38
rs138125037 Chr.5: 136056760-136056760 on Build GRCh38
rs774852613 Chr.5: 136056660-136056660 on Build GRCh38
rs367944058 Chr.5: 136056759-136056759 on Build GRCh38
rs756136153 Chr.5: 136056714-136056714 on Build GRCh38
rs951305517 Chr.5: 136056637-136056637 on Build GRCh38
rs2072239 Chr.5: 136056818-136056818 on Build GRCh38
rs121909209 Chr.5: 136056781-136056781 on Build GRCh38
rs202066916 Chr.5: 136056762-136056762 on Build GRCh38
rs571643353 Chr.5: 136056432-136056432 on Build GRCh38

View Full Product Details