Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138467698-138468241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR1
Entrez Gene ID
1958
Gene Name
early growth response 1
Gene Aliases
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225
UniGene
Hs.326035
Gene Chromosome Location
Chr.5: 138465492-138469315 on Build GRCh38
Transcripts
NM_001964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327807 c.1448C>T p.P483L Chr.5: 138467897-138467897 on Build GRCh38
7082476 c.1379C>A p.S460Y Chr.5: 138467828-138467828 on Build GRCh38
6327131 c.1429C>T p.P477S Chr.5: 138467878-138467878 on Build GRCh38
5455345 c.1613G>T p.R538M Chr.5: 138468062-138468062 on Build GRCh38
6323871 c.1481C>T p.S494L Chr.5: 138467930-138467930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767924969 Chr.5: 138467919-138467922 on Build GRCh38
rs751040100 Chr.5: 138468005-138468005 on Build GRCh38
rs768733193 Chr.5: 138467816-138467816 on Build GRCh38
rs562998786 Chr.5: 138467826-138467826 on Build GRCh38
rs956754060 Chr.5: 138467922-138467922 on Build GRCh38
rs139669376 Chr.5: 138467784-138467784 on Build GRCh38
rs758418994 Chr.5: 138468013-138468013 on Build GRCh38
rs915368252 Chr.5: 138467978-138467978 on Build GRCh38
rs922456828 Chr.5: 138467891-138467891 on Build GRCh38
rs202136756 Chr.5: 138468050-138468050 on Build GRCh38
rs546954332 Chr.5: 138468167-138468167 on Build GRCh38
rs911570660 Chr.5: 138468150-138468150 on Build GRCh38
rs753554596 Chr.5: 138467724-138467724 on Build GRCh38
rs778876846 Chr.5: 138467850-138467850 on Build GRCh38
rs571997670 Chr.5: 138467789-138467812 on Build GRCh38
rs534778183 Chr.5: 138467771-138467771 on Build GRCh38
rs200048883 Chr.5: 138467840-138467840 on Build GRCh38
rs893194804 Chr.5: 138468159-138468159 on Build GRCh38
rs202226814 Chr.5: 138467878-138467878 on Build GRCh38
rs760644725 Chr.5: 138467843-138467843 on Build GRCh38
rs746308508 Chr.5: 138467774-138467774 on Build GRCh38
rs757997080 Chr.5: 138467998-138467998 on Build GRCh38
rs201012682 Chr.5: 138467865-138467865 on Build GRCh38
rs949012675 Chr.5: 138468084-138468084 on Build GRCh38
rs199889910 Chr.5: 138468190-138468190 on Build GRCh38
rs183982123 Chr.5: 138468147-138468147 on Build GRCh38
rs865951226 Chr.5: 138467810-138467810 on Build GRCh38
rs780082755 Chr.5: 138468175-138468175 on Build GRCh38
rs955227243 Chr.5: 138467903-138467903 on Build GRCh38
rs752378281 Chr.5: 138467997-138467997 on Build GRCh38
rs759468344 Chr.5: 138467773-138467796 on Build GRCh38
rs202203243 Chr.5: 138467880-138467880 on Build GRCh38
rs199632861 Chr.5: 138468115-138468115 on Build GRCh38
rs199867643 Chr.5: 138467923-138467923 on Build GRCh38
rs978648148 Chr.5: 138468018-138468018 on Build GRCh38
rs767650411 Chr.5: 138467836-138467836 on Build GRCh38
rs28365162 Chr.5: 138467976-138467976 on Build GRCh38
rs764267087 Chr.5: 138467815-138467838 on Build GRCh38
rs140128606 Chr.5: 138467939-138467939 on Build GRCh38
rs779466738 Chr.5: 138468130-138468130 on Build GRCh38
rs763644281 Chr.5: 138468003-138468003 on Build GRCh38
rs773373715 Chr.5: 138467956-138467956 on Build GRCh38
rs200993915 Chr.5: 138468193-138468193 on Build GRCh38
rs201514005 Chr.5: 138468085-138468085 on Build GRCh38
rs373664381 Chr.5: 138467874-138467874 on Build GRCh38
rs145540497 Chr.5: 138467984-138467984 on Build GRCh38
rs202120386 Chr.5: 138467958-138467958 on Build GRCh38
rs774453334 Chr.5: 138467935-138467935 on Build GRCh38
rs201552434 Chr.5: 138467788-138467788 on Build GRCh38
rs201250503 Chr.5: 138467970-138467970 on Build GRCh38
rs759479225 Chr.5: 138468073-138468073 on Build GRCh38
rs776445332 Chr.5: 138468067-138468067 on Build GRCh38
rs201207618 Chr.5: 138467943-138467943 on Build GRCh38
rs777227296 Chr.5: 138467883-138467883 on Build GRCh38
rs766194467 Chr.5: 138467962-138467962 on Build GRCh38
rs898940454 Chr.5: 138468209-138468209 on Build GRCh38
rs772398081 Chr.5: 138468041-138468041 on Build GRCh38
rs553137681 Chr.5: 138467754-138467754 on Build GRCh38
rs201394271 Chr.5: 138467758-138467758 on Build GRCh38
rs200025697 Chr.5: 138468213-138468213 on Build GRCh38
rs147903263 Chr.5: 138467838-138467838 on Build GRCh38
rs748400232 Chr.5: 138468038-138468038 on Build GRCh38
rs201253879 Chr.5: 138468126-138468126 on Build GRCh38
rs773465307 Chr.5: 138468043-138468043 on Build GRCh38
rs753553653 Chr.5: 138467863-138467863 on Build GRCh38
rs995383525 Chr.5: 138468212-138468212 on Build GRCh38
rs763735848 Chr.5: 138468100-138468100 on Build GRCh38
rs779644730 Chr.5: 138467720-138467720 on Build GRCh38
rs762635880 Chr.5: 138468089-138468089 on Build GRCh38
rs149798677 Chr.5: 138467812-138467812 on Build GRCh38
rs759581124 Chr.5: 138468153-138468153 on Build GRCh38
rs921818491 Chr.5: 138468033-138468033 on Build GRCh38
rs199574070 Chr.5: 138467778-138467778 on Build GRCh38
rs756735460 Chr.5: 138468113-138468113 on Build GRCh38
rs201306291 Chr.5: 138468157-138468157 on Build GRCh38
rs200636536 Chr.5: 138468051-138468051 on Build GRCh38
rs775246141 Chr.5: 138468083-138468083 on Build GRCh38
rs756713503 Chr.5: 138468006-138468006 on Build GRCh38
rs779377556 Chr.5: 138468036-138468036 on Build GRCh38
rs762983434 Chr.5: 138467803-138467826 on Build GRCh38
rs778533252 Chr.5: 138467742-138467742 on Build GRCh38
rs541522909 Chr.5: 138467823-138467823 on Build GRCh38
rs768984243 Chr.5: 138467924-138467924 on Build GRCh38
rs200649112 Chr.5: 138468122-138468122 on Build GRCh38
rs199731696 Chr.5: 138468142-138468142 on Build GRCh38
rs200476410 Chr.5: 138467832-138467832 on Build GRCh38
rs775668956 Chr.5: 138467802-138467802 on Build GRCh38
rs746396391 Chr.5: 138467904-138467904 on Build GRCh38
rs373411522 Chr.5: 138467741-138467741 on Build GRCh38
rs371059427 Chr.5: 138468035-138468035 on Build GRCh38
rs770180164 Chr.5: 138467910-138467910 on Build GRCh38
rs771492229 Chr.5: 138467757-138467757 on Build GRCh38
rs749750277 Chr.5: 138468025-138468025 on Build GRCh38
rs751130334 Chr.5: 138468111-138468111 on Build GRCh38
rs769682068 Chr.5: 138467772-138467773 on Build GRCh38
rs147752848 Chr.5: 138468019-138468019 on Build GRCh38
rs766990363 Chr.5: 138468114-138468114 on Build GRCh38
rs780511164 Chr.5: 138467919-138467919 on Build GRCh38
rs763286707 Chr.5: 138467809-138467809 on Build GRCh38
rs774561952 Chr.5: 138467824-138467824 on Build GRCh38
rs762137361 Chr.5: 138467823-138467824 on Build GRCh38
rs774669212 Chr.5: 138467824-138467847 on Build GRCh38
rs376532290 Chr.5: 138467968-138467968 on Build GRCh38
rs879053776 Chr.5: 138467829-138467829 on Build GRCh38
rs776418216 Chr.5: 138467736-138467738 on Build GRCh38
rs370248873 Chr.5: 138467855-138467855 on Build GRCh38
rs200268457 Chr.5: 138467969-138467969 on Build GRCh38
rs754329993 Chr.5: 138468117-138468117 on Build GRCh38
rs764964083 Chr.5: 138468077-138468077 on Build GRCh38
rs201813217 Chr.5: 138468195-138468195 on Build GRCh38

View Full Product Details