Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141124432-141124945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB4
Entrez Gene ID
56131
Gene Name
protocadherin beta 4
Gene Aliases
PCDH-BETA4
UniGene
Hs.591250
Gene Chromosome Location
Chr.5: 141121750-141125623 on Build GRCh38
Transcripts
NM_018938.3

SNP Detail

SNP ID Chromosome Location
rs782238460 Chr.5: 141124490-141124490 on Build GRCh38
rs984490908 Chr.5: 141124798-141124798 on Build GRCh38
rs973004740 Chr.5: 141124463-141124463 on Build GRCh38
rs553841407 Chr.5: 141124491-141124491 on Build GRCh38
rs917744602 Chr.5: 141124911-141124911 on Build GRCh38
rs991892688 Chr.5: 141124902-141124902 on Build GRCh38
rs1042250238 Chr.5: 141124917-141124917 on Build GRCh38
rs146103699 Chr.5: 141124483-141124483 on Build GRCh38
rs1042322397 Chr.5: 141124650-141124650 on Build GRCh38
rs146020036 Chr.5: 141124865-141124865 on Build GRCh38
rs1056240240 Chr.5: 141124506-141124506 on Build GRCh38
rs961577783 Chr.5: 141124757-141124757 on Build GRCh38
rs945274719 Chr.5: 141124641-141124641 on Build GRCh38
rs113839187 Chr.5: 141124796-141124796 on Build GRCh38
rs572977888 Chr.5: 141124690-141124690 on Build GRCh38
rs181487745 Chr.5: 141124861-141124861 on Build GRCh38
rs3088331 Chr.5: 141124728-141124728 on Build GRCh38
rs544205467 Chr.5: 141124784-141124784 on Build GRCh38
rs538183510 Chr.5: 141124477-141124477 on Build GRCh38
rs903842743 Chr.5: 141124662-141124662 on Build GRCh38
rs782354763 Chr.5: 141124499-141124499 on Build GRCh38
rs994413639 Chr.5: 141124766-141124766 on Build GRCh38
rs75741396 Chr.5: 141124677-141124677 on Build GRCh38
rs569200348 Chr.5: 141124461-141124461 on Build GRCh38
rs782790193 Chr.5: 141124792-141124792 on Build GRCh38
rs945243648 Chr.5: 141124913-141124913 on Build GRCh38
rs896352147 Chr.5: 141124563-141124563 on Build GRCh38
rs189810600 Chr.5: 141124774-141124774 on Build GRCh38
rs1000879815 Chr.5: 141124666-141124666 on Build GRCh38
rs565638147 Chr.5: 141124504-141124504 on Build GRCh38
rs542042097 Chr.5: 141124726-141124726 on Build GRCh38
rs918923969 Chr.5: 141124479-141124479 on Build GRCh38
rs3822339 Chr.5: 141124850-141124850 on Build GRCh38
rs959139467 Chr.5: 141124856-141124856 on Build GRCh38
rs539434600 Chr.5: 141124560-141124560 on Build GRCh38
rs1008759830 Chr.5: 141124727-141124727 on Build GRCh38

View Full Product Details