Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141365030-141365523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619721 c.1086C>T p.P362P Chr.5: 141365416-141365416 on Build GRCh38
3851807 c.721C>A p.L241M Chr.5: 141365051-141365051 on Build GRCh38
6031229 c.744C>T p.S248S Chr.5: 141365074-141365074 on Build GRCh38
6359189 c.973G>A p.G325S Chr.5: 141365303-141365303 on Build GRCh38
5709528 c.1084delC p.G363fs*3 Chr.5: 141365414-141365414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548514157 Chr.5: 141365164-141365164 on Build GRCh38
rs200560640 Chr.5: 141365309-141365309 on Build GRCh38
rs369502854 Chr.5: 141365120-141365120 on Build GRCh38
rs34496075 Chr.5: 141365418-141365419 on Build GRCh38
rs1010180647 Chr.5: 141365439-141365439 on Build GRCh38
rs752434944 Chr.5: 141365375-141365375 on Build GRCh38
rs552296703 Chr.5: 141365321-141365321 on Build GRCh38
rs757034193 Chr.5: 141365307-141365307 on Build GRCh38
rs554930491 Chr.5: 141365396-141365396 on Build GRCh38
rs373921082 Chr.5: 141365125-141365125 on Build GRCh38
rs764671143 Chr.5: 141365117-141365117 on Build GRCh38
rs774588043 Chr.5: 141365090-141365090 on Build GRCh38
rs961817012 Chr.5: 141365418-141365418 on Build GRCh38
rs761258930 Chr.5: 141365365-141365365 on Build GRCh38
rs546328235 Chr.5: 141365449-141365449 on Build GRCh38
rs748153099 Chr.5: 141365079-141365079 on Build GRCh38
rs777231006 Chr.5: 141365220-141365220 on Build GRCh38
rs538677777 Chr.5: 141365297-141365297 on Build GRCh38
rs184618404 Chr.5: 141365071-141365071 on Build GRCh38
rs962224901 Chr.5: 141365224-141365224 on Build GRCh38
rs199512708 Chr.5: 141365274-141365274 on Build GRCh38
rs765751985 Chr.5: 141365237-141365237 on Build GRCh38
rs781264093 Chr.5: 141365429-141365429 on Build GRCh38
rs751303366 Chr.5: 141365131-141365131 on Build GRCh38
rs758172625 Chr.5: 141365486-141365486 on Build GRCh38
rs937776299 Chr.5: 141365489-141365489 on Build GRCh38
rs753864685 Chr.5: 141365305-141365305 on Build GRCh38
rs765123370 Chr.5: 141365478-141365478 on Build GRCh38
rs75563633 Chr.5: 141365084-141365084 on Build GRCh38
rs537066811 Chr.5: 141365422-141365422 on Build GRCh38
rs749640886 Chr.5: 141365441-141365441 on Build GRCh38
rs770375441 Chr.5: 141365434-141365434 on Build GRCh38
rs926241628 Chr.5: 141365458-141365458 on Build GRCh38
rs748996894 Chr.5: 141365174-141365174 on Build GRCh38
rs1048742374 Chr.5: 141365077-141365077 on Build GRCh38
rs769625651 Chr.5: 141365082-141365082 on Build GRCh38
rs1056307044 Chr.5: 141365108-141365108 on Build GRCh38
rs777540442 Chr.5: 141365299-141365299 on Build GRCh38
rs779776168 Chr.5: 141365064-141365064 on Build GRCh38
rs896268643 Chr.5: 141365137-141365137 on Build GRCh38
rs750087491 Chr.5: 141365060-141365060 on Build GRCh38
rs747695499 Chr.5: 141365152-141365152 on Build GRCh38
rs931548015 Chr.5: 141365447-141365447 on Build GRCh38
rs529687184 Chr.5: 141365180-141365180 on Build GRCh38
rs372623504 Chr.5: 141365325-141365325 on Build GRCh38
rs549713754 Chr.5: 141365181-141365181 on Build GRCh38
rs754097120 Chr.5: 141365118-141365118 on Build GRCh38
rs151160787 Chr.5: 141365430-141365430 on Build GRCh38
rs747026177 Chr.5: 141365327-141365327 on Build GRCh38
rs751252073 Chr.5: 141365260-141365260 on Build GRCh38
rs771010337 Chr.5: 141365087-141365087 on Build GRCh38
rs536146431 Chr.5: 141365242-141365242 on Build GRCh38
rs761340226 Chr.5: 141365467-141365467 on Build GRCh38
rs540403118 Chr.5: 141365112-141365112 on Build GRCh38
rs747311878 Chr.5: 141365419-141365419 on Build GRCh38
rs775485996 Chr.5: 141365186-141365186 on Build GRCh38
rs764735476 Chr.5: 141365366-141365366 on Build GRCh38
rs752099096 Chr.5: 141365281-141365281 on Build GRCh38
rs754573655 Chr.5: 141365075-141365075 on Build GRCh38
rs762059066 Chr.5: 141365232-141365232 on Build GRCh38
rs780914811 Chr.5: 141365146-141365146 on Build GRCh38
rs763719415 Chr.5: 141365391-141365391 on Build GRCh38
rs767635242 Chr.5: 141365099-141365099 on Build GRCh38
rs189137040 Chr.5: 141365239-141365239 on Build GRCh38
rs75982128 Chr.5: 141365139-141365139 on Build GRCh38
rs761155361 Chr.5: 141365110-141365110 on Build GRCh38
rs569659080 Chr.5: 141365219-141365219 on Build GRCh38
rs766260971 Chr.5: 141365497-141365497 on Build GRCh38
rs568315403 Chr.5: 141365414-141365414 on Build GRCh38
rs762446364 Chr.5: 141365347-141365347 on Build GRCh38
rs776988694 Chr.5: 141365340-141365340 on Build GRCh38
rs115453161 Chr.5: 141365386-141365386 on Build GRCh38
rs763661542 Chr.5: 141365450-141365450 on Build GRCh38
rs565922694 Chr.5: 141365353-141365353 on Build GRCh38
rs774993961 Chr.5: 141365445-141365445 on Build GRCh38
rs745669256 Chr.5: 141365319-141365319 on Build GRCh38
rs769039827 Chr.5: 141365058-141365058 on Build GRCh38
rs553043540 Chr.5: 141365451-141365451 on Build GRCh38
rs750173961 Chr.5: 141365401-141365401 on Build GRCh38
rs750405300 Chr.5: 141365483-141365483 on Build GRCh38
rs182594355 Chr.5: 141365423-141365423 on Build GRCh38
rs376872993 Chr.5: 141365210-141365210 on Build GRCh38
rs759196627 Chr.5: 141365265-141365265 on Build GRCh38
rs754518751 Chr.5: 141365145-141365145 on Build GRCh38
rs753717509 Chr.5: 141365395-141365395 on Build GRCh38
rs201444039 Chr.5: 141365167-141365167 on Build GRCh38
rs757924486 Chr.5: 141365063-141365063 on Build GRCh38
rs528661266 Chr.5: 141365093-141365093 on Build GRCh38
rs770428432 Chr.5: 141365348-141365348 on Build GRCh38
rs906812137 Chr.5: 141365463-141365463 on Build GRCh38
rs771471249 Chr.5: 141365444-141365444 on Build GRCh38
rs766093939 Chr.5: 141365130-141365130 on Build GRCh38
rs181194736 Chr.5: 141365055-141365055 on Build GRCh38
rs775966290 Chr.5: 141365104-141365104 on Build GRCh38
rs376885852 Chr.5: 141365326-141365326 on Build GRCh38
rs780113082 Chr.5: 141365416-141365416 on Build GRCh38
rs768826451 Chr.5: 141365329-141365329 on Build GRCh38
rs562030939 Chr.5: 141365175-141365175 on Build GRCh38

View Full Product Details