Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141954880-141955387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH12
Entrez Gene ID
51294
Gene Name
protocadherin 12
Gene Aliases
VE-cadherin-2, VECAD2
UniGene
Hs.439474
Gene Chromosome Location
Chr.5: 141943585-141959062 on Build GRCh38
Transcripts
NM_016580.3 XM_017010070.1 XM_017010076.1

Gene Symbol
RNF14
Entrez Gene ID
9604
Gene Name
ring finger protein 14
Gene Aliases
ARA54, HFB30, HRIHFB2038, TRIAD2
UniGene
Hs.483616
Gene Chromosome Location
Chr.5: 141949337-141993709 on Build GRCh38
Transcripts
NM_016580.3 XM_017010070.1 XM_017010076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3611817 c.2662G>A p.G888R Chr.5: 141955190-141955190 on Build GRCh38
1496013 c.2504C>T p.P835L Chr.5: 141955348-141955348 on Build GRCh38
5554369 c.2829C>T p.F943F Chr.5: 141955023-141955023 on Build GRCh38
5902746 c.2617G>A p.G873S Chr.5: 141955235-141955235 on Build GRCh38
5560063 c.2466G>A p.L822L Chr.5: 141955386-141955386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750087149 Chr.5: 141955172-141955172 on Build GRCh38
rs768146683 Chr.5: 141955163-141955163 on Build GRCh38
rs746450107 Chr.5: 141955253-141955253 on Build GRCh38
rs777396555 Chr.5: 141955059-141955059 on Build GRCh38
rs1048025807 Chr.5: 141955289-141955289 on Build GRCh38
rs1035719913 Chr.5: 141955130-141955130 on Build GRCh38
rs747183572 Chr.5: 141955046-141955046 on Build GRCh38
rs374569241 Chr.5: 141955260-141955260 on Build GRCh38
rs747907583 Chr.5: 141955001-141955001 on Build GRCh38
rs749595350 Chr.5: 141955325-141955325 on Build GRCh38
rs776847273 Chr.5: 141955350-141955350 on Build GRCh38
rs1040074840 Chr.5: 141955354-141955354 on Build GRCh38
rs758295361 Chr.5: 141955248-141955248 on Build GRCh38
rs780448801 Chr.5: 141955000-141955000 on Build GRCh38
rs538775199 Chr.5: 141955347-141955347 on Build GRCh38
rs200214038 Chr.5: 141955204-141955204 on Build GRCh38
rs764333172 Chr.5: 141955292-141955292 on Build GRCh38
rs2434322 Chr.5: 141955356-141955356 on Build GRCh38
rs773755967 Chr.5: 141955186-141955186 on Build GRCh38
rs775129851 Chr.5: 141955016-141955016 on Build GRCh38
rs375840810 Chr.5: 141955015-141955015 on Build GRCh38
rs750172243 Chr.5: 141955296-141955296 on Build GRCh38
rs751543859 Chr.5: 141955214-141955214 on Build GRCh38
rs375346212 Chr.5: 141955337-141955337 on Build GRCh38
rs767176274 Chr.5: 141955073-141955074 on Build GRCh38
rs771321515 Chr.5: 141955330-141955330 on Build GRCh38
rs546412869 Chr.5: 141954908-141954908 on Build GRCh38
rs769469035 Chr.5: 141955002-141955002 on Build GRCh38
rs372052882 Chr.5: 141955038-141955038 on Build GRCh38
rs569368318 Chr.5: 141955114-141955114 on Build GRCh38
rs199848714 Chr.5: 141955079-141955079 on Build GRCh38
rs1026100054 Chr.5: 141955057-141955057 on Build GRCh38
rs771283801 Chr.5: 141955269-141955269 on Build GRCh38
rs776824903 Chr.5: 141955099-141955099 on Build GRCh38
rs760445554 Chr.5: 141955019-141955019 on Build GRCh38
rs149882545 Chr.5: 141955023-141955023 on Build GRCh38
rs755272537 Chr.5: 141955055-141955055 on Build GRCh38
rs182860036 Chr.5: 141955067-141955067 on Build GRCh38
rs439293 Chr.5: 141955124-141955124 on Build GRCh38
rs752769310 Chr.5: 141955279-141955279 on Build GRCh38
rs755776573 Chr.5: 141955321-141955321 on Build GRCh38
rs756386790 Chr.5: 141955234-141955234 on Build GRCh38
rs757050421 Chr.5: 141955140-141955140 on Build GRCh38
rs773668098 Chr.5: 141955265-141955265 on Build GRCh38
rs779117992 Chr.5: 141954981-141954981 on Build GRCh38
rs147558478 Chr.5: 141955139-141955139 on Build GRCh38
rs770265325 Chr.5: 141955185-141955185 on Build GRCh38
rs778658899 Chr.5: 141955235-141955235 on Build GRCh38
rs372344966 Chr.5: 141954954-141954954 on Build GRCh38
rs369976753 Chr.5: 141955028-141955028 on Build GRCh38
rs906145048 Chr.5: 141955071-141955071 on Build GRCh38
rs140005536 Chr.5: 141955195-141955195 on Build GRCh38
rs141990944 Chr.5: 141955007-141955007 on Build GRCh38
rs779793170 Chr.5: 141955317-141955317 on Build GRCh38
rs767622193 Chr.5: 141955276-141955276 on Build GRCh38
rs763432404 Chr.5: 141955266-141955266 on Build GRCh38
rs758205450 Chr.5: 141955311-141955311 on Build GRCh38
rs750958908 Chr.5: 141955117-141955117 on Build GRCh38
rs754901453 Chr.5: 141955363-141955363 on Build GRCh38
rs142096693 Chr.5: 141955151-141955151 on Build GRCh38
rs979374348 Chr.5: 141954906-141954906 on Build GRCh38
rs772386316 Chr.5: 141954995-141954995 on Build GRCh38
rs778748473 Chr.5: 141955143-141955143 on Build GRCh38
rs373022057 Chr.5: 141955112-141955112 on Build GRCh38
rs565373949 Chr.5: 141955249-141955249 on Build GRCh38
rs61737138 Chr.5: 141955217-141955217 on Build GRCh38
rs745989411 Chr.5: 141954990-141954990 on Build GRCh38
rs964316740 Chr.5: 141955089-141955089 on Build GRCh38
rs375930414 Chr.5: 141955020-141955020 on Build GRCh38
rs373091541 Chr.5: 141955066-141955066 on Build GRCh38
rs763851054 Chr.5: 141955092-141955092 on Build GRCh38
rs775039753 Chr.5: 141955085-141955085 on Build GRCh38
rs932328568 Chr.5: 141954989-141954989 on Build GRCh38
rs767567681 Chr.5: 141955225-141955225 on Build GRCh38
rs760857936 Chr.5: 141955285-141955285 on Build GRCh38
rs780344780 Chr.5: 141955042-141955042 on Build GRCh38
rs368872362 Chr.5: 141955318-141955318 on Build GRCh38
rs920894091 Chr.5: 141954950-141954950 on Build GRCh38
rs915423832 Chr.5: 141954918-141954918 on Build GRCh38
rs747688840 Chr.5: 141955320-141955320 on Build GRCh38
rs574087581 Chr.5: 141955118-141955118 on Build GRCh38
rs762204744 Chr.5: 141955182-141955182 on Build GRCh38
rs757841190 Chr.5: 141954966-141954966 on Build GRCh38
rs760027626 Chr.5: 141955274-141955274 on Build GRCh38
rs762262497 Chr.5: 141955100-141955100 on Build GRCh38
rs778277706 Chr.5: 141954927-141954927 on Build GRCh38
rs767020872 Chr.5: 141955206-141955206 on Build GRCh38
rs746563397 Chr.5: 141955158-141955158 on Build GRCh38
rs377464118 Chr.5: 141955135-141955135 on Build GRCh38
rs781497781 Chr.5: 141955138-141955138 on Build GRCh38
rs777580158 Chr.5: 141955322-141955322 on Build GRCh38
rs774638036 Chr.5: 141955336-141955336 on Build GRCh38
rs551312542 Chr.5: 141955022-141955022 on Build GRCh38
rs148879785 Chr.5: 141955254-141955254 on Build GRCh38
rs759067548 Chr.5: 141955014-141955014 on Build GRCh38
rs751347629 Chr.5: 141955360-141955360 on Build GRCh38
rs760223768 Chr.5: 141955086-141955086 on Build GRCh38
rs199668488 Chr.5: 141955344-141955344 on Build GRCh38
rs141083707 Chr.5: 141955008-141955008 on Build GRCh38
rs533737041 Chr.5: 141955226-141955226 on Build GRCh38
rs975363306 Chr.5: 141954909-141954909 on Build GRCh38
rs773810499 Chr.5: 141955010-141955010 on Build GRCh38
rs143534629 Chr.5: 141955273-141955273 on Build GRCh38
rs769608793 Chr.5: 141955261-141955261 on Build GRCh38
rs370507314 Chr.5: 141955109-141955109 on Build GRCh38
rs749138024 Chr.5: 141955062-141955062 on Build GRCh38
rs1044682229 Chr.5: 141955026-141955026 on Build GRCh38
rs375734807 Chr.5: 141955355-141955355 on Build GRCh38
rs61737140 Chr.5: 141955156-141955156 on Build GRCh38
rs750485056 Chr.5: 141955035-141955035 on Build GRCh38
rs776363111 Chr.5: 141955167-141955167 on Build GRCh38
rs764245491 Chr.5: 141955348-141955348 on Build GRCh38
rs376177062 Chr.5: 141955132-141955132 on Build GRCh38

View Full Product Details