Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154456616-154457109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP30L
Entrez Gene ID
79685
Gene Name
SAP30 like
Gene Aliases
NS4ATP2
UniGene
Hs.592566
Gene Chromosome Location
Chr.5: 154445957-154461053 on Build GRCh38
Transcripts
NM_001131062.1 NM_001131063.1 NM_024632.5 NR_024084.1

SNP Detail

SNP ID Chromosome Location
rs777403982 Chr.5: 154456707-154456707 on Build GRCh38
rs915528968 Chr.5: 154456647-154456647 on Build GRCh38
rs906418210 Chr.5: 154456897-154456897 on Build GRCh38
rs1013115161 Chr.5: 154456986-154456986 on Build GRCh38
rs979463120 Chr.5: 154456700-154456700 on Build GRCh38
rs978440169 Chr.5: 154457045-154457045 on Build GRCh38
rs932702927 Chr.5: 154456812-154456812 on Build GRCh38
rs139576612 Chr.5: 154456811-154456811 on Build GRCh38
rs146776079 Chr.5: 154457040-154457041 on Build GRCh38
rs542470931 Chr.5: 154456832-154456832 on Build GRCh38
rs560734922 Chr.5: 154456835-154456835 on Build GRCh38
rs1041887766 Chr.5: 154456790-154456790 on Build GRCh38
rs922811941 Chr.5: 154456740-154456740 on Build GRCh38
rs776460524 Chr.5: 154456878-154456878 on Build GRCh38
rs571193378 Chr.5: 154456975-154456975 on Build GRCh38
rs578062783 Chr.5: 154456932-154456932 on Build GRCh38
rs150039435 Chr.5: 154456888-154456888 on Build GRCh38
rs746657709 Chr.5: 154456765-154456765 on Build GRCh38
rs750543562 Chr.5: 154456879-154456879 on Build GRCh38
rs895577531 Chr.5: 154456985-154456985 on Build GRCh38
rs993348843 Chr.5: 154456896-154456896 on Build GRCh38
rs1022710746 Chr.5: 154456993-154456993 on Build GRCh38
rs975684179 Chr.5: 154456716-154456716 on Build GRCh38
rs35475649 Chr.5: 154456817-154456818 on Build GRCh38
rs924848345 Chr.5: 154456701-154456701 on Build GRCh38
rs767706869 Chr.5: 154456751-154456751 on Build GRCh38
rs1049826976 Chr.5: 154456825-154456825 on Build GRCh38
rs572146731 Chr.5: 154456766-154456766 on Build GRCh38
rs968406413 Chr.5: 154456659-154456659 on Build GRCh38
rs528216525 Chr.5: 154456848-154456848 on Build GRCh38
rs145315671 Chr.5: 154457057-154457057 on Build GRCh38
rs1038051699 Chr.5: 154456860-154456862 on Build GRCh38
rs935057376 Chr.5: 154456827-154456827 on Build GRCh38

View Full Product Details