Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163444488-163445032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG1
Entrez Gene ID
900
Gene Name
cyclin G1
Gene Aliases
CCNG
UniGene
Hs.79101
Gene Chromosome Location
Chr.5: 163437567-163445016 on Build GRCh38
Transcripts
NM_004060.3 NM_199246.1

SNP Detail

SNP ID Chromosome Location
rs967204973 Chr.5: 163444650-163444650 on Build GRCh38
rs779450810 Chr.5: 163444753-163444753 on Build GRCh38
rs1050616118 Chr.5: 163444955-163444955 on Build GRCh38
rs756949850 Chr.5: 163444841-163444841 on Build GRCh38
rs973401967 Chr.5: 163444738-163444738 on Build GRCh38
rs999220016 Chr.5: 163444597-163444598 on Build GRCh38
rs908250717 Chr.5: 163444824-163444824 on Build GRCh38
rs571921717 Chr.5: 163444657-163444659 on Build GRCh38
rs949214090 Chr.5: 163444512-163444513 on Build GRCh38
rs902401376 Chr.5: 163444530-163444532 on Build GRCh38
rs1014085108 Chr.5: 163444620-163444620 on Build GRCh38
rs12657994 Chr.5: 163444761-163444761 on Build GRCh38
rs192535026 Chr.5: 163444835-163444835 on Build GRCh38
rs565278287 Chr.5: 163444879-163444879 on Build GRCh38
rs17883275 Chr.5: 163444718-163444718 on Build GRCh38
rs923877142 Chr.5: 163444813-163444813 on Build GRCh38
rs1053481657 Chr.5: 163444870-163444870 on Build GRCh38
rs896519936 Chr.5: 163444616-163444616 on Build GRCh38
rs1804633 Chr.5: 163444854-163444854 on Build GRCh38
rs1015145022 Chr.5: 163444687-163444687 on Build GRCh38
rs112172238 Chr.5: 163444805-163444805 on Build GRCh38
rs532529210 Chr.5: 163444923-163444923 on Build GRCh38
rs962258420 Chr.5: 163444691-163444691 on Build GRCh38
rs560246257 Chr.5: 163444685-163444685 on Build GRCh38
rs369896651 Chr.5: 163444655-163444655 on Build GRCh38
rs893481582 Chr.5: 163444911-163444911 on Build GRCh38
rs975023210 Chr.5: 163444786-163444786 on Build GRCh38
rs190798116 Chr.5: 163444789-163444789 on Build GRCh38
rs572046790 Chr.5: 163444757-163444757 on Build GRCh38
rs1055006009 Chr.5: 163444980-163444980 on Build GRCh38
rs1046747597 Chr.5: 163444518-163444518 on Build GRCh38
rs1028705124 Chr.5: 163444660-163444660 on Build GRCh38
rs547589899 Chr.5: 163444945-163444945 on Build GRCh38
rs896397036 Chr.5: 163444984-163444984 on Build GRCh38
rs1033622254 Chr.5: 163444520-163444520 on Build GRCh38
rs534968287 Chr.5: 163444939-163444939 on Build GRCh38
rs986873915 Chr.5: 163444792-163444792 on Build GRCh38
rs958852779 Chr.5: 163444781-163444781 on Build GRCh38
rs1038144142 Chr.5: 163444898-163444898 on Build GRCh38
rs1020999567 Chr.5: 163444647-163444647 on Build GRCh38
rs955046302 Chr.5: 163444678-163444678 on Build GRCh38
rs898357254 Chr.5: 163444919-163444919 on Build GRCh38
rs185471595 Chr.5: 163444566-163444566 on Build GRCh38
rs920921860 Chr.5: 163444788-163444788 on Build GRCh38

View Full Product Details