Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178884077-178884594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF354B
Entrez Gene ID
117608
Gene Name
zinc finger protein 354B
Gene Aliases
KID2
UniGene
Hs.724051
Gene Chromosome Location
Chr.5: 178859813-178901135 on Build GRCh38
Transcripts
NM_058230.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829624 c.1823C>T p.A608V Chr.5: 178884275-178884275 on Build GRCh38
1066535 c.1625G>T p.R542M Chr.5: 178884077-178884077 on Build GRCh38
4585757 c.1696A>G p.I566V Chr.5: 178884148-178884148 on Build GRCh38
1066536 c.1825G>A p.D609N Chr.5: 178884277-178884277 on Build GRCh38
5696229 c.1769G>A p.R590K Chr.5: 178884221-178884221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770628874 Chr.5: 178884105-178884105 on Build GRCh38
rs536000292 Chr.5: 178884552-178884555 on Build GRCh38
rs1043071871 Chr.5: 178884501-178884501 on Build GRCh38
rs533913294 Chr.5: 178884558-178884558 on Build GRCh38
rs1052308854 Chr.5: 178884535-178884535 on Build GRCh38
rs376195716 Chr.5: 178884293-178884293 on Build GRCh38
rs749238105 Chr.5: 178884228-178884228 on Build GRCh38
rs572833440 Chr.5: 178884540-178884540 on Build GRCh38
rs191967030 Chr.5: 178884567-178884567 on Build GRCh38
rs947922169 Chr.5: 178884536-178884536 on Build GRCh38
rs771001150 Chr.5: 178884240-178884240 on Build GRCh38
rs936542926 Chr.5: 178884499-178884499 on Build GRCh38
rs77568902 Chr.5: 178884259-178884259 on Build GRCh38
rs982916252 Chr.5: 178884411-178884411 on Build GRCh38
rs765748276 Chr.5: 178884148-178884148 on Build GRCh38
rs550469209 Chr.5: 178884371-178884371 on Build GRCh38
rs755901047 Chr.5: 178884198-178884198 on Build GRCh38
rs890523620 Chr.5: 178884537-178884537 on Build GRCh38
rs755921133 Chr.5: 178884310-178884310 on Build GRCh38
rs763734854 Chr.5: 178884313-178884313 on Build GRCh38
rs142040814 Chr.5: 178884276-178884276 on Build GRCh38
rs946923186 Chr.5: 178884425-178884425 on Build GRCh38
rs763383670 Chr.5: 178884268-178884268 on Build GRCh38
rs559023412 Chr.5: 178884277-178884277 on Build GRCh38
rs755154369 Chr.5: 178884185-178884185 on Build GRCh38
rs947634615 Chr.5: 178884472-178884472 on Build GRCh38
rs74462474 Chr.5: 178884352-178884352 on Build GRCh38
rs528254971 Chr.5: 178884342-178884342 on Build GRCh38
rs919550584 Chr.5: 178884368-178884368 on Build GRCh38
rs773410824 Chr.5: 178884330-178884330 on Build GRCh38
rs779577479 Chr.5: 178884239-178884241 on Build GRCh38
rs903144960 Chr.5: 178884518-178884518 on Build GRCh38
rs78387453 Chr.5: 178884389-178884389 on Build GRCh38
rs76329310 Chr.5: 178884467-178884467 on Build GRCh38
rs978108867 Chr.5: 178884454-178884454 on Build GRCh38
rs144854572 Chr.5: 178884170-178884170 on Build GRCh38
rs769211575 Chr.5: 178884233-178884237 on Build GRCh38
rs55889185 Chr.5: 178884164-178884164 on Build GRCh38
rs747493506 Chr.5: 178884230-178884231 on Build GRCh38
rs998960231 Chr.5: 178884531-178884531 on Build GRCh38
rs779046389 Chr.5: 178884318-178884318 on Build GRCh38
rs944039116 Chr.5: 178884415-178884415 on Build GRCh38
rs778496928 Chr.5: 178884242-178884242 on Build GRCh38
rs748716907 Chr.5: 178884309-178884311 on Build GRCh38
rs747599443 Chr.5: 178884448-178884448 on Build GRCh38
rs779747553 Chr.5: 178884322-178884322 on Build GRCh38
rs752512493 Chr.5: 178884194-178884194 on Build GRCh38
rs546462926 Chr.5: 178884348-178884348 on Build GRCh38
rs182051593 Chr.5: 178884317-178884317 on Build GRCh38
rs554613721 Chr.5: 178884519-178884519 on Build GRCh38
rs751645649 Chr.5: 178884171-178884171 on Build GRCh38
rs768345457 Chr.5: 178884311-178884315 on Build GRCh38
rs777008335 Chr.5: 178884144-178884144 on Build GRCh38
rs752928959 Chr.5: 178884301-178884301 on Build GRCh38
rs368083878 Chr.5: 178884138-178884138 on Build GRCh38
rs894717836 Chr.5: 178884151-178884151 on Build GRCh38
rs189588149 Chr.5: 178884264-178884264 on Build GRCh38
rs745342195 Chr.5: 178884319-178884319 on Build GRCh38
rs546466918 Chr.5: 178884169-178884169 on Build GRCh38
rs777761465 Chr.5: 178884216-178884216 on Build GRCh38
rs773979905 Chr.5: 178884329-178884330 on Build GRCh38
rs140053195 Chr.5: 178884321-178884321 on Build GRCh38
rs762136494 Chr.5: 178884146-178884146 on Build GRCh38
rs750963394 Chr.5: 178884149-178884149 on Build GRCh38
rs906306662 Chr.5: 178884554-178884554 on Build GRCh38
rs867552856 Chr.5: 178884250-178884250 on Build GRCh38
rs372012541 Chr.5: 178884186-178884186 on Build GRCh38
rs780410610 Chr.5: 178884241-178884241 on Build GRCh38
rs985559288 Chr.5: 178884374-178884374 on Build GRCh38
rs768360431 Chr.5: 178884325-178884325 on Build GRCh38
rs61740831 Chr.5: 178884249-178884249 on Build GRCh38
rs187659702 Chr.5: 178884355-178884355 on Build GRCh38
rs75617526 Chr.5: 178884566-178884566 on Build GRCh38
rs372542910 Chr.5: 178884116-178884116 on Build GRCh38
rs753687416 Chr.5: 178884314-178884314 on Build GRCh38
rs369181418 Chr.5: 178884333-178884333 on Build GRCh38
rs771404130 Chr.5: 178884340-178884340 on Build GRCh38
rs746585082 Chr.5: 178884323-178884323 on Build GRCh38
rs1053709638 Chr.5: 178884508-178884508 on Build GRCh38
rs938881975 Chr.5: 178884441-178884441 on Build GRCh38
rs1030839060 Chr.5: 178884297-178884297 on Build GRCh38
rs771830989 Chr.5: 178884245-178884245 on Build GRCh38
rs778431617 Chr.5: 178884209-178884212 on Build GRCh38
rs995790246 Chr.5: 178884567-178884567 on Build GRCh38

View Full Product Details