Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179798059-179798560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT4B
Entrez Gene ID
11282
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
Gene Aliases
GNT-IV, GNT-IVB
UniGene
Hs.567419
Gene Chromosome Location
Chr.5: 179797597-179806952 on Build GRCh38
Transcripts
NM_014275.4 NM_054013.3 XM_017008998.1 NR_031598.1

Gene Symbol
MIR1229
Entrez Gene ID
100302156
Gene Name
microRNA 1229
Gene Aliases
MIRN1229, hsa-mir-1229
UniGene
Gene Chromosome Location
Chr.5: 179798278-179798346 on Build GRCh38
Transcripts
NM_014275.4 NM_054013.3 XM_017008998.1 NR_031598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1636843 c.1467+1G>C p.? Chr.5: 179798512-179798512 on Build GRCh38
6170897 c.1420G>A p.D474N Chr.5: 179798515-179798515 on Build GRCh38
5171603 c.1397delC p.T466fs*72 Chr.5: 179798538-179798538 on Build GRCh38
1486668 c.1474C>T p.Q492* Chr.5: 179798428-179798428 on Build GRCh38
5601808 c.1579G>A p.E527K Chr.5: 179798254-179798254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777501919 Chr.5: 179798137-179798138 on Build GRCh38
rs772771034 Chr.5: 179798134-179798134 on Build GRCh38
rs745479806 Chr.5: 179798332-179798332 on Build GRCh38
rs750019683 Chr.5: 179798463-179798463 on Build GRCh38
rs770355423 Chr.5: 179798343-179798343 on Build GRCh38
rs776625019 Chr.5: 179798118-179798118 on Build GRCh38
rs139490632 Chr.5: 179798106-179798106 on Build GRCh38
rs567672196 Chr.5: 179798301-179798301 on Build GRCh38
rs973155282 Chr.5: 179798338-179798338 on Build GRCh38
rs567386290 Chr.5: 179798527-179798527 on Build GRCh38
rs762055070 Chr.5: 179798340-179798340 on Build GRCh38
rs552336424 Chr.5: 179798513-179798513 on Build GRCh38
rs200918506 Chr.5: 179798274-179798274 on Build GRCh38
rs996111054 Chr.5: 179798158-179798158 on Build GRCh38
rs370845309 Chr.5: 179798376-179798376 on Build GRCh38
rs767835386 Chr.5: 179798428-179798428 on Build GRCh38
rs771976741 Chr.5: 179798515-179798515 on Build GRCh38
rs778966833 Chr.5: 179798455-179798455 on Build GRCh38
rs142783315 Chr.5: 179798348-179798348 on Build GRCh38
rs751811285 Chr.5: 179798363-179798363 on Build GRCh38
rs770776158 Chr.5: 179798239-179798239 on Build GRCh38
rs776812428 Chr.5: 179798374-179798374 on Build GRCh38
rs1031042416 Chr.5: 179798238-179798238 on Build GRCh38
rs371210487 Chr.5: 179798136-179798136 on Build GRCh38
rs748599210 Chr.5: 179798382-179798382 on Build GRCh38
rs79572903 Chr.5: 179798504-179798504 on Build GRCh38
rs771882220 Chr.5: 179798104-179798104 on Build GRCh38
rs774529356 Chr.5: 179798412-179798412 on Build GRCh38
rs765636550 Chr.5: 179798276-179798276 on Build GRCh38
rs150645557 Chr.5: 179798142-179798142 on Build GRCh38
rs748605549 Chr.5: 179798397-179798397 on Build GRCh38
rs149467882 Chr.5: 179798167-179798167 on Build GRCh38
rs753934806 Chr.5: 179798089-179798089 on Build GRCh38
rs114723280 Chr.5: 179798099-179798099 on Build GRCh38
rs374415178 Chr.5: 179798211-179798211 on Build GRCh38
rs770087871 Chr.5: 179798398-179798398 on Build GRCh38
rs750663440 Chr.5: 179798179-179798179 on Build GRCh38
rs754734738 Chr.5: 179798300-179798300 on Build GRCh38
rs762647419 Chr.5: 179798135-179798135 on Build GRCh38
rs2291417 Chr.5: 179798480-179798480 on Build GRCh38
rs763937478 Chr.5: 179798441-179798441 on Build GRCh38
rs537588120 Chr.5: 179798195-179798195 on Build GRCh38
rs747872224 Chr.5: 179798304-179798304 on Build GRCh38
rs760926700 Chr.5: 179798536-179798536 on Build GRCh38
rs770539407 Chr.5: 179798147-179798148 on Build GRCh38
rs937298997 Chr.5: 179798323-179798323 on Build GRCh38
rs781014343 Chr.5: 179798322-179798322 on Build GRCh38
rs746576885 Chr.5: 179798486-179798486 on Build GRCh38
rs747668331 Chr.5: 179798119-179798119 on Build GRCh38
rs145401676 Chr.5: 179798499-179798499 on Build GRCh38
rs141885208 Chr.5: 179798538-179798538 on Build GRCh38
rs1046158939 Chr.5: 179798084-179798084 on Build GRCh38
rs2291418 Chr.5: 179798324-179798324 on Build GRCh38
rs763035047 Chr.5: 179798354-179798354 on Build GRCh38
rs370863172 Chr.5: 179798360-179798360 on Build GRCh38
rs200916134 Chr.5: 179798399-179798399 on Build GRCh38
rs368430313 Chr.5: 179798215-179798215 on Build GRCh38
rs745658853 Chr.5: 179798507-179798531 on Build GRCh38
rs769290385 Chr.5: 179798126-179798126 on Build GRCh38
rs370796263 Chr.5: 179798313-179798313 on Build GRCh38
rs778314434 Chr.5: 179798080-179798080 on Build GRCh38
rs368655234 Chr.5: 179798393-179798393 on Build GRCh38
rs967482366 Chr.5: 179798241-179798241 on Build GRCh38
rs371351604 Chr.5: 179798235-179798235 on Build GRCh38
rs761119315 Chr.5: 179798142-179798142 on Build GRCh38
rs1020161322 Chr.5: 179798418-179798418 on Build GRCh38
rs993360746 Chr.5: 179798096-179798096 on Build GRCh38
rs753845071 Chr.5: 179798447-179798447 on Build GRCh38
rs769211175 Chr.5: 179798231-179798231 on Build GRCh38
rs377397954 Chr.5: 179798306-179798306 on Build GRCh38
rs750539490 Chr.5: 179798277-179798277 on Build GRCh38
rs746019568 Chr.5: 179798100-179798100 on Build GRCh38
rs200647784 Chr.5: 179798292-179798292 on Build GRCh38
rs757349151 Chr.5: 179798093-179798093 on Build GRCh38
rs758465824 Chr.5: 179798283-179798283 on Build GRCh38
rs768617757 Chr.5: 179798110-179798110 on Build GRCh38
rs908719160 Chr.5: 179798537-179798537 on Build GRCh38
rs530090723 Chr.5: 179798438-179798438 on Build GRCh38
rs770504411 Chr.5: 179798321-179798321 on Build GRCh38
rs576648308 Chr.5: 179798168-179798168 on Build GRCh38
rs573493750 Chr.5: 179798366-179798366 on Build GRCh38
rs777106337 Chr.5: 179798339-179798339 on Build GRCh38
rs745351353 Chr.5: 179798390-179798390 on Build GRCh38
rs780215914 Chr.5: 179798204-179798204 on Build GRCh38
rs1021742130 Chr.5: 179798432-179798432 on Build GRCh38
rs544679540 Chr.5: 179798409-179798409 on Build GRCh38
rs771494011 Chr.5: 179798505-179798505 on Build GRCh38
rs535125650 Chr.5: 179798317-179798317 on Build GRCh38
rs143182365 Chr.5: 179798249-179798249 on Build GRCh38
rs767602330 Chr.5: 179798373-179798373 on Build GRCh38
rs917239218 Chr.5: 179798512-179798512 on Build GRCh38
rs764456989 Chr.5: 179798260-179798260 on Build GRCh38
rs537403321 Chr.5: 179798392-179798392 on Build GRCh38
rs778521452 Chr.5: 179798388-179798388 on Build GRCh38
rs756855303 Chr.5: 179798387-179798387 on Build GRCh38
rs894553421 Chr.5: 179798111-179798111 on Build GRCh38
rs757836393 Chr.5: 179798475-179798475 on Build GRCh38
rs757316494 Chr.5: 179798453-179798453 on Build GRCh38
rs754317194 Chr.5: 179798153-179798153 on Build GRCh38
rs756503298 Chr.5: 179798497-179798497 on Build GRCh38
rs763411349 Chr.5: 179798408-179798408 on Build GRCh38
rs138931499 Chr.5: 179798254-179798254 on Build GRCh38
rs766644214 Chr.5: 179798286-179798286 on Build GRCh38
rs748051364 Chr.5: 179798224-179798224 on Build GRCh38
rs746872303 Chr.5: 179798207-179798207 on Build GRCh38
rs759529045 Chr.5: 179798413-179798413 on Build GRCh38
rs764570093 Chr.5: 179798146-179798146 on Build GRCh38
rs1046319774 Chr.5: 179798098-179798098 on Build GRCh38
rs772898504 Chr.5: 179798233-179798233 on Build GRCh38
rs765316503 Chr.5: 179798173-179798173 on Build GRCh38
rs1008799936 Chr.5: 179798401-179798401 on Build GRCh38
rs139945722 Chr.5: 179798434-179798434 on Build GRCh38
rs199546175 Chr.5: 179798383-179798383 on Build GRCh38
rs773714351 Chr.5: 179798407-179798407 on Build GRCh38
rs777847628 Chr.5: 179798310-179798310 on Build GRCh38
rs777031623 Chr.5: 179798273-179798273 on Build GRCh38
rs377385523 Chr.5: 179798333-179798333 on Build GRCh38
rs775809340 Chr.5: 179798516-179798516 on Build GRCh38
rs776039475 Chr.5: 179798138-179798138 on Build GRCh38
rs17854722 Chr.5: 179798386-179798386 on Build GRCh38

View Full Product Details