Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180118529-180119027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGEF1C
Entrez Gene ID
255426
Gene Name
RasGEF domain family member 1C
Gene Aliases
-
UniGene
Hs.190559
Gene Chromosome Location
Chr.5: 180100795-180209130 on Build GRCh38
Transcripts
NM_175062.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682553 c.1045G>A p.A349T Chr.5: 180118647-180118647 on Build GRCh38
3994249 c.967G>A p.A323T Chr.5: 180118807-180118807 on Build GRCh38
4978071 c.991C>T p.Q331* Chr.5: 180118701-180118701 on Build GRCh38
3012123 c.910G>A p.G304S Chr.5: 180118864-180118864 on Build GRCh38
5496508 c.1083+8G>A p.? Chr.5: 180118601-180118601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981855082 Chr.5: 180118996-180118996 on Build GRCh38
rs916283817 Chr.5: 180118977-180118977 on Build GRCh38
rs996218696 Chr.5: 180118738-180118738 on Build GRCh38
rs146808625 Chr.5: 180118653-180118653 on Build GRCh38
rs949036403 Chr.5: 180118995-180118995 on Build GRCh38
rs114180160 Chr.5: 180119005-180119005 on Build GRCh38
rs374277989 Chr.5: 180118921-180118921 on Build GRCh38
rs977924506 Chr.5: 180118944-180118944 on Build GRCh38
rs62405054 Chr.5: 180118562-180118562 on Build GRCh38
rs767903141 Chr.5: 180118769-180118769 on Build GRCh38
rs936149962 Chr.5: 180118871-180118871 on Build GRCh38
rs140960830 Chr.5: 180118800-180118800 on Build GRCh38
rs759350273 Chr.5: 180118579-180118579 on Build GRCh38
rs758380429 Chr.5: 180118625-180118626 on Build GRCh38
rs765366182 Chr.5: 180118814-180118814 on Build GRCh38
rs144869075 Chr.5: 180118722-180118722 on Build GRCh38
rs376949886 Chr.5: 180118809-180118809 on Build GRCh38
rs760031898 Chr.5: 180118641-180118641 on Build GRCh38
rs138202063 Chr.5: 180118790-180118790 on Build GRCh38
rs376536496 Chr.5: 180118630-180118630 on Build GRCh38
rs756851704 Chr.5: 180118895-180118895 on Build GRCh38
rs758671647 Chr.5: 180119001-180119001 on Build GRCh38
rs1023311379 Chr.5: 180118793-180118793 on Build GRCh38
rs773505140 Chr.5: 180118827-180118827 on Build GRCh38
rs532002423 Chr.5: 180118931-180118931 on Build GRCh38
rs763228959 Chr.5: 180118650-180118650 on Build GRCh38
rs751308865 Chr.5: 180118852-180118852 on Build GRCh38
rs1038372749 Chr.5: 180118803-180118803 on Build GRCh38
rs568718625 Chr.5: 180118907-180118907 on Build GRCh38
rs779488171 Chr.5: 180118612-180118612 on Build GRCh38
rs768340365 Chr.5: 180118580-180118580 on Build GRCh38
rs569708520 Chr.5: 180118865-180118865 on Build GRCh38
rs140595237 Chr.5: 180118677-180118677 on Build GRCh38
rs559248645 Chr.5: 180118791-180118791 on Build GRCh38
rs370547572 Chr.5: 180118587-180118587 on Build GRCh38
rs746879303 Chr.5: 180118685-180118685 on Build GRCh38
rs772499093 Chr.5: 180118728-180118728 on Build GRCh38
rs747354084 Chr.5: 180118904-180118904 on Build GRCh38
rs751585560 Chr.5: 180118688-180118703 on Build GRCh38
rs554007078 Chr.5: 180118687-180118687 on Build GRCh38
rs545722886 Chr.5: 180118654-180118654 on Build GRCh38
rs777452980 Chr.5: 180118773-180118773 on Build GRCh38
rs764270377 Chr.5: 180118747-180118747 on Build GRCh38
rs574978411 Chr.5: 180118643-180118643 on Build GRCh38
rs747647226 Chr.5: 180118768-180118768 on Build GRCh38
rs758242021 Chr.5: 180118758-180118758 on Build GRCh38
rs780009147 Chr.5: 180118684-180118684 on Build GRCh38
rs773614037 Chr.5: 180118583-180118583 on Build GRCh38
rs926124297 Chr.5: 180118676-180118676 on Build GRCh38
rs200460338 Chr.5: 180118601-180118601 on Build GRCh38
rs758316782 Chr.5: 180118682-180118682 on Build GRCh38
rs576077247 Chr.5: 180118957-180118957 on Build GRCh38
rs200390928 Chr.5: 180118887-180118887 on Build GRCh38
rs138948699 Chr.5: 180118864-180118864 on Build GRCh38
rs746926298 Chr.5: 180118627-180118627 on Build GRCh38
rs932015074 Chr.5: 180118931-180118931 on Build GRCh38
rs765186468 Chr.5: 180118655-180118655 on Build GRCh38
rs201318370 Chr.5: 180118602-180118602 on Build GRCh38
rs752249143 Chr.5: 180118877-180118877 on Build GRCh38
rs907601944 Chr.5: 180118999-180118999 on Build GRCh38
rs936333940 Chr.5: 180118568-180118568 on Build GRCh38
rs746253194 Chr.5: 180118909-180118909 on Build GRCh38
rs781772374 Chr.5: 180118908-180118908 on Build GRCh38
rs764285338 Chr.5: 180118672-180118674 on Build GRCh38
rs765288627 Chr.5: 180118578-180118579 on Build GRCh38
rs755654807 Chr.5: 180118883-180118883 on Build GRCh38
rs954735332 Chr.5: 180118674-180118674 on Build GRCh38
rs1032692301 Chr.5: 180118945-180118945 on Build GRCh38
rs780748587 Chr.5: 180118764-180118764 on Build GRCh38
rs780521475 Chr.5: 180118900-180118900 on Build GRCh38
rs371834834 Chr.5: 180118604-180118604 on Build GRCh38
rs763271495 Chr.5: 180118584-180118584 on Build GRCh38
rs776083514 Chr.5: 180118729-180118729 on Build GRCh38
rs772040968 Chr.5: 180118611-180118611 on Build GRCh38
rs763028564 Chr.5: 180118829-180118829 on Build GRCh38
rs201116631 Chr.5: 180118647-180118647 on Build GRCh38
rs535737868 Chr.5: 180118913-180118913 on Build GRCh38
rs990901372 Chr.5: 180118967-180118967 on Build GRCh38
rs766203532 Chr.5: 180118847-180118847 on Build GRCh38
rs77441079 Chr.5: 180118947-180118947 on Build GRCh38
rs372297909 Chr.5: 180118592-180118592 on Build GRCh38
rs766676980 Chr.5: 180118928-180118928 on Build GRCh38
rs539557701 Chr.5: 180118940-180118940 on Build GRCh38
rs369825038 Chr.5: 180118727-180118727 on Build GRCh38
rs556805624 Chr.5: 180118640-180118640 on Build GRCh38
rs200174941 Chr.5: 180118718-180118718 on Build GRCh38
rs925213141 Chr.5: 180118739-180118739 on Build GRCh38
rs760310022 Chr.5: 180118573-180118573 on Build GRCh38
rs149114740 Chr.5: 180118808-180118808 on Build GRCh38
rs1040313113 Chr.5: 180118925-180118925 on Build GRCh38
rs755217508 Chr.5: 180119003-180119003 on Build GRCh38
rs376436919 Chr.5: 180118754-180118754 on Build GRCh38
rs1032494507 Chr.5: 180119001-180119001 on Build GRCh38
rs562013738 Chr.5: 180118762-180118762 on Build GRCh38
rs550233163 Chr.5: 180118888-180118888 on Build GRCh38
rs373002378 Chr.5: 180118751-180118751 on Build GRCh38
rs781214094 Chr.5: 180118908-180118909 on Build GRCh38
rs778619612 Chr.5: 180119001-180119002 on Build GRCh38
rs745748475 Chr.5: 180118561-180118561 on Build GRCh38
rs756351614 Chr.5: 180118594-180118594 on Build GRCh38
rs144013368 Chr.5: 180118593-180118593 on Build GRCh38
rs1013519486 Chr.5: 180118972-180118972 on Build GRCh38
rs900570931 Chr.5: 180118736-180118736 on Build GRCh38
rs369386547 Chr.5: 180118631-180118631 on Build GRCh38
rs752642323 Chr.5: 180118608-180118628 on Build GRCh38
rs896367141 Chr.5: 180118952-180118953 on Build GRCh38
rs991801524 Chr.5: 180118778-180118778 on Build GRCh38
rs572261747 Chr.5: 180118934-180118934 on Build GRCh38
rs777086782 Chr.5: 180118886-180118886 on Build GRCh38
rs202079156 Chr.5: 180118763-180118763 on Build GRCh38
rs564145629 Chr.5: 180118753-180118753 on Build GRCh38
rs1023594348 Chr.5: 180118975-180118975 on Build GRCh38
rs761026839 Chr.5: 180118734-180118734 on Build GRCh38
rs757503598 Chr.5: 180118717-180118718 on Build GRCh38
rs371026256 Chr.5: 180118755-180118755 on Build GRCh38
rs778122623 Chr.5: 180118597-180118597 on Build GRCh38
rs764217466 Chr.5: 180118649-180118649 on Build GRCh38
rs201724936 Chr.5: 180118648-180118648 on Build GRCh38
rs776576455 Chr.5: 180118560-180118560 on Build GRCh38
rs775373995 Chr.5: 180118616-180118616 on Build GRCh38
rs780624297 Chr.5: 180118708-180118708 on Build GRCh38
rs980228666 Chr.5: 180118664-180118664 on Build GRCh38
rs143395580 Chr.5: 180118632-180118632 on Build GRCh38
rs145725435 Chr.5: 180118688-180118688 on Build GRCh38
rs748855721 Chr.5: 180118774-180118774 on Build GRCh38

View Full Product Details