Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180301011-180301510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
UniGene
Hs.696497
Gene Chromosome Location
Chr.5: 180300690-180353387 on Build GRCh38
Transcripts
NM_005110.3 XM_006714942.2 XM_017010100.1

SNP Detail

SNP ID Chromosome Location
rs1016791517 Chr.5: 180301316-180301316 on Build GRCh38
rs1020976348 Chr.5: 180301300-180301300 on Build GRCh38
rs866058267 Chr.5: 180301068-180301068 on Build GRCh38
rs537580048 Chr.5: 180301209-180301209 on Build GRCh38
rs574310035 Chr.5: 180301268-180301268 on Build GRCh38
rs909765072 Chr.5: 180301135-180301135 on Build GRCh38
rs949723671 Chr.5: 180301399-180301399 on Build GRCh38
rs1011571299 Chr.5: 180301294-180301294 on Build GRCh38
rs985227852 Chr.5: 180301342-180301342 on Build GRCh38
rs369204312 Chr.5: 180301216-180301216 on Build GRCh38
rs913575023 Chr.5: 180301163-180301163 on Build GRCh38
rs577128473 Chr.5: 180301369-180301369 on Build GRCh38
rs184546685 Chr.5: 180301457-180301457 on Build GRCh38
rs568606250 Chr.5: 180301131-180301131 on Build GRCh38
rs538262053 Chr.5: 180301093-180301093 on Build GRCh38
rs998292461 Chr.5: 180301307-180301307 on Build GRCh38
rs776903720 Chr.5: 180301429-180301429 on Build GRCh38
rs542632245 Chr.5: 180301215-180301215 on Build GRCh38
rs1006220446 Chr.5: 180301269-180301269 on Build GRCh38
rs973043067 Chr.5: 180301387-180301387 on Build GRCh38
rs967097425 Chr.5: 180301302-180301302 on Build GRCh38
rs138920932 Chr.5: 180301458-180301458 on Build GRCh38
rs374880930 Chr.5: 180301310-180301310 on Build GRCh38
rs558168688 Chr.5: 180301112-180301112 on Build GRCh38
rs531822885 Chr.5: 180301449-180301449 on Build GRCh38
rs147231426 Chr.5: 180301368-180301368 on Build GRCh38
rs559730979 Chr.5: 180301348-180301348 on Build GRCh38
rs1045504365 Chr.5: 180301406-180301406 on Build GRCh38
rs932456633 Chr.5: 180301085-180301085 on Build GRCh38
rs894095877 Chr.5: 180301231-180301232 on Build GRCh38
rs922488504 Chr.5: 180301064-180301064 on Build GRCh38
rs899771432 Chr.5: 180301318-180301318 on Build GRCh38
rs963845994 Chr.5: 180301073-180301073 on Build GRCh38
rs59738929 Chr.5: 180301328-180301328 on Build GRCh38
rs926995521 Chr.5: 180301407-180301407 on Build GRCh38
rs878968148 Chr.5: 180301125-180301125 on Build GRCh38
rs979120463 Chr.5: 180301440-180301440 on Build GRCh38
rs143370482 Chr.5: 180301127-180301127 on Build GRCh38
rs767402898 Chr.5: 180301184-180301184 on Build GRCh38
rs979678057 Chr.5: 180301167-180301167 on Build GRCh38
rs1033938491 Chr.5: 180301226-180301226 on Build GRCh38
rs926886410 Chr.5: 180301173-180301173 on Build GRCh38
rs1045878868 Chr.5: 180301210-180301210 on Build GRCh38
rs941041522 Chr.5: 180301146-180301146 on Build GRCh38
rs918288335 Chr.5: 180301398-180301398 on Build GRCh38
rs938148556 Chr.5: 180301183-180301183 on Build GRCh38
rs946398878 Chr.5: 180301166-180301166 on Build GRCh38
rs988171168 Chr.5: 180301149-180301149 on Build GRCh38
rs1036735838 Chr.5: 180301158-180301158 on Build GRCh38
rs760410564 Chr.5: 180301148-180301149 on Build GRCh38
rs562918481 Chr.5: 180301428-180301428 on Build GRCh38
rs545869943 Chr.5: 180301410-180301410 on Build GRCh38
rs1029498170 Chr.5: 180301338-180301338 on Build GRCh38
rs896811933 Chr.5: 180301159-180301159 on Build GRCh38
rs975298559 Chr.5: 180301120-180301120 on Build GRCh38
rs1048160509 Chr.5: 180301223-180301223 on Build GRCh38

View Full Product Details