Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180850929-180851432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP62
Entrez Gene ID
643836
Gene Name
ZFP62 zinc finger protein
Gene Aliases
ZET, ZNF755
UniGene
Hs.509227
Gene Chromosome Location
Chr.5: 180843580-180861327 on Build GRCh38
Transcripts
NM_001172638.1 NM_152283.4 XM_017009710.1 XM_017009711.1 XM_017009712.1 XM_017009713.1 XM_017009714.1 XM_017009715.1 XM_017009716.1 XM_017009717.1 XM_017009718.1 XM_017009719.1 XM_017009720.1 XM_017009721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1233408 c.317A>C p.K106T Chr.5: 180851079-180851079 on Build GRCh38
7275821 c.23A>G p.D8G Chr.5: 180851373-180851373 on Build GRCh38
5682103 c.15G>A p.K5K Chr.5: 180851381-180851381 on Build GRCh38
7398561 c.297G>C p.Q99H Chr.5: 180851099-180851099 on Build GRCh38
4931713 c.296A>T p.Q99L Chr.5: 180851100-180851100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044005466 Chr.5: 180851120-180851120 on Build GRCh38
rs764983847 Chr.5: 180851230-180851230 on Build GRCh38
rs963255266 Chr.5: 180851247-180851247 on Build GRCh38
rs757752723 Chr.5: 180851207-180851207 on Build GRCh38
rs905430611 Chr.5: 180851140-180851140 on Build GRCh38
rs771081745 Chr.5: 180851086-180851087 on Build GRCh38
rs765194069 Chr.5: 180851031-180851031 on Build GRCh38
rs766006939 Chr.5: 180851330-180851330 on Build GRCh38
rs563545184 Chr.5: 180851253-180851253 on Build GRCh38
rs769681813 Chr.5: 180851307-180851307 on Build GRCh38
rs192511596 Chr.5: 180851254-180851254 on Build GRCh38
rs142442586 Chr.5: 180851027-180851027 on Build GRCh38
rs773200063 Chr.5: 180851314-180851314 on Build GRCh38
rs776454755 Chr.5: 180851289-180851289 on Build GRCh38
rs773797830 Chr.5: 180850960-180850960 on Build GRCh38
rs151304370 Chr.5: 180851088-180851088 on Build GRCh38
rs949374798 Chr.5: 180851044-180851044 on Build GRCh38
rs561460081 Chr.5: 180851357-180851357 on Build GRCh38
rs561822206 Chr.5: 180851395-180851395 on Build GRCh38
rs747170293 Chr.5: 180851003-180851003 on Build GRCh38
rs1040103205 Chr.5: 180851067-180851067 on Build GRCh38
rs754625126 Chr.5: 180851113-180851113 on Build GRCh38
rs1042677060 Chr.5: 180851159-180851159 on Build GRCh38
rs754220224 Chr.5: 180851196-180851196 on Build GRCh38
rs1010211762 Chr.5: 180851143-180851143 on Build GRCh38
rs892605100 Chr.5: 180851094-180851094 on Build GRCh38
rs368580289 Chr.5: 180851004-180851004 on Build GRCh38
rs867443966 Chr.5: 180851202-180851202 on Build GRCh38
rs140539381 Chr.5: 180851142-180851142 on Build GRCh38
rs545456906 Chr.5: 180851240-180851240 on Build GRCh38
rs964984660 Chr.5: 180851385-180851385 on Build GRCh38
rs140265621 Chr.5: 180851359-180851359 on Build GRCh38
rs746000114 Chr.5: 180851213-180851213 on Build GRCh38
rs141026256 Chr.5: 180850992-180850992 on Build GRCh38
rs373641672 Chr.5: 180851226-180851226 on Build GRCh38
rs199942165 Chr.5: 180851091-180851091 on Build GRCh38
rs146692011 Chr.5: 180851222-180851222 on Build GRCh38
rs762550416 Chr.5: 180851281-180851281 on Build GRCh38
rs970658558 Chr.5: 180851261-180851261 on Build GRCh38
rs34392747 Chr.5: 180851289-180851290 on Build GRCh38
rs747957192 Chr.5: 180851291-180851291 on Build GRCh38
rs946987642 Chr.5: 180851117-180851117 on Build GRCh38
rs901456143 Chr.5: 180851161-180851162 on Build GRCh38
rs776877201 Chr.5: 180851382-180851383 on Build GRCh38
rs762738991 Chr.5: 180851326-180851326 on Build GRCh38
rs778900307 Chr.5: 180850953-180850953 on Build GRCh38
rs759298827 Chr.5: 180851377-180851377 on Build GRCh38
rs745937989 Chr.5: 180850959-180850959 on Build GRCh38
rs767255924 Chr.5: 180851378-180851378 on Build GRCh38
rs705441 Chr.5: 180851393-180851393 on Build GRCh38
rs772650474 Chr.5: 180851071-180851071 on Build GRCh38
rs971900727 Chr.5: 180851250-180851250 on Build GRCh38
rs150126330 Chr.5: 180851209-180851209 on Build GRCh38
rs570135527 Chr.5: 180851021-180851021 on Build GRCh38
rs771989179 Chr.5: 180850949-180850949 on Build GRCh38
rs762513973 Chr.5: 180851082-180851082 on Build GRCh38

View Full Product Details