Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181045200-181045694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_001308245.1 NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_011534444.2 XM_017009113.1 XM_017009114.1 XM_017009115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7302004 c.64T>A p.F22I Chr.5: 181045553-181045553 on Build GRCh38
4003488 c.24A>G p.P8P Chr.5: 181045513-181045513 on Build GRCh38
7302006 c.64T>A p.F22I Chr.5: 181045553-181045553 on Build GRCh38
4003485 c.9C>T p.D3D Chr.5: 181045498-181045498 on Build GRCh38
3941283 c.44C>T p.S15L Chr.5: 181045533-181045533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013747970 Chr.5: 181045361-181045361 on Build GRCh38
rs775890914 Chr.5: 181045519-181045519 on Build GRCh38
rs139601076 Chr.5: 181045443-181045443 on Build GRCh38
rs541887791 Chr.5: 181045472-181045472 on Build GRCh38
rs1001137939 Chr.5: 181045317-181045317 on Build GRCh38
rs1042398467 Chr.5: 181045302-181045302 on Build GRCh38
rs6894087 Chr.5: 181045513-181045513 on Build GRCh38
rs763046355 Chr.5: 181045537-181045537 on Build GRCh38
rs747367645 Chr.5: 181045561-181045561 on Build GRCh38
rs201027444 Chr.5: 181045558-181045558 on Build GRCh38
rs1055302785 Chr.5: 181045522-181045522 on Build GRCh38
rs553821930 Chr.5: 181045284-181045284 on Build GRCh38
rs370543834 Chr.5: 181045671-181045671 on Build GRCh38
rs372675288 Chr.5: 181045484-181045484 on Build GRCh38
rs756297675 Chr.5: 181045528-181045529 on Build GRCh38
rs776624538 Chr.5: 181045482-181045482 on Build GRCh38
rs766845674 Chr.5: 181045638-181045638 on Build GRCh38
rs74668793 Chr.5: 181045405-181045405 on Build GRCh38
rs76366126 Chr.5: 181045420-181045420 on Build GRCh38
rs199807644 Chr.5: 181045602-181045602 on Build GRCh38
rs371450738 Chr.5: 181045554-181045554 on Build GRCh38
rs1018580001 Chr.5: 181045364-181045364 on Build GRCh38
rs961832767 Chr.5: 181045592-181045592 on Build GRCh38
rs184327346 Chr.5: 181045670-181045670 on Build GRCh38
rs767747403 Chr.5: 181045646-181045646 on Build GRCh38
rs996283533 Chr.5: 181045578-181045578 on Build GRCh38
rs755563388 Chr.5: 181045641-181045641 on Build GRCh38
rs759271300 Chr.5: 181045583-181045583 on Build GRCh38
rs773479898 Chr.5: 181045572-181045572 on Build GRCh38
rs758285569 Chr.5: 181045486-181045486 on Build GRCh38
rs369441917 Chr.5: 181045621-181045621 on Build GRCh38
rs1041191790 Chr.5: 181045656-181045656 on Build GRCh38
rs576870904 Chr.5: 181045312-181045312 on Build GRCh38
rs113682037 Chr.5: 181045626-181045626 on Build GRCh38
rs144716927 Chr.5: 181045563-181045563 on Build GRCh38
rs749408906 Chr.5: 181045623-181045623 on Build GRCh38
rs769800106 Chr.5: 181045531-181045531 on Build GRCh38
rs541056452 Chr.5: 181045499-181045499 on Build GRCh38
rs951823697 Chr.5: 181045421-181045421 on Build GRCh38
rs111777066 Chr.5: 181045438-181045438 on Build GRCh38
rs771760064 Chr.5: 181045632-181045632 on Build GRCh38
rs376394422 Chr.5: 181045601-181045601 on Build GRCh38
rs956975502 Chr.5: 181045338-181045338 on Build GRCh38
rs368802401 Chr.5: 181045451-181045451 on Build GRCh38
rs945360669 Chr.5: 181045288-181045288 on Build GRCh38
rs868558757 Chr.5: 181045465-181045465 on Build GRCh38
rs933167429 Chr.5: 181045243-181045243 on Build GRCh38
rs201955770 Chr.5: 181045549-181045549 on Build GRCh38
rs780258641 Chr.5: 181045583-181045585 on Build GRCh38
rs772421635 Chr.5: 181045517-181045517 on Build GRCh38
rs553488243 Chr.5: 181045648-181045648 on Build GRCh38
rs900093347 Chr.5: 181045562-181045562 on Build GRCh38
rs1024675083 Chr.5: 181045371-181045371 on Build GRCh38
rs781287474 Chr.5: 181045564-181045564 on Build GRCh38
rs553994701 Chr.5: 181045397-181045397 on Build GRCh38
rs755148732 Chr.5: 181045646-181045647 on Build GRCh38
rs775404752 Chr.5: 181045634-181045634 on Build GRCh38
rs950480906 Chr.5: 181045381-181045381 on Build GRCh38
rs748196562 Chr.5: 181045565-181045565 on Build GRCh38
rs376133595 Chr.5: 181045404-181045404 on Build GRCh38
rs965650332 Chr.5: 181045400-181045400 on Build GRCh38
rs750916979 Chr.5: 181045553-181045553 on Build GRCh38
rs752009254 Chr.5: 181045639-181045639 on Build GRCh38
rs749578991 Chr.5: 181045585-181045586 on Build GRCh38
rs763274407 Chr.5: 181045239-181045239 on Build GRCh38
rs200387419 Chr.5: 181045591-181045591 on Build GRCh38
rs376351605 Chr.5: 181045636-181045636 on Build GRCh38
rs28677846 Chr.5: 181045498-181045498 on Build GRCh38
rs767139790 Chr.5: 181045585-181045585 on Build GRCh38
rs145827131 Chr.5: 181045512-181045512 on Build GRCh38
rs1031156367 Chr.5: 181045320-181045320 on Build GRCh38
rs369677072 Chr.5: 181045650-181045650 on Build GRCh38
rs771388236 Chr.5: 181045444-181045444 on Build GRCh38
rs1008411256 Chr.5: 181045353-181045353 on Build GRCh38
rs780727852 Chr.5: 181045507-181045507 on Build GRCh38
rs772233736 Chr.5: 181045456-181045456 on Build GRCh38
rs568029331 Chr.5: 181045267-181045267 on Build GRCh38
rs74555255 Chr.5: 181045448-181045448 on Build GRCh38
rs761824020 Chr.5: 181045528-181045528 on Build GRCh38
rs375394652 Chr.5: 181045586-181045586 on Build GRCh38
rs963595198 Chr.5: 181045435-181045435 on Build GRCh38
rs1052491012 Chr.5: 181045640-181045640 on Build GRCh38
rs904079911 Chr.5: 181045306-181045306 on Build GRCh38
rs775702530 Chr.5: 181045463-181045463 on Build GRCh38
rs903963845 Chr.5: 181045471-181045471 on Build GRCh38
rs1055058836 Chr.5: 181045653-181045653 on Build GRCh38
rs398093927 Chr.5: 181045424-181045424 on Build GRCh38
rs997081979 Chr.5: 181045663-181045663 on Build GRCh38
rs1051162514 Chr.5: 181045264-181045264 on Build GRCh38
rs755306047 Chr.5: 181045599-181045599 on Build GRCh38
rs184114719 Chr.5: 181045439-181045439 on Build GRCh38
rs1028647688 Chr.5: 181045567-181045567 on Build GRCh38
rs201472333 Chr.5: 181045533-181045533 on Build GRCh38
rs200561150 Chr.5: 181045485-181045485 on Build GRCh38
rs188720349 Chr.5: 181045526-181045526 on Build GRCh38
rs773291591 Chr.5: 181045534-181045534 on Build GRCh38
rs974957012 Chr.5: 181045227-181045227 on Build GRCh38
rs759193687 Chr.5: 181045540-181045540 on Build GRCh38
rs1034376348 Chr.5: 181045319-181045319 on Build GRCh38
rs755592532 Chr.5: 181045544-181045544 on Build GRCh38
rs760391517 Chr.5: 181045658-181045658 on Build GRCh38
rs751895917 Chr.5: 181045598-181045598 on Build GRCh38
rs760290066 Chr.5: 181045635-181045635 on Build GRCh38
rs752201567 Chr.5: 181045543-181045543 on Build GRCh38
rs983481475 Chr.5: 181045396-181045396 on Build GRCh38
rs746173464 Chr.5: 181045516-181045516 on Build GRCh38
rs202143567 Chr.5: 181045539-181045539 on Build GRCh38
rs771029730 Chr.5: 181045580-181045580 on Build GRCh38
rs111396549 Chr.5: 181045398-181045398 on Build GRCh38
rs766059030 Chr.5: 181045487-181045487 on Build GRCh38
rs774154303 Chr.5: 181045581-181045581 on Build GRCh38
rs201199198 Chr.5: 181045573-181045573 on Build GRCh38
rs777966251 Chr.5: 181045607-181045607 on Build GRCh38
rs921773250 Chr.5: 181045228-181045228 on Build GRCh38
rs1046518407 Chr.5: 181045669-181045669 on Build GRCh38
rs138527915 Chr.5: 181045570-181045570 on Build GRCh38
rs770185561 Chr.5: 181045604-181045604 on Build GRCh38
rs779325563 Chr.5: 181045550-181045550 on Build GRCh38
rs900114661 Chr.5: 181045661-181045661 on Build GRCh38
rs760874768 Chr.5: 181045466-181045466 on Build GRCh38
rs985051078 Chr.5: 181045425-181045425 on Build GRCh38
rs1045516035 Chr.5: 181045255-181045255 on Build GRCh38
rs1029096414 Chr.5: 181045665-181045665 on Build GRCh38
rs533558911 Chr.5: 181045334-181045334 on Build GRCh38
rs747234210 Chr.5: 181045520-181045520 on Build GRCh38
rs1045065211 Chr.5: 181045449-181045449 on Build GRCh38
rs373481901 Chr.5: 181045627-181045627 on Build GRCh38
rs780562974 Chr.5: 181045594-181045594 on Build GRCh38
rs949653658 Chr.5: 181045668-181045668 on Build GRCh38
rs369415772 Chr.5: 181045590-181045590 on Build GRCh38

View Full Product Details