Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 14135428-14135926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD83
Entrez Gene ID
9308
Gene Name
CD83 molecule
Gene Aliases
BL11, HB15
UniGene
Hs.595133
Gene Chromosome Location
Chr.6: 14117256-14136918 on Build GRCh38
Transcripts
NM_001040280.1 NM_001251901.1 NM_004233.3

SNP Detail

SNP ID Chromosome Location
rs374372249 Chr.6: 14135746-14135746 on Build GRCh38
rs917613619 Chr.6: 14135547-14135547 on Build GRCh38
rs144093044 Chr.6: 14135708-14135708 on Build GRCh38
rs555062784 Chr.6: 14135586-14135586 on Build GRCh38
rs1024595084 Chr.6: 14135808-14135808 on Build GRCh38
rs548894139 Chr.6: 14135461-14135461 on Build GRCh38
rs921221961 Chr.6: 14135550-14135550 on Build GRCh38
rs538871670 Chr.6: 14135530-14135530 on Build GRCh38
rs982945408 Chr.6: 14135814-14135814 on Build GRCh38
rs951524803 Chr.6: 14135454-14135454 on Build GRCh38
rs956377592 Chr.6: 14135472-14135472 on Build GRCh38
rs559405851 Chr.6: 14135580-14135580 on Build GRCh38
rs1001127342 Chr.6: 14135742-14135742 on Build GRCh38
rs980454615 Chr.6: 14135845-14135845 on Build GRCh38
rs146007792 Chr.6: 14135760-14135760 on Build GRCh38
rs543876843 Chr.6: 14135679-14135679 on Build GRCh38
rs746267828 Chr.6: 14135836-14135836 on Build GRCh38
rs569986737 Chr.6: 14135475-14135475 on Build GRCh38
rs537417005 Chr.6: 14135683-14135683 on Build GRCh38
rs951119926 Chr.6: 14135799-14135799 on Build GRCh38
rs370058890 Chr.6: 14135731-14135731 on Build GRCh38
rs1027119090 Chr.6: 14135449-14135449 on Build GRCh38
rs940974902 Chr.6: 14135594-14135594 on Build GRCh38
rs552769541 Chr.6: 14135558-14135558 on Build GRCh38
rs574891772 Chr.6: 14135680-14135680 on Build GRCh38
rs975477399 Chr.6: 14135518-14135518 on Build GRCh38
rs965658775 Chr.6: 14135885-14135885 on Build GRCh38
rs1026714396 Chr.6: 14135774-14135774 on Build GRCh38
rs367694662 Chr.6: 14135572-14135572 on Build GRCh38
rs1049288212 Chr.6: 14135584-14135584 on Build GRCh38
rs1019735813 Chr.6: 14135817-14135817 on Build GRCh38
rs1043136095 Chr.6: 14135565-14135565 on Build GRCh38
rs1023302898 Chr.6: 14135688-14135688 on Build GRCh38
rs1040384752 Chr.6: 14135648-14135649 on Build GRCh38
rs992072593 Chr.6: 14135783-14135783 on Build GRCh38
rs376308924 Chr.6: 14135569-14135569 on Build GRCh38
rs1029082241 Chr.6: 14135691-14135691 on Build GRCh38
rs768443002 Chr.6: 14135800-14135800 on Build GRCh38
rs982894630 Chr.6: 14135455-14135455 on Build GRCh38
rs755912096 Chr.6: 14135473-14135473 on Build GRCh38

View Full Product Details