Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 19839086-19839583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ID4
Entrez Gene ID
3400
Gene Name
inhibitor of DNA binding 4, HLH protein
Gene Aliases
IDB4, bHLHb27
UniGene
Hs.519601
Gene Chromosome Location
Chr.6: 19837370-19842200 on Build GRCh38
Transcripts
NM_001546.3

SNP Detail

SNP ID Chromosome Location
rs1028929737 Chr.6: 19839516-19839516 on Build GRCh38
rs1010671115 Chr.6: 19839373-19839373 on Build GRCh38
rs1030428097 Chr.6: 19839186-19839186 on Build GRCh38
rs72836085 Chr.6: 19839457-19839457 on Build GRCh38
rs146408427 Chr.6: 19839393-19839393 on Build GRCh38
rs143075248 Chr.6: 19839552-19839552 on Build GRCh38
rs890153211 Chr.6: 19839311-19839311 on Build GRCh38
rs41271301 Chr.6: 19839198-19839198 on Build GRCh38
rs184423744 Chr.6: 19839201-19839201 on Build GRCh38
rs189313667 Chr.6: 19839251-19839251 on Build GRCh38
rs911643954 Chr.6: 19839517-19839517 on Build GRCh38
rs890874239 Chr.6: 19839169-19839169 on Build GRCh38
rs921514440 Chr.6: 19839267-19839267 on Build GRCh38
rs578134981 Chr.6: 19839341-19839342 on Build GRCh38
rs934319470 Chr.6: 19839406-19839406 on Build GRCh38
rs974650668 Chr.6: 19839344-19839344 on Build GRCh38
rs115821285 Chr.6: 19839274-19839274 on Build GRCh38
rs756565949 Chr.6: 19839156-19839156 on Build GRCh38
rs200526011 Chr.6: 19839272-19839273 on Build GRCh38
rs41271299 Chr.6: 19839184-19839184 on Build GRCh38
rs1050904995 Chr.6: 19839168-19839168 on Build GRCh38
rs941473192 Chr.6: 19839257-19839257 on Build GRCh38
rs765651982 Chr.6: 19839120-19839120 on Build GRCh38
rs1802806 Chr.6: 19839410-19839410 on Build GRCh38
rs377628202 Chr.6: 19839330-19839330 on Build GRCh38
rs1015681059 Chr.6: 19839341-19839342 on Build GRCh38
rs1023914345 Chr.6: 19839380-19839380 on Build GRCh38
rs975587754 Chr.6: 19839454-19839454 on Build GRCh38
rs999426681 Chr.6: 19839192-19839192 on Build GRCh38
rs927123727 Chr.6: 19839428-19839428 on Build GRCh38
rs941613831 Chr.6: 19839128-19839128 on Build GRCh38
rs754476131 Chr.6: 19839375-19839378 on Build GRCh38
rs1023294489 Chr.6: 19839199-19839199 on Build GRCh38
rs1802807 Chr.6: 19839315-19839315 on Build GRCh38
rs957905662 Chr.6: 19839200-19839200 on Build GRCh38
rs72552347 Chr.6: 19839453-19839453 on Build GRCh38
rs531990076 Chr.6: 19839277-19839277 on Build GRCh38
rs571362838 Chr.6: 19839123-19839123 on Build GRCh38
rs912200989 Chr.6: 19839274-19839275 on Build GRCh38
rs896472011 Chr.6: 19839331-19839331 on Build GRCh38
rs983073630 Chr.6: 19839254-19839254 on Build GRCh38
rs140374613 Chr.6: 19839255-19839255 on Build GRCh38
rs900255026 Chr.6: 19839183-19839183 on Build GRCh38
rs956251563 Chr.6: 19839551-19839551 on Build GRCh38
rs8214 Chr.6: 19839548-19839548 on Build GRCh38
rs898589845 Chr.6: 19839467-19839467 on Build GRCh38
rs76847683 Chr.6: 19839343-19839343 on Build GRCh38
rs764321690 Chr.6: 19839171-19839171 on Build GRCh38
rs770411984 Chr.6: 19839429-19839429 on Build GRCh38
rs758031722 Chr.6: 19839339-19839339 on Build GRCh38
rs1052263938 Chr.6: 19839327-19839327 on Build GRCh38
rs995576007 Chr.6: 19839476-19839476 on Build GRCh38
rs754498148 Chr.6: 19839386-19839386 on Build GRCh38
rs528136321 Chr.6: 19839542-19839542 on Build GRCh38
rs930205779 Chr.6: 19839247-19839247 on Build GRCh38
rs931740912 Chr.6: 19839488-19839488 on Build GRCh38
rs9689909 Chr.6: 19839242-19839242 on Build GRCh38
rs778648357 Chr.6: 19839272-19839272 on Build GRCh38
rs1037418608 Chr.6: 19839179-19839179 on Build GRCh38
rs941760098 Chr.6: 19839178-19839178 on Build GRCh38
rs2819 Chr.6: 19839364-19839364 on Build GRCh38
rs951323830 Chr.6: 19839426-19839426 on Build GRCh38
rs564941466 Chr.6: 19839249-19839254 on Build GRCh38
rs1004071804 Chr.6: 19839392-19839392 on Build GRCh38
rs910105071 Chr.6: 19839438-19839438 on Build GRCh38
rs1040286425 Chr.6: 19839269-19839269 on Build GRCh38
rs187917612 Chr.6: 19839135-19839135 on Build GRCh38
rs374239156 Chr.6: 19839275-19839275 on Build GRCh38
rs947120686 Chr.6: 19839417-19839417 on Build GRCh38
rs72552348 Chr.6: 19839519-19839519 on Build GRCh38
rs551220382 Chr.6: 19839558-19839558 on Build GRCh38
rs988560211 Chr.6: 19839409-19839409 on Build GRCh38
rs1002761479 Chr.6: 19839316-19839319 on Build GRCh38
rs1051987438 Chr.6: 19839512-19839512 on Build GRCh38
rs777327799 Chr.6: 19839377-19839377 on Build GRCh38
rs890359321 Chr.6: 19839189-19839189 on Build GRCh38
rs1044196861 Chr.6: 19839313-19839313 on Build GRCh38
rs868438768 Chr.6: 19839424-19839424 on Build GRCh38
rs982117765 Chr.6: 19839237-19839237 on Build GRCh38
rs753752927 Chr.6: 19839232-19839232 on Build GRCh38
rs1008928321 Chr.6: 19839525-19839526 on Build GRCh38
rs766231470 Chr.6: 19839223-19839223 on Build GRCh38
rs930236834 Chr.6: 19839137-19839137 on Build GRCh38
rs893738474 Chr.6: 19839328-19839328 on Build GRCh38
rs1039010846 Chr.6: 19839133-19839133 on Build GRCh38
rs1036996840 Chr.6: 19839463-19839463 on Build GRCh38
rs544812817 Chr.6: 19839308-19839308 on Build GRCh38
rs749782821 Chr.6: 19839157-19839158 on Build GRCh38
rs146732020 Chr.6: 19839219-19839219 on Build GRCh38
rs914325928 Chr.6: 19839410-19839411 on Build GRCh38
rs1002616573 Chr.6: 19839228-19839228 on Build GRCh38
rs549892274 Chr.6: 19839222-19839222 on Build GRCh38
rs969487868 Chr.6: 19839381-19839381 on Build GRCh38
rs192526260 Chr.6: 19839352-19839352 on Build GRCh38
rs987047614 Chr.6: 19839378-19839379 on Build GRCh38
rs778572202 Chr.6: 19839451-19839451 on Build GRCh38
rs916564735 Chr.6: 19839119-19839119 on Build GRCh38

View Full Product Details