Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152348970-152349493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

SNP Detail

SNP ID Chromosome Location
rs531974136 Chr.1: 152349372-152349372 on Build GRCh38
rs560732476 Chr.1: 152349299-152349299 on Build GRCh38
rs572497947 Chr.1: 152349275-152349275 on Build GRCh38
rs1021457792 Chr.1: 152349197-152349201 on Build GRCh38
rs1011022856 Chr.1: 152349220-152349220 on Build GRCh38
rs374465443 Chr.1: 152349229-152349229 on Build GRCh38
rs959868565 Chr.1: 152349470-152349470 on Build GRCh38
rs939739134 Chr.1: 152349364-152349364 on Build GRCh38
rs955739278 Chr.1: 152349163-152349163 on Build GRCh38
rs778365753 Chr.1: 152349436-152349436 on Build GRCh38
rs909435238 Chr.1: 152349302-152349303 on Build GRCh38
rs549634514 Chr.1: 152349336-152349336 on Build GRCh38
rs576329994 Chr.1: 152349211-152349211 on Build GRCh38
rs982108831 Chr.1: 152349240-152349240 on Build GRCh38
rs763298870 Chr.1: 152349201-152349201 on Build GRCh38
rs574470163 Chr.1: 152349145-152349148 on Build GRCh38
rs894055540 Chr.1: 152349441-152349441 on Build GRCh38
rs932927670 Chr.1: 152349381-152349381 on Build GRCh38
rs115206579 Chr.1: 152349355-152349355 on Build GRCh38
rs1035406346 Chr.1: 152349247-152349247 on Build GRCh38
rs1035928841 Chr.1: 152349452-152349452 on Build GRCh38
rs1040714076 Chr.1: 152349055-152349055 on Build GRCh38
rs574412094 Chr.1: 152349020-152349020 on Build GRCh38
rs1028508649 Chr.1: 152349209-152349209 on Build GRCh38
rs901454711 Chr.1: 152349108-152349108 on Build GRCh38
rs900810909 Chr.1: 152349380-152349380 on Build GRCh38
rs151314663 Chr.1: 152349425-152349425 on Build GRCh38
rs928515835 Chr.1: 152349258-152349258 on Build GRCh38
rs992293535 Chr.1: 152349290-152349290 on Build GRCh38
rs1011053863 Chr.1: 152349443-152349443 on Build GRCh38
rs370463607 Chr.1: 152349110-152349110 on Build GRCh38
rs943665654 Chr.1: 152349326-152349326 on Build GRCh38
rs997477374 Chr.1: 152349145-152349145 on Build GRCh38
rs1011171525 Chr.1: 152349394-152349394 on Build GRCh38
rs969934972 Chr.1: 152349435-152349435 on Build GRCh38
rs540854553 Chr.1: 152349296-152349296 on Build GRCh38
rs764344877 Chr.1: 152349239-152349239 on Build GRCh38
rs766024568 Chr.1: 152349366-152349366 on Build GRCh38
rs989781991 Chr.1: 152349177-152349177 on Build GRCh38
rs560004314 Chr.1: 152349295-152349295 on Build GRCh38
rs1031752416 Chr.1: 152349068-152349068 on Build GRCh38
rs183767339 Chr.1: 152349036-152349036 on Build GRCh38
rs541755110 Chr.1: 152349187-152349187 on Build GRCh38
rs529951798 Chr.1: 152349327-152349327 on Build GRCh38
rs772710070 Chr.1: 152349266-152349266 on Build GRCh38
rs527488290 Chr.1: 152349300-152349300 on Build GRCh38
rs775661140 Chr.1: 152349189-152349189 on Build GRCh38
rs1017785385 Chr.1: 152349009-152349010 on Build GRCh38
rs747106367 Chr.1: 152349440-152349440 on Build GRCh38
rs552309626 Chr.1: 152349335-152349335 on Build GRCh38
rs565275936 Chr.1: 152349231-152349231 on Build GRCh38
rs919349621 Chr.1: 152349297-152349297 on Build GRCh38

View Full Product Details