Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152510616-152511114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCE5A
Entrez Gene ID
254910
Gene Name
late cornified envelope 5A
Gene Aliases
LEP18, SPRL5A
UniGene
Hs.516410
Gene Chromosome Location
Chr.1: 152510844-152512177 on Build GRCh38
Transcripts
NM_178438.4

SNP Detail

SNP ID Chromosome Location
rs938293183 Chr.1: 152510913-152510913 on Build GRCh38
rs967866563 Chr.1: 152510926-152510926 on Build GRCh38
rs917228290 Chr.1: 152511025-152511025 on Build GRCh38
rs900491579 Chr.1: 152511038-152511038 on Build GRCh38
rs548768738 Chr.1: 152510729-152510729 on Build GRCh38
rs563913127 Chr.1: 152510830-152510830 on Build GRCh38
rs992750951 Chr.1: 152510753-152510753 on Build GRCh38
rs910106614 Chr.1: 152511060-152511060 on Build GRCh38
rs1034401070 Chr.1: 152510974-152510974 on Build GRCh38
rs539856507 Chr.1: 152510812-152510812 on Build GRCh38
rs143267367 Chr.1: 152510757-152510757 on Build GRCh38
rs769889306 Chr.1: 152510724-152510724 on Build GRCh38
rs774235242 Chr.1: 152510738-152510738 on Build GRCh38
rs1018923585 Chr.1: 152510715-152510715 on Build GRCh38
rs73004848 Chr.1: 152510985-152510985 on Build GRCh38
rs1020722162 Chr.1: 152510803-152510803 on Build GRCh38
rs1022187127 Chr.1: 152510925-152510925 on Build GRCh38
rs1047255764 Chr.1: 152510762-152510762 on Build GRCh38
rs191054418 Chr.1: 152511081-152511081 on Build GRCh38
rs142434659 Chr.1: 152510678-152510678 on Build GRCh38
rs549237294 Chr.1: 152511066-152511066 on Build GRCh38
rs997204991 Chr.1: 152511047-152511047 on Build GRCh38
rs1039486641 Chr.1: 152510819-152510819 on Build GRCh38
rs868096319 Chr.1: 152510672-152510672 on Build GRCh38
rs1008860855 Chr.1: 152510686-152510686 on Build GRCh38
rs113870585 Chr.1: 152510761-152510761 on Build GRCh38
rs996762108 Chr.1: 152510737-152510737 on Build GRCh38
rs186435143 Chr.1: 152510804-152510804 on Build GRCh38
rs944773808 Chr.1: 152511024-152511024 on Build GRCh38
rs1022532810 Chr.1: 152510701-152510701 on Build GRCh38
rs920800977 Chr.1: 152510924-152510924 on Build GRCh38
rs941518904 Chr.1: 152511083-152511083 on Build GRCh38
rs370339235 Chr.1: 152510697-152510697 on Build GRCh38
rs1009878173 Chr.1: 152510909-152510909 on Build GRCh38
rs553052046 Chr.1: 152510828-152510828 on Build GRCh38
rs575079103 Chr.1: 152511072-152511072 on Build GRCh38
rs537783111 Chr.1: 152510764-152510764 on Build GRCh38
rs151260858 Chr.1: 152511036-152511036 on Build GRCh38
rs76517809 Chr.1: 152510780-152510780 on Build GRCh38
rs991080648 Chr.1: 152510918-152510918 on Build GRCh38
rs892411184 Chr.1: 152510875-152510875 on Build GRCh38
rs930816493 Chr.1: 152510946-152510946 on Build GRCh38
rs981793646 Chr.1: 152510822-152510822 on Build GRCh38
rs867072728 Chr.1: 152510807-152510807 on Build GRCh38
rs528943832 Chr.1: 152510728-152510728 on Build GRCh38
rs564829392 Chr.1: 152510680-152510680 on Build GRCh38
rs566580416 Chr.1: 152510679-152510679 on Build GRCh38
rs573937954 Chr.1: 152510973-152510973 on Build GRCh38
rs930764138 Chr.1: 152510662-152510662 on Build GRCh38

View Full Product Details