Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153070115-153070650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRR2B
Entrez Gene ID
6701
Gene Name
small proline rich protein 2B
Gene Aliases
-
UniGene
Hs.568239
Gene Chromosome Location
Chr.1: 153070226-153071608 on Build GRCh38
Transcripts
NM_001017418.2 XM_017002174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4166689 c.207G>A p.P69P Chr.1: 153070633-153070633 on Build GRCh38
7422837 c.209A>T p.K70M Chr.1: 153070631-153070631 on Build GRCh38
6789311 c.194C>A p.P65Q Chr.1: 153070646-153070646 on Build GRCh38
1639510 c.203C>A p.P68Q Chr.1: 153070637-153070637 on Build GRCh38
6338396 c.200A>G p.Y67C Chr.1: 153070640-153070640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911390991 Chr.1: 153070521-153070521 on Build GRCh38
rs547667988 Chr.1: 153070182-153070182 on Build GRCh38
rs1047644824 Chr.1: 153070318-153070318 on Build GRCh38
rs984269709 Chr.1: 153070503-153070503 on Build GRCh38
rs2275301 Chr.1: 153070316-153070316 on Build GRCh38
rs969093493 Chr.1: 153070387-153070387 on Build GRCh38
rs112227433 Chr.1: 153070497-153070497 on Build GRCh38
rs539547635 Chr.1: 153070212-153070212 on Build GRCh38
rs371258955 Chr.1: 153070613-153070613 on Build GRCh38
rs767447141 Chr.1: 153070600-153070600 on Build GRCh38
rs538555480 Chr.1: 153070302-153070302 on Build GRCh38
rs551532840 Chr.1: 153070246-153070246 on Build GRCh38
rs918858378 Chr.1: 153070220-153070220 on Build GRCh38
rs1040326029 Chr.1: 153070567-153070567 on Build GRCh38
rs989726234 Chr.1: 153070248-153070248 on Build GRCh38
rs914161151 Chr.1: 153070268-153070268 on Build GRCh38
rs996913504 Chr.1: 153070342-153070342 on Build GRCh38
rs574498044 Chr.1: 153070439-153070439 on Build GRCh38
rs576864561 Chr.1: 153070476-153070476 on Build GRCh38
rs868740266 Chr.1: 153070292-153070292 on Build GRCh38
rs971754491 Chr.1: 153070491-153070491 on Build GRCh38
rs1021155272 Chr.1: 153070398-153070398 on Build GRCh38
rs748610140 Chr.1: 153070573-153070573 on Build GRCh38
rs1008730719 Chr.1: 153070339-153070339 on Build GRCh38
rs766833018 Chr.1: 153070590-153070590 on Build GRCh38
rs983152365 Chr.1: 153070525-153070525 on Build GRCh38
rs961689491 Chr.1: 153070468-153070468 on Build GRCh38
rs767097822 Chr.1: 153070156-153070156 on Build GRCh38
rs543116794 Chr.1: 153070448-153070448 on Build GRCh38
rs879106923 Chr.1: 153070628-153070628 on Build GRCh38
rs371939310 Chr.1: 153070593-153070593 on Build GRCh38
rs567779298 Chr.1: 153070191-153070191 on Build GRCh38
rs527977173 Chr.1: 153070516-153070516 on Build GRCh38
rs776257142 Chr.1: 153070570-153070570 on Build GRCh38
rs997234506 Chr.1: 153070196-153070196 on Build GRCh38
rs545815691 Chr.1: 153070477-153070477 on Build GRCh38
rs759850837 Chr.1: 153070599-153070599 on Build GRCh38
rs773505734 Chr.1: 153070580-153070580 on Build GRCh38
rs750096414 Chr.1: 153070173-153070173 on Build GRCh38
rs1010872130 Chr.1: 153070396-153070396 on Build GRCh38
rs565620222 Chr.1: 153070502-153070502 on Build GRCh38
rs918839399 Chr.1: 153070494-153070494 on Build GRCh38
rs368525974 Chr.1: 153070611-153070611 on Build GRCh38
rs1008743650 Chr.1: 153070164-153070164 on Build GRCh38
rs550168370 Chr.1: 153070261-153070261 on Build GRCh38
rs901239154 Chr.1: 153070341-153070341 on Build GRCh38
rs886388770 Chr.1: 153070324-153070324 on Build GRCh38
rs527937563 Chr.1: 153070148-153070148 on Build GRCh38
rs953077971 Chr.1: 153070501-153070501 on Build GRCh38
rs773183303 Chr.1: 153070564-153070564 on Build GRCh38
rs765911241 Chr.1: 153070297-153070297 on Build GRCh38
rs931811756 Chr.1: 153070589-153070589 on Build GRCh38
rs546954373 Chr.1: 153070515-153070515 on Build GRCh38
rs752856669 Chr.1: 153070601-153070601 on Build GRCh38
rs1028845058 Chr.1: 153070368-153070368 on Build GRCh38
rs907773340 Chr.1: 153070531-153070531 on Build GRCh38
rs117850648 Chr.1: 153070436-153070436 on Build GRCh38
rs960250596 Chr.1: 153070185-153070185 on Build GRCh38
rs966818073 Chr.1: 153070270-153070270 on Build GRCh38
rs930318298 Chr.1: 153070519-153070519 on Build GRCh38
rs999505289 Chr.1: 153070313-153070313 on Build GRCh38
rs966973188 Chr.1: 153070401-153070401 on Build GRCh38
rs1041090197 Chr.1: 153070563-153070563 on Build GRCh38
rs530112125 Chr.1: 153070276-153070276 on Build GRCh38
rs900311786 Chr.1: 153070568-153070568 on Build GRCh38
rs958028414 Chr.1: 153070388-153070388 on Build GRCh38
rs1040253276 Chr.1: 153070340-153070340 on Build GRCh38
rs770286075 Chr.1: 153070575-153070575 on Build GRCh38
rs180980083 Chr.1: 153070363-153070363 on Build GRCh38
rs772126897 Chr.1: 153070350-153070350 on Build GRCh38
rs748423915 Chr.1: 153070299-153070299 on Build GRCh38
rs982227878 Chr.1: 153070422-153070422 on Build GRCh38
rs763287872 Chr.1: 153070582-153070582 on Build GRCh38
rs750446544 Chr.1: 153070614-153070614 on Build GRCh38
rs1023893094 Chr.1: 153070379-153070379 on Build GRCh38
rs958294043 Chr.1: 153070227-153070227 on Build GRCh38
rs556926193 Chr.1: 153070455-153070455 on Build GRCh38
rs554447125 Chr.1: 153070437-153070437 on Build GRCh38

View Full Product Details