Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 45902927-45903425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC5
Entrez Gene ID
53405
Gene Name
chloride intracellular channel 5
Gene Aliases
DFNB102, DFNB103, MST130, MSTP130
UniGene
Hs.485489
Gene Chromosome Location
Chr.6: 45898451-46129809 on Build GRCh38
Transcripts
NM_001114086.1 NM_016929.4 NR_045672.1 NR_045673.1 XM_017010953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672784 c.1157T>C p.F386S Chr.6: 45903164-45903164 on Build GRCh38
3627883 c.749G>A p.R250Q Chr.6: 45903095-45903095 on Build GRCh38
5823713 c.1070T>A p.V357E Chr.6: 45903251-45903251 on Build GRCh38
7146907 c.706G>C p.E236Q Chr.6: 45903138-45903138 on Build GRCh38
356193 c.708G>T p.E236D Chr.6: 45903136-45903136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934157327 Chr.6: 45903011-45903012 on Build GRCh38
rs774779406 Chr.6: 45903202-45903202 on Build GRCh38
rs879014681 Chr.6: 45903174-45903174 on Build GRCh38
rs375247542 Chr.6: 45903063-45903063 on Build GRCh38
rs770060180 Chr.6: 45903269-45903269 on Build GRCh38
rs1040988406 Chr.6: 45903159-45903159 on Build GRCh38
rs113982625 Chr.6: 45903342-45903342 on Build GRCh38
rs536635423 Chr.6: 45902959-45902959 on Build GRCh38
rs150143344 Chr.6: 45903121-45903121 on Build GRCh38
rs914052565 Chr.6: 45903260-45903260 on Build GRCh38
rs772026065 Chr.6: 45903102-45903102 on Build GRCh38
rs755017418 Chr.6: 45903247-45903247 on Build GRCh38
rs533995355 Chr.6: 45903104-45903104 on Build GRCh38
rs193274454 Chr.6: 45903095-45903095 on Build GRCh38
rs753616506 Chr.6: 45903137-45903137 on Build GRCh38
rs748226068 Chr.6: 45903087-45903087 on Build GRCh38
rs764817817 Chr.6: 45903337-45903337 on Build GRCh38
rs754440328 Chr.6: 45903057-45903057 on Build GRCh38
rs192479279 Chr.6: 45903221-45903221 on Build GRCh38
rs767093009 Chr.6: 45903059-45903059 on Build GRCh38
rs765745426 Chr.6: 45903040-45903040 on Build GRCh38
rs554810737 Chr.6: 45902993-45902993 on Build GRCh38
rs763828992 Chr.6: 45903135-45903135 on Build GRCh38
rs969178808 Chr.6: 45903257-45903257 on Build GRCh38
rs777645379 Chr.6: 45903071-45903071 on Build GRCh38
rs747157176 Chr.6: 45903109-45903109 on Build GRCh38
rs999203258 Chr.6: 45902979-45902979 on Build GRCh38
rs548572455 Chr.6: 45903371-45903371 on Build GRCh38
rs1043716893 Chr.6: 45903200-45903200 on Build GRCh38
rs569565327 Chr.6: 45903358-45903358 on Build GRCh38
rs772163210 Chr.6: 45903093-45903093 on Build GRCh38
rs957805647 Chr.6: 45902992-45902992 on Build GRCh38
rs930759206 Chr.6: 45903120-45903120 on Build GRCh38
rs777943609 Chr.6: 45903281-45903281 on Build GRCh38
rs1031163403 Chr.6: 45902991-45902991 on Build GRCh38
rs371423137 Chr.6: 45903240-45903240 on Build GRCh38
rs187468500 Chr.6: 45902958-45902958 on Build GRCh38
rs569204750 Chr.6: 45903103-45903103 on Build GRCh38
rs368704000 Chr.6: 45903132-45903132 on Build GRCh38
rs202102019 Chr.6: 45903171-45903171 on Build GRCh38
rs755770652 Chr.6: 45903133-45903133 on Build GRCh38
rs530370366 Chr.6: 45903369-45903369 on Build GRCh38
rs965382184 Chr.6: 45902973-45902973 on Build GRCh38
rs776828172 Chr.6: 45903183-45903183 on Build GRCh38
rs916438769 Chr.6: 45903050-45903050 on Build GRCh38
rs770120642 Chr.6: 45903195-45903195 on Build GRCh38
rs35735653 Chr.6: 45903263-45903264 on Build GRCh38
rs748319669 Chr.6: 45903265-45903265 on Build GRCh38
rs544931083 Chr.6: 45903140-45903140 on Build GRCh38
rs759216865 Chr.6: 45903201-45903201 on Build GRCh38
rs745741071 Chr.6: 45903146-45903146 on Build GRCh38
rs979665702 Chr.6: 45903293-45903293 on Build GRCh38
rs750232438 Chr.6: 45903237-45903237 on Build GRCh38
rs187897856 Chr.6: 45903148-45903148 on Build GRCh38
rs202022202 Chr.6: 45903105-45903105 on Build GRCh38
rs138583424 Chr.6: 45903184-45903184 on Build GRCh38
rs781297978 Chr.6: 45903252-45903252 on Build GRCh38
rs532735433 Chr.6: 45903315-45903315 on Build GRCh38
rs551334536 Chr.6: 45903326-45903326 on Build GRCh38
rs373644994 Chr.6: 45903154-45903154 on Build GRCh38
rs920619958 Chr.6: 45903387-45903387 on Build GRCh38
rs199935144 Chr.6: 45903264-45903264 on Build GRCh38
rs896196395 Chr.6: 45903117-45903117 on Build GRCh38
rs771269822 Chr.6: 45903307-45903307 on Build GRCh38
rs766899135 Chr.6: 45903130-45903130 on Build GRCh38
rs542607424 Chr.6: 45903173-45903173 on Build GRCh38
rs372375671 Chr.6: 45903092-45903092 on Build GRCh38
rs539916161 Chr.6: 45903220-45903220 on Build GRCh38
rs779927896 Chr.6: 45903170-45903170 on Build GRCh38
rs981972348 Chr.6: 45903323-45903323 on Build GRCh38
rs370470268 Chr.6: 45903045-45903045 on Build GRCh38
rs749652445 Chr.6: 45903306-45903306 on Build GRCh38
rs745860376 Chr.6: 45903096-45903096 on Build GRCh38
rs376835561 Chr.6: 45903223-45903223 on Build GRCh38
rs3831297 Chr.6: 45903266-45903267 on Build GRCh38
rs140858725 Chr.6: 45903112-45903112 on Build GRCh38
rs563808750 Chr.6: 45903111-45903111 on Build GRCh38
rs972461807 Chr.6: 45903384-45903384 on Build GRCh38
rs372300556 Chr.6: 45903197-45903197 on Build GRCh38
rs1009027669 Chr.6: 45903155-45903155 on Build GRCh38
rs763539403 Chr.6: 45903129-45903129 on Build GRCh38
rs757089924 Chr.6: 45903138-45903138 on Build GRCh38
rs200361326 Chr.6: 45903198-45903198 on Build GRCh38
rs930657719 Chr.6: 45903396-45903396 on Build GRCh38
rs905168946 Chr.6: 45903343-45903343 on Build GRCh38
rs986143723 Chr.6: 45903399-45903399 on Build GRCh38
rs200204343 Chr.6: 45903233-45903233 on Build GRCh38
rs368110807 Chr.6: 45903064-45903064 on Build GRCh38
rs766742096 Chr.6: 45903139-45903139 on Build GRCh38
rs764896698 Chr.6: 45903236-45903236 on Build GRCh38
rs758208609 Chr.6: 45903238-45903238 on Build GRCh38

View Full Product Details