Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83582124-83582626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
UniGene
Hs.368046
Gene Chromosome Location
Chr.6: 83552885-83709408 on Build GRCh38
Transcripts
NM_001242792.1 NM_001242793.1 NM_001242794.1 NM_001256717.1 NM_001256718.1 NM_014841.2 XM_005248770.4 XM_006715615.1 XM_011536265.1 XM_011536266.1 XM_011536269.1 XM_011536271.1 XM_011536273.2 XM_011536275.2 XM_011536276.2 XM_011536277.1 XM_017011554.1 XM_017011555.1 XM_017011556.1 XM_017011557.1 XM_017011558.1 XM_017011559.1 XM_017011560.1 XM_017011561.1 XM_017011562.1 XM_017011563.1 XM_017011564.1 XM_017011565.1 XM_017011566.1 XM_017011567.1 XM_017011568.1 XM_017011569.1 XM_017011570.1 XM_017011571.1 XM_017011572.1 XM_017011573.1 XM_017011574.1 XM_017011575.1 XM_017011576.1 XM_017011577.1 XM_017011578.1 XM_017011579.1 XM_017011580.1 XM_017011581.1 XM_017011582.1 XM_017011583.1 XM_017011584.1 XM_017011585.1 XM_017011586.1 XM_017011587.1 XM_017011588.1 XM_017011589.1 XM_017011590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
323498 c.2015-2A>C p.? Chr.6: 83582358-83582358 on Build GRCh38
3875850 c.2108C>A p.T703K Chr.6: 83582263-83582263 on Build GRCh38
3411318 c.2037G>A p.A679A Chr.6: 83582334-83582334 on Build GRCh38
1722715 c.2135C>T p.S712F Chr.6: 83582236-83582236 on Build GRCh38
6253859 c.2080C>A p.Q694K Chr.6: 83582291-83582291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980190989 Chr.6: 83582412-83582412 on Build GRCh38
rs775083073 Chr.6: 83582372-83582373 on Build GRCh38
rs762929970 Chr.6: 83582197-83582197 on Build GRCh38
rs923245910 Chr.6: 83582548-83582551 on Build GRCh38
rs372318317 Chr.6: 83582430-83582431 on Build GRCh38
rs757812922 Chr.6: 83582335-83582335 on Build GRCh38
rs764466153 Chr.6: 83582260-83582260 on Build GRCh38
rs770958151 Chr.6: 83582287-83582287 on Build GRCh38
rs908441566 Chr.6: 83582577-83582579 on Build GRCh38
rs1053109122 Chr.6: 83582444-83582444 on Build GRCh38
rs779006343 Chr.6: 83582230-83582230 on Build GRCh38
rs750572065 Chr.6: 83582338-83582338 on Build GRCh38
rs766006917 Chr.6: 83582198-83582198 on Build GRCh38
rs759797947 Chr.6: 83582155-83582155 on Build GRCh38
rs774049654 Chr.6: 83582292-83582292 on Build GRCh38
rs773096842 Chr.6: 83582262-83582262 on Build GRCh38
rs772614396 Chr.6: 83582198-83582198 on Build GRCh38
rs547755750 Chr.6: 83582552-83582552 on Build GRCh38
rs377703742 Chr.6: 83582214-83582214 on Build GRCh38
rs569210256 Chr.6: 83582259-83582259 on Build GRCh38
rs1010213193 Chr.6: 83582426-83582426 on Build GRCh38
rs764786244 Chr.6: 83582366-83582367 on Build GRCh38
rs772242497 Chr.6: 83582240-83582240 on Build GRCh38
rs754286115 Chr.6: 83582276-83582276 on Build GRCh38
rs752398203 Chr.6: 83582303-83582303 on Build GRCh38
rs537691670 Chr.6: 83582409-83582409 on Build GRCh38
rs6912747 Chr.6: 83582313-83582313 on Build GRCh38
rs757723064 Chr.6: 83582229-83582229 on Build GRCh38
rs963406649 Chr.6: 83582281-83582281 on Build GRCh38
rs749963326 Chr.6: 83582459-83582459 on Build GRCh38
rs202058742 Chr.6: 83582334-83582334 on Build GRCh38
rs200135770 Chr.6: 83582206-83582206 on Build GRCh38
rs779944407 Chr.6: 83582352-83582352 on Build GRCh38
rs747281309 Chr.6: 83582366-83582366 on Build GRCh38
rs762379488 Chr.6: 83582384-83582385 on Build GRCh38
rs773421224 Chr.6: 83582260-83582262 on Build GRCh38
rs114037634 Chr.6: 83582271-83582271 on Build GRCh38
rs1027005429 Chr.6: 83582328-83582328 on Build GRCh38
rs773289618 Chr.6: 83582185-83582185 on Build GRCh38
rs1007337601 Chr.6: 83582542-83582542 on Build GRCh38
rs751070611 Chr.6: 83582200-83582200 on Build GRCh38
rs1051610955 Chr.6: 83582528-83582528 on Build GRCh38
rs201039955 Chr.6: 83582237-83582237 on Build GRCh38
rs61729902 Chr.6: 83582258-83582258 on Build GRCh38
rs568934446 Chr.6: 83582437-83582437 on Build GRCh38
rs892407559 Chr.6: 83582473-83582473 on Build GRCh38
rs759303755 Chr.6: 83582360-83582360 on Build GRCh38
rs917440560 Chr.6: 83582351-83582351 on Build GRCh38
rs545137919 Chr.6: 83582168-83582169 on Build GRCh38
rs758049574 Chr.6: 83582541-83582541 on Build GRCh38
rs556774005 Chr.6: 83582427-83582428 on Build GRCh38
rs777747462 Chr.6: 83582369-83582369 on Build GRCh38
rs759072157 Chr.6: 83582297-83582297 on Build GRCh38
rs754355476 Chr.6: 83582216-83582216 on Build GRCh38
rs950738351 Chr.6: 83582550-83582550 on Build GRCh38
rs529641584 Chr.6: 83582443-83582443 on Build GRCh38
rs926052366 Chr.6: 83582414-83582414 on Build GRCh38
rs749066858 Chr.6: 83582371-83582371 on Build GRCh38
rs760187135 Chr.6: 83582305-83582305 on Build GRCh38
rs73484871 Chr.6: 83582489-83582489 on Build GRCh38
rs1033653 Chr.6: 83582420-83582420 on Build GRCh38
rs1017809540 Chr.6: 83582576-83582576 on Build GRCh38
rs898894595 Chr.6: 83582585-83582585 on Build GRCh38
rs754714655 Chr.6: 83582202-83582202 on Build GRCh38
rs1018230777 Chr.6: 83582460-83582460 on Build GRCh38
rs757933639 Chr.6: 83582398-83582398 on Build GRCh38
rs545632479 Chr.6: 83582204-83582204 on Build GRCh38
rs758512167 Chr.6: 83582341-83582341 on Build GRCh38

View Full Product Details