Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89080603-89081094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNRC1
Entrez Gene ID
10957
Gene Name
proline rich nuclear receptor coactivator 1
Gene Aliases
B4-2, PNAS-145, PROL2, PRR2
UniGene
Hs.75969
Gene Chromosome Location
Chr.6: 89080710-89085160 on Build GRCh38
Transcripts
NM_006813.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6479829 c.99G>A p.P33P Chr.6: 89080993-89080993 on Build GRCh38
3876032 c.158C>A p.P53Q Chr.6: 89081052-89081052 on Build GRCh38
6980458 c.59C>A p.P20Q Chr.6: 89080953-89080953 on Build GRCh38
3876031 c.84C>A p.Y28* Chr.6: 89080978-89080978 on Build GRCh38
3831189 c.28G>C p.E10Q Chr.6: 89080922-89080922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752318923 Chr.6: 89080856-89080858 on Build GRCh38
rs1017268237 Chr.6: 89080947-89080947 on Build GRCh38
rs201384456 Chr.6: 89080762-89080764 on Build GRCh38
rs752226092 Chr.6: 89081051-89081051 on Build GRCh38
rs930063910 Chr.6: 89080834-89080834 on Build GRCh38
rs138996422 Chr.6: 89081055-89081055 on Build GRCh38
rs778642099 Chr.6: 89080683-89080683 on Build GRCh38
rs909989803 Chr.6: 89081033-89081033 on Build GRCh38
rs749652952 Chr.6: 89081012-89081012 on Build GRCh38
rs1048047852 Chr.6: 89080633-89080633 on Build GRCh38
rs959787896 Chr.6: 89080764-89080764 on Build GRCh38
rs968964244 Chr.6: 89080685-89080685 on Build GRCh38
rs771663275 Chr.6: 89080728-89080728 on Build GRCh38
rs771346878 Chr.6: 89081015-89081015 on Build GRCh38
rs948355072 Chr.6: 89080629-89080631 on Build GRCh38
rs770204672 Chr.6: 89080893-89080893 on Build GRCh38
rs754617058 Chr.6: 89081042-89081042 on Build GRCh38
rs998766112 Chr.6: 89080640-89080640 on Build GRCh38
rs750246225 Chr.6: 89080866-89080866 on Build GRCh38
rs111592760 Chr.6: 89080750-89080750 on Build GRCh38
rs1001227809 Chr.6: 89080752-89080752 on Build GRCh38
rs376425738 Chr.6: 89080909-89080909 on Build GRCh38
rs933126191 Chr.6: 89080726-89080726 on Build GRCh38
rs750221705 Chr.6: 89080981-89080981 on Build GRCh38
rs759602909 Chr.6: 89080934-89080934 on Build GRCh38
rs1037202069 Chr.6: 89080638-89080638 on Build GRCh38
rs767095345 Chr.6: 89081048-89081048 on Build GRCh38
rs766217585 Chr.6: 89081039-89081039 on Build GRCh38
rs967139857 Chr.6: 89080681-89080681 on Build GRCh38
rs757768258 Chr.6: 89081057-89081057 on Build GRCh38
rs553253595 Chr.6: 89080699-89080699 on Build GRCh38
rs142290803 Chr.6: 89080915-89080915 on Build GRCh38
rs779847527 Chr.6: 89080989-89080989 on Build GRCh38
rs901319189 Chr.6: 89080639-89080639 on Build GRCh38
rs397686554 Chr.6: 89080845-89080848 on Build GRCh38
rs77509309 Chr.6: 89080844-89080844 on Build GRCh38
rs1011200560 Chr.6: 89080669-89080669 on Build GRCh38
rs2231267 Chr.6: 89081009-89081009 on Build GRCh38
rs574604782 Chr.6: 89080713-89080713 on Build GRCh38
rs113633573 Chr.6: 89080845-89080845 on Build GRCh38
rs200652105 Chr.6: 89080889-89080889 on Build GRCh38
rs1056859873 Chr.6: 89080845-89080854 on Build GRCh38
rs1016345350 Chr.6: 89080630-89080630 on Build GRCh38
rs758065103 Chr.6: 89080876-89080876 on Build GRCh38
rs1041547383 Chr.6: 89080708-89080708 on Build GRCh38
rs144161245 Chr.6: 89080930-89080930 on Build GRCh38
rs976791930 Chr.6: 89080804-89080804 on Build GRCh38
rs141030892 Chr.6: 89081018-89081018 on Build GRCh38
rs933708411 Chr.6: 89081007-89081007 on Build GRCh38
rs765111426 Chr.6: 89080966-89080966 on Build GRCh38
rs761772180 Chr.6: 89080956-89080956 on Build GRCh38
rs760592947 Chr.6: 89080844-89080847 on Build GRCh38
rs978780884 Chr.6: 89080696-89080696 on Build GRCh38
rs116999034 Chr.6: 89080693-89080693 on Build GRCh38
rs759152308 Chr.6: 89080782-89080782 on Build GRCh38
rs1035821572 Chr.6: 89080763-89080763 on Build GRCh38
rs530280979 Chr.6: 89080858-89080858 on Build GRCh38
rs767579194 Chr.6: 89080936-89080936 on Build GRCh38
rs999935976 Chr.6: 89080850-89080850 on Build GRCh38
rs942783953 Chr.6: 89080704-89080704 on Build GRCh38
rs909524456 Chr.6: 89080731-89080731 on Build GRCh38
rs145119263 Chr.6: 89081023-89081023 on Build GRCh38
rs1049972577 Chr.6: 89080641-89080642 on Build GRCh38
rs199759749 Chr.6: 89081053-89081053 on Build GRCh38
rs988994671 Chr.6: 89080680-89080680 on Build GRCh38
rs370111073 Chr.6: 89080768-89080768 on Build GRCh38
rs772316901 Chr.6: 89081063-89081063 on Build GRCh38
rs959861984 Chr.6: 89080691-89080691 on Build GRCh38
rs748629618 Chr.6: 89080890-89080890 on Build GRCh38
rs751313758 Chr.6: 89080992-89080992 on Build GRCh38
rs775179680 Chr.6: 89080843-89080846 on Build GRCh38
rs909576779 Chr.6: 89080634-89080634 on Build GRCh38
rs201541377 Chr.6: 89080847-89080848 on Build GRCh38
rs968949673 Chr.6: 89080772-89080772 on Build GRCh38
rs534640155 Chr.6: 89080687-89080687 on Build GRCh38
rs775615765 Chr.6: 89081070-89081070 on Build GRCh38
rs745403270 Chr.6: 89080700-89080700 on Build GRCh38
rs1010371245 Chr.6: 89080767-89080767 on Build GRCh38
rs1035532710 Chr.6: 89080688-89080688 on Build GRCh38
rs754598018 Chr.6: 89080997-89080997 on Build GRCh38
rs905550135 Chr.6: 89080751-89080751 on Build GRCh38
rs377610042 Chr.6: 89081029-89081029 on Build GRCh38
rs547644613 Chr.6: 89080815-89080815 on Build GRCh38
rs755615300 Chr.6: 89081052-89081052 on Build GRCh38
rs746243332 Chr.6: 89081062-89081062 on Build GRCh38
rs1056804363 Chr.6: 89080922-89080922 on Build GRCh38
rs905796221 Chr.6: 89080910-89080910 on Build GRCh38
rs1018913481 Chr.6: 89081000-89081000 on Build GRCh38
rs911245906 Chr.6: 89080701-89080701 on Build GRCh38
rs147772027 Chr.6: 89080931-89080931 on Build GRCh38
rs760718382 Chr.6: 89081024-89081024 on Build GRCh38
rs765190042 Chr.6: 89081025-89081025 on Build GRCh38
rs372473933 Chr.6: 89081006-89081006 on Build GRCh38
rs563272457 Chr.6: 89080729-89080729 on Build GRCh38
rs886317742 Chr.6: 89080736-89080736 on Build GRCh38
rs969826602 Chr.6: 89080647-89080647 on Build GRCh38
rs979451452 Chr.6: 89080783-89080783 on Build GRCh38
rs199615708 Chr.6: 89080881-89080881 on Build GRCh38
rs777085037 Chr.6: 89081062-89081062 on Build GRCh38
rs939773897 Chr.6: 89080637-89080637 on Build GRCh38
rs746037781 Chr.6: 89081022-89081022 on Build GRCh38
rs751401768 Chr.6: 89081041-89081041 on Build GRCh38
rs575452099 Chr.6: 89080743-89080743 on Build GRCh38
rs1035845551 Chr.6: 89080668-89080668 on Build GRCh38
rs946884142 Chr.6: 89081016-89081016 on Build GRCh38
rs576611010 Chr.6: 89080676-89080676 on Build GRCh38
rs148836277 Chr.6: 89080937-89080937 on Build GRCh38
rs114364131 Chr.6: 89080829-89080829 on Build GRCh38
rs922728217 Chr.6: 89080710-89080710 on Build GRCh38
rs939700917 Chr.6: 89080737-89080737 on Build GRCh38
rs66688820 Chr.6: 89080841-89080844 on Build GRCh38
rs200126302 Chr.6: 89080846-89080848 on Build GRCh38
rs758308333 Chr.6: 89080983-89080983 on Build GRCh38
rs541812702 Chr.6: 89080722-89080722 on Build GRCh38
rs189259512 Chr.6: 89080745-89080745 on Build GRCh38
rs778727274 Chr.6: 89080863-89080863 on Build GRCh38
rs148016809 Chr.6: 89081011-89081011 on Build GRCh38
rs1005806869 Chr.6: 89080627-89080627 on Build GRCh38
rs753860284 Chr.6: 89080848-89080848 on Build GRCh38
rs964364541 Chr.6: 89080798-89080798 on Build GRCh38
rs1037999465 Chr.6: 89080903-89080903 on Build GRCh38
rs757974562 Chr.6: 89080895-89080896 on Build GRCh38
rs904713083 Chr.6: 89080988-89080988 on Build GRCh38
rs747107708 Chr.6: 89081071-89081071 on Build GRCh38
rs780684799 Chr.6: 89080998-89080998 on Build GRCh38
rs1001720072 Chr.6: 89080665-89080665 on Build GRCh38
rs532766439 Chr.6: 89080787-89080787 on Build GRCh38
rs907744260 Chr.6: 89080859-89080859 on Build GRCh38
rs565509667 Chr.6: 89080771-89080771 on Build GRCh38
rs762828322 Chr.6: 89081031-89081031 on Build GRCh38

View Full Product Details