Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89631905-89632413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD6
Entrez Gene ID
22881
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
UniGene
Hs.702213
Gene Chromosome Location
Chr.6: 89433178-89633834 on Build GRCh38
Transcripts
NM_001242809.1 NM_001242811.1 NM_001242813.1 NM_001242814.1 NM_014942.4 XM_005248679.3 XM_005248680.3 XM_005248681.2 XM_005248682.3 XM_005248683.3 XM_005248684.3 XM_011535596.2 XM_011535597.2 XM_011535598.2 XM_011535599.2 XM_011535600.2 XM_017010493.1 XM_017010494.1 XM_017010495.1 XM_017010496.1 XM_017010497.1 XM_017010498.1 XM_017010499.1 XM_017010500.1 XM_017010501.1 XM_017010502.1 XM_017010503.1 XM_017010504.1 XM_017010505.1 XM_017010506.1 XM_017010507.1 XM_017010508.1 XM_017010509.1 XM_017010510.1 XM_017010511.1 XM_017010512.1 XM_017010513.1 XM_017010514.1 XM_017010515.1 NM_020466.4 NR_028493.1 NR_028494.1 NR_028495.1

Gene Symbol
LYRM2
Entrez Gene ID
57226
Gene Name
LYR motif containing 2
Gene Aliases
DJ122O8.2
UniGene
Hs.177275
Gene Chromosome Location
Chr.6: 89632224-89638755 on Build GRCh38
Transcripts
NM_001242809.1 NM_001242811.1 NM_001242813.1 NM_001242814.1 NM_014942.4 XM_005248679.3 XM_005248680.3 XM_005248681.2 XM_005248682.3 XM_005248683.3 XM_005248684.3 XM_011535596.2 XM_011535597.2 XM_011535598.2 XM_011535599.2 XM_011535600.2 XM_017010493.1 XM_017010494.1 XM_017010495.1 XM_017010496.1 XM_017010497.1 XM_017010498.1 XM_017010499.1 XM_017010500.1 XM_017010501.1 XM_017010502.1 XM_017010503.1 XM_017010504.1 XM_017010505.1 XM_017010506.1 XM_017010507.1 XM_017010508.1 XM_017010509.1 XM_017010510.1 XM_017010511.1 XM_017010512.1 XM_017010513.1 XM_017010514.1 XM_017010515.1 NM_020466.4 NR_028493.1 NR_028494.1 NR_028495.1

SNP Detail

SNP ID Chromosome Location
rs1029679478 Chr.6: 89632131-89632131 on Build GRCh38
rs76045374 Chr.6: 89632256-89632256 on Build GRCh38
rs1005681281 Chr.6: 89632270-89632271 on Build GRCh38
rs537574740 Chr.6: 89632138-89632138 on Build GRCh38
rs529954781 Chr.6: 89632291-89632291 on Build GRCh38
rs915828934 Chr.6: 89632379-89632379 on Build GRCh38
rs149337546 Chr.6: 89632170-89632170 on Build GRCh38
rs67417326 Chr.6: 89632369-89632370 on Build GRCh38
rs891326537 Chr.6: 89632220-89632220 on Build GRCh38
rs186820118 Chr.6: 89632222-89632222 on Build GRCh38
rs202213868 Chr.6: 89632366-89632366 on Build GRCh38
rs1016084228 Chr.6: 89631931-89631931 on Build GRCh38
rs951914270 Chr.6: 89632035-89632035 on Build GRCh38
rs376578168 Chr.6: 89631949-89631949 on Build GRCh38
rs776128271 Chr.6: 89631975-89631975 on Build GRCh38
rs562479361 Chr.6: 89632320-89632322 on Build GRCh38
rs1044701814 Chr.6: 89632247-89632249 on Build GRCh38
rs1049889620 Chr.6: 89632206-89632206 on Build GRCh38
rs987431780 Chr.6: 89632151-89632151 on Build GRCh38
rs991439207 Chr.6: 89632351-89632351 on Build GRCh38
rs3186412 Chr.6: 89632330-89632330 on Build GRCh38
rs1052790490 Chr.6: 89631935-89631935 on Build GRCh38
rs920539605 Chr.6: 89631995-89631995 on Build GRCh38
rs974461241 Chr.6: 89631980-89631980 on Build GRCh38
rs577214650 Chr.6: 89632194-89632194 on Build GRCh38
rs1008807307 Chr.6: 89632250-89632250 on Build GRCh38
rs934373742 Chr.6: 89632165-89632165 on Build GRCh38
rs925666496 Chr.6: 89632177-89632177 on Build GRCh38
rs991323406 Chr.6: 89632043-89632043 on Build GRCh38
rs753112638 Chr.6: 89632125-89632127 on Build GRCh38
rs950345772 Chr.6: 89632244-89632244 on Build GRCh38
rs561076809 Chr.6: 89632199-89632199 on Build GRCh38
rs896027744 Chr.6: 89631942-89631942 on Build GRCh38
rs924339241 Chr.6: 89632140-89632140 on Build GRCh38
rs145492547 Chr.6: 89632371-89632372 on Build GRCh38
rs750068223 Chr.6: 89632283-89632283 on Build GRCh38
rs951951513 Chr.6: 89632340-89632340 on Build GRCh38
rs10664096 Chr.6: 89632374-89632375 on Build GRCh38
rs994293512 Chr.6: 89632322-89632323 on Build GRCh38
rs895506401 Chr.6: 89632231-89632231 on Build GRCh38
rs138725569 Chr.6: 89632085-89632085 on Build GRCh38
rs541590622 Chr.6: 89632337-89632337 on Build GRCh38
rs111692349 Chr.6: 89632368-89632369 on Build GRCh38
rs563180470 Chr.6: 89632378-89632378 on Build GRCh38
rs971279500 Chr.6: 89632385-89632385 on Build GRCh38
rs906119884 Chr.6: 89632264-89632264 on Build GRCh38
rs138426132 Chr.6: 89632386-89632386 on Build GRCh38
rs544187509 Chr.6: 89632019-89632019 on Build GRCh38
rs866980048 Chr.6: 89632128-89632128 on Build GRCh38
rs528270817 Chr.6: 89631974-89631974 on Build GRCh38
rs200541805 Chr.6: 89632368-89632368 on Build GRCh38
rs555210124 Chr.6: 89632009-89632013 on Build GRCh38
rs111626561 Chr.6: 89632171-89632171 on Build GRCh38
rs906069095 Chr.6: 89632024-89632024 on Build GRCh38
rs1005630236 Chr.6: 89632030-89632030 on Build GRCh38
rs961454604 Chr.6: 89632102-89632108 on Build GRCh38
rs973119639 Chr.6: 89632117-89632117 on Build GRCh38

View Full Product Details