Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 97144745-97145277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMS22L
Entrez Gene ID
253714
Gene Name
MMS22 like, DNA repair protein
Gene Aliases
C6orf167, dJ39B17.2
UniGene
Hs.444292
Gene Chromosome Location
Chr.6: 97142161-97283437 on Build GRCh38
Transcripts
NM_198468.2 XM_006715430.1 XM_017010657.1

SNP Detail

SNP ID Chromosome Location
rs370882315 Chr.6: 97145010-97145010 on Build GRCh38
rs545681601 Chr.6: 97145068-97145069 on Build GRCh38
rs942951105 Chr.6: 97145196-97145196 on Build GRCh38
rs879002608 Chr.6: 97145069-97145069 on Build GRCh38
rs766940135 Chr.6: 97144856-97144858 on Build GRCh38
rs3839355 Chr.6: 97145024-97145025 on Build GRCh38
rs1018697213 Chr.6: 97144844-97144844 on Build GRCh38
rs796497491 Chr.6: 97145025-97145026 on Build GRCh38
rs926636939 Chr.6: 97145160-97145160 on Build GRCh38
rs918046075 Chr.6: 97145073-97145073 on Build GRCh38
rs533100804 Chr.6: 97144886-97144886 on Build GRCh38
rs1006814223 Chr.6: 97144838-97144838 on Build GRCh38
rs905160185 Chr.6: 97144794-97144794 on Build GRCh38
rs919031251 Chr.6: 97145048-97145067 on Build GRCh38
rs3798287 Chr.6: 97144909-97144909 on Build GRCh38
rs998227305 Chr.6: 97145242-97145242 on Build GRCh38
rs949648505 Chr.6: 97145140-97145140 on Build GRCh38
rs550789502 Chr.6: 97145028-97145028 on Build GRCh38
rs553898368 Chr.6: 97144874-97144874 on Build GRCh38
rs535922442 Chr.6: 97144957-97144957 on Build GRCh38
rs3822909 Chr.6: 97145067-97145067 on Build GRCh38
rs956515209 Chr.6: 97144857-97144857 on Build GRCh38
rs972210522 Chr.6: 97145042-97145067 on Build GRCh38
rs59932333 Chr.6: 97145066-97145067 on Build GRCh38
rs1037768970 Chr.6: 97144850-97144850 on Build GRCh38
rs901220726 Chr.6: 97144847-97144847 on Build GRCh38
rs918203141 Chr.6: 97144781-97144781 on Build GRCh38
rs1017293643 Chr.6: 97144947-97144947 on Build GRCh38
rs888661903 Chr.6: 97145178-97145178 on Build GRCh38
rs754243176 Chr.6: 97145065-97145066 on Build GRCh38
rs532557810 Chr.6: 97145170-97145170 on Build GRCh38
rs763245756 Chr.6: 97144878-97144878 on Build GRCh38
rs927616968 Chr.6: 97144962-97144962 on Build GRCh38
rs890378425 Chr.6: 97144898-97144898 on Build GRCh38
rs978814731 Chr.6: 97145019-97145019 on Build GRCh38
rs34588294 Chr.6: 97145067-97145068 on Build GRCh38
rs796992315 Chr.6: 97145066-97145067 on Build GRCh38
rs9400593 Chr.6: 97145026-97145026 on Build GRCh38
rs575625495 Chr.6: 97145063-97145063 on Build GRCh38
rs751472617 Chr.6: 97144809-97144812 on Build GRCh38
rs939064536 Chr.6: 97144963-97144963 on Build GRCh38
rs948614112 Chr.6: 97145148-97145148 on Build GRCh38
rs781379996 Chr.6: 97145170-97145170 on Build GRCh38
rs929333685 Chr.6: 97145202-97145202 on Build GRCh38
rs201716334 Chr.6: 97145022-97145022 on Build GRCh38
rs564631324 Chr.6: 97145168-97145168 on Build GRCh38
rs578167798 Chr.6: 97145201-97145201 on Build GRCh38
rs144674349 Chr.6: 97144802-97144802 on Build GRCh38
rs1038097652 Chr.6: 97145200-97145200 on Build GRCh38
rs775378721 Chr.6: 97144852-97144852 on Build GRCh38
rs71720288 Chr.6: 97145056-97145057 on Build GRCh38
rs1045048071 Chr.6: 97144790-97144790 on Build GRCh38
rs955975460 Chr.6: 97145065-97145065 on Build GRCh38
rs949635508 Chr.6: 97144782-97144782 on Build GRCh38

View Full Product Details