Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 101924522-101925021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIK2
Entrez Gene ID
2898
Gene Name
glutamate ionotropic receptor kainate type subunit 2
Gene Aliases
EAA4, GLR6, GLUK6, GLUR6, GluK2, MRT6
UniGene
Hs.98262
Gene Chromosome Location
Chr.6: 101393708-102070083 on Build GRCh38
Transcripts
NM_001166247.1 NM_021956.4 NM_175768.3 XM_005266945.2 XM_005266946.3 XM_011535777.2 XM_011535778.2 XM_017010781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3153817 c.1758T>C p.P586P Chr.6: 101924610-101924610 on Build GRCh38
5869039 c.1774C>T p.P592S Chr.6: 101924626-101924626 on Build GRCh38
3153822 c.1766G>C p.W589S Chr.6: 101924618-101924618 on Build GRCh38
6105012 c.1789_1790CC>AA p.P597N Chr.6: 101924641-101924642 on Build GRCh38
3776953 c.1867G>A p.G623S Chr.6: 101924719-101924719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767151881 Chr.6: 101924641-101924641 on Build GRCh38
rs540726824 Chr.6: 101924917-101924917 on Build GRCh38
rs374524150 Chr.6: 101924725-101924725 on Build GRCh38
rs113961601 Chr.6: 101924551-101924551 on Build GRCh38
rs1004497104 Chr.6: 101924773-101924773 on Build GRCh38
rs781375354 Chr.6: 101924734-101924734 on Build GRCh38
rs546584086 Chr.6: 101924722-101924722 on Build GRCh38
rs556867979 Chr.6: 101924768-101924768 on Build GRCh38
rs532551421 Chr.6: 101924991-101924991 on Build GRCh38
rs773566956 Chr.6: 101924756-101924756 on Build GRCh38
rs777724559 Chr.6: 101924553-101924553 on Build GRCh38
rs969170081 Chr.6: 101924832-101924832 on Build GRCh38
rs762188093 Chr.6: 101924578-101924579 on Build GRCh38
rs538971046 Chr.6: 101924760-101924760 on Build GRCh38
rs770115532 Chr.6: 101924745-101924745 on Build GRCh38
rs770026337 Chr.6: 101924597-101924597 on Build GRCh38
rs112535925 Chr.6: 101924577-101924577 on Build GRCh38
rs749657225 Chr.6: 101924758-101924758 on Build GRCh38
rs751038289 Chr.6: 101924800-101924800 on Build GRCh38
rs751004500 Chr.6: 101924632-101924632 on Build GRCh38
rs759061308 Chr.6: 101924640-101924640 on Build GRCh38
rs763440330 Chr.6: 101924612-101924612 on Build GRCh38
rs773678882 Chr.6: 101924598-101924598 on Build GRCh38
rs150033177 Chr.6: 101924665-101924665 on Build GRCh38
rs747149258 Chr.6: 101924562-101924562 on Build GRCh38
rs766942576 Chr.6: 101924622-101924622 on Build GRCh38
rs1010083197 Chr.6: 101924996-101924996 on Build GRCh38
rs1052059971 Chr.6: 101924979-101924979 on Build GRCh38
rs752321887 Chr.6: 101924646-101924646 on Build GRCh38
rs560256794 Chr.6: 101924919-101924919 on Build GRCh38
rs753744358 Chr.6: 101924680-101924680 on Build GRCh38
rs371605924 Chr.6: 101924595-101924595 on Build GRCh38
rs890359305 Chr.6: 101924980-101924980 on Build GRCh38
rs757251684 Chr.6: 101924715-101924715 on Build GRCh38
rs140414588 Chr.6: 101924978-101924978 on Build GRCh38
rs1014419083 Chr.6: 101924788-101924788 on Build GRCh38
rs749206072 Chr.6: 101924555-101924555 on Build GRCh38
rs541029730 Chr.6: 101924998-101924998 on Build GRCh38
rs780267075 Chr.6: 101924560-101924560 on Build GRCh38
rs187472444 Chr.6: 101924945-101924945 on Build GRCh38
rs528042314 Chr.6: 101924564-101924564 on Build GRCh38
rs368718306 Chr.6: 101924578-101924578 on Build GRCh38
rs759008368 Chr.6: 101924765-101924765 on Build GRCh38
rs1039751266 Chr.6: 101924902-101924902 on Build GRCh38
rs960526761 Chr.6: 101924780-101924780 on Build GRCh38
rs941349088 Chr.6: 101924829-101924829 on Build GRCh38
rs148242386 Chr.6: 101924851-101924851 on Build GRCh38
rs748299713 Chr.6: 101924736-101924736 on Build GRCh38
rs1023521836 Chr.6: 101924807-101924807 on Build GRCh38
rs184232517 Chr.6: 101924884-101924884 on Build GRCh38
rs755830629 Chr.6: 101924652-101924652 on Build GRCh38
rs888420223 Chr.6: 101924752-101924752 on Build GRCh38
rs1015836558 Chr.6: 101924776-101924776 on Build GRCh38
rs771039873 Chr.6: 101924557-101924557 on Build GRCh38
rs924961127 Chr.6: 101924912-101924913 on Build GRCh38

View Full Product Details