Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118314739-118315249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F1
Entrez Gene ID
222553
Gene Name
solute carrier family 35 member F1
Gene Aliases
C6orf169, dJ230I3.1
UniGene
Hs.654841
Gene Chromosome Location
Chr.6: 117907226-118317676 on Build GRCh38
Transcripts
NM_001029858.3 XM_005266865.4

SNP Detail

SNP ID Chromosome Location
rs996510094 Chr.6: 118315060-118315060 on Build GRCh38
rs183907507 Chr.6: 118315025-118315025 on Build GRCh38
rs138786535 Chr.6: 118314971-118314971 on Build GRCh38
rs1019013563 Chr.6: 118314788-118314788 on Build GRCh38
rs762552933 Chr.6: 118315057-118315057 on Build GRCh38
rs775146152 Chr.6: 118315012-118315012 on Build GRCh38
rs889392341 Chr.6: 118315080-118315080 on Build GRCh38
rs963260761 Chr.6: 118315225-118315225 on Build GRCh38
rs1015781962 Chr.6: 118315191-118315191 on Build GRCh38
rs182031707 Chr.6: 118315037-118315037 on Build GRCh38
rs768167448 Chr.6: 118315063-118315063 on Build GRCh38
rs958381441 Chr.6: 118314958-118314958 on Build GRCh38
rs1009614338 Chr.6: 118315124-118315124 on Build GRCh38
rs572319996 Chr.6: 118314906-118314906 on Build GRCh38
rs76531026 Chr.6: 118314847-118314847 on Build GRCh38
rs747306384 Chr.6: 118314962-118314962 on Build GRCh38
rs946672262 Chr.6: 118315047-118315047 on Build GRCh38
rs955728679 Chr.6: 118314859-118314859 on Build GRCh38
rs944714026 Chr.6: 118315109-118315109 on Build GRCh38
rs1020255700 Chr.6: 118315032-118315032 on Build GRCh38
rs996562309 Chr.6: 118314814-118314814 on Build GRCh38
rs890909661 Chr.6: 118315132-118315132 on Build GRCh38
rs1050795967 Chr.6: 118315111-118315111 on Build GRCh38
rs1004421464 Chr.6: 118314893-118314893 on Build GRCh38
rs576168862 Chr.6: 118315158-118315158 on Build GRCh38
rs1000008793 Chr.6: 118315168-118315168 on Build GRCh38
rs769488146 Chr.6: 118314998-118314998 on Build GRCh38
rs1040813977 Chr.6: 118315118-118315118 on Build GRCh38
rs150653494 Chr.6: 118314920-118314920 on Build GRCh38
rs543559067 Chr.6: 118314929-118314929 on Build GRCh38
rs763772626 Chr.6: 118315100-118315100 on Build GRCh38
rs554726918 Chr.6: 118314919-118314919 on Build GRCh38
rs775853988 Chr.6: 118315022-118315022 on Build GRCh38
rs913787554 Chr.6: 118315036-118315036 on Build GRCh38
rs113182136 Chr.6: 118315129-118315129 on Build GRCh38
rs34689949 Chr.6: 118315001-118315002 on Build GRCh38
rs757570109 Chr.6: 118314913-118314913 on Build GRCh38
rs963017050 Chr.6: 118314917-118314917 on Build GRCh38
rs1043344184 Chr.6: 118315054-118315054 on Build GRCh38
rs1009335392 Chr.6: 118314779-118314779 on Build GRCh38
rs760090277 Chr.6: 118314799-118314799 on Build GRCh38
rs981525399 Chr.6: 118314973-118314973 on Build GRCh38
rs111515894 Chr.6: 118315034-118315034 on Build GRCh38
rs546066013 Chr.6: 118315074-118315074 on Build GRCh38
rs892510825 Chr.6: 118315123-118315124 on Build GRCh38
rs535030794 Chr.6: 118314915-118314915 on Build GRCh38

View Full Product Details