Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154989320-154989835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLAD1
Entrez Gene ID
80308
Gene Name
flavin adenine dinucleotide synthetase 1
Gene Aliases
FAD1, FADS, LSMFLAD, PP591
UniGene
Hs.118666
Gene Chromosome Location
Chr.1: 154983294-154993111 on Build GRCh38
Transcripts
NM_001184891.1 NM_025207.4 NM_201398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232343 c.1118G>A p.G373E Chr.1: 154989560-154989560 on Build GRCh38
5352721 c.1200T>C p.C400C Chr.1: 154989642-154989642 on Build GRCh38
234168 c.1118-1G>A p.? Chr.1: 154989559-154989559 on Build GRCh38
463078 c.1125T>G p.S375S Chr.1: 154989567-154989567 on Build GRCh38
6664391 c.1237C>T p.L413L Chr.1: 154989679-154989679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374605995 Chr.1: 154989544-154989544 on Build GRCh38
rs141161249 Chr.1: 154989594-154989594 on Build GRCh38
rs989865194 Chr.1: 154989781-154989781 on Build GRCh38
rs756159770 Chr.1: 154989519-154989519 on Build GRCh38
rs777240008 Chr.1: 154989764-154989764 on Build GRCh38
rs191570212 Chr.1: 154989527-154989527 on Build GRCh38
rs769162563 Chr.1: 154989599-154989599 on Build GRCh38
rs751108469 Chr.1: 154989781-154989782 on Build GRCh38
rs755714541 Chr.1: 154989676-154989676 on Build GRCh38
rs371270707 Chr.1: 154989351-154989351 on Build GRCh38
rs754788619 Chr.1: 154989655-154989655 on Build GRCh38
rs377444183 Chr.1: 154989513-154989513 on Build GRCh38
rs34294567 Chr.1: 154989610-154989611 on Build GRCh38
rs188363426 Chr.1: 154989439-154989439 on Build GRCh38
rs149756913 Chr.1: 154989613-154989613 on Build GRCh38
rs774708539 Chr.1: 154989615-154989615 on Build GRCh38
rs908112813 Chr.1: 154989503-154989503 on Build GRCh38
rs751432092 Chr.1: 154989654-154989654 on Build GRCh38
rs541501421 Chr.1: 154989813-154989813 on Build GRCh38
rs763362103 Chr.1: 154989606-154989606 on Build GRCh38
rs766291659 Chr.1: 154989653-154989653 on Build GRCh38
rs772415435 Chr.1: 154989728-154989728 on Build GRCh38
rs759840345 Chr.1: 154989616-154989616 on Build GRCh38
rs184603461 Chr.1: 154989420-154989420 on Build GRCh38
rs748771067 Chr.1: 154989707-154989707 on Build GRCh38
rs780951656 Chr.1: 154989593-154989593 on Build GRCh38
rs774796492 Chr.1: 154989721-154989721 on Build GRCh38
rs760425674 Chr.1: 154989486-154989486 on Build GRCh38
rs764079017 Chr.1: 154989631-154989631 on Build GRCh38
rs767770610 Chr.1: 154989623-154989623 on Build GRCh38
rs570263731 Chr.1: 154989340-154989340 on Build GRCh38
rs923428469 Chr.1: 154989522-154989522 on Build GRCh38
rs372031707 Chr.1: 154989397-154989398 on Build GRCh38
rs556010860 Chr.1: 154989740-154989740 on Build GRCh38
rs878865631 Chr.1: 154989791-154989791 on Build GRCh38
rs771399093 Chr.1: 154989607-154989607 on Build GRCh38
rs957033729 Chr.1: 154989769-154989769 on Build GRCh38
rs535077896 Chr.1: 154989592-154989592 on Build GRCh38
rs76665200 Chr.1: 154989545-154989545 on Build GRCh38
rs142224458 Chr.1: 154989561-154989561 on Build GRCh38
rs368097196 Chr.1: 154989609-154989609 on Build GRCh38
rs985054172 Chr.1: 154989496-154989496 on Build GRCh38
rs189649333 Chr.1: 154989628-154989628 on Build GRCh38
rs924293795 Chr.1: 154989419-154989419 on Build GRCh38
rs574458525 Chr.1: 154989757-154989757 on Build GRCh38
rs747788623 Chr.1: 154989674-154989674 on Build GRCh38
rs779719543 Chr.1: 154989551-154989551 on Build GRCh38
rs202098739 Chr.1: 154989671-154989671 on Build GRCh38
rs962275886 Chr.1: 154989506-154989506 on Build GRCh38
rs939937701 Chr.1: 154989529-154989529 on Build GRCh38
rs1012300593 Chr.1: 154989375-154989375 on Build GRCh38
rs775757993 Chr.1: 154989738-154989738 on Build GRCh38
rs1028379351 Chr.1: 154989755-154989755 on Build GRCh38
rs754741967 Chr.1: 154989722-154989722 on Build GRCh38
rs773824432 Chr.1: 154989604-154989604 on Build GRCh38
rs749931262 Chr.1: 154989353-154989353 on Build GRCh38
rs370458690 Chr.1: 154989518-154989518 on Build GRCh38
rs771361489 Chr.1: 154989717-154989717 on Build GRCh38
rs1043646797 Chr.1: 154989605-154989605 on Build GRCh38
rs1023645105 Chr.1: 154989415-154989415 on Build GRCh38
rs146796757 Chr.1: 154989626-154989626 on Build GRCh38
rs371251686 Chr.1: 154989756-154989756 on Build GRCh38
rs753687017 Chr.1: 154989659-154989659 on Build GRCh38
rs758282575 Chr.1: 154989546-154989546 on Build GRCh38
rs148102813 Chr.1: 154989647-154989647 on Build GRCh38
rs80218412 Chr.1: 154989469-154989469 on Build GRCh38
rs576674138 Chr.1: 154989639-154989639 on Build GRCh38
rs999070892 Chr.1: 154989762-154989762 on Build GRCh38
rs547663677 Chr.1: 154989684-154989684 on Build GRCh38

View Full Product Details