Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127968687-127969183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRK
Entrez Gene ID
5796
Gene Name
protein tyrosine phosphatase, receptor type K
Gene Aliases
R-PTP-kappa
UniGene
Hs.155919
Gene Chromosome Location
Chr.6: 127968779-128520847 on Build GRCh38
Transcripts
NM_001135648.2 NM_001291981.1 NM_001291984.1 NM_002844.3 XM_006715537.2 XM_011536014.2 XM_011536015.2 XM_011536016.2 XM_011536017.2 XM_011536018.2 XM_017011145.1 XM_017011146.1 XM_017011147.1 XM_017011148.1 XM_017011149.1 XM_017011150.1

SNP Detail

SNP ID Chromosome Location
rs904809188 Chr.6: 127969003-127969003 on Build GRCh38
rs1013049846 Chr.6: 127968919-127968919 on Build GRCh38
rs952761934 Chr.6: 127969124-127969124 on Build GRCh38
rs868180405 Chr.6: 127969029-127969029 on Build GRCh38
rs1803926 Chr.6: 127969110-127969110 on Build GRCh38
rs897567454 Chr.6: 127968861-127968861 on Build GRCh38
rs758249710 Chr.6: 127969015-127969015 on Build GRCh38
rs1035820250 Chr.6: 127968871-127968871 on Build GRCh38
rs186554999 Chr.6: 127968922-127968922 on Build GRCh38
rs1002614571 Chr.6: 127968865-127968865 on Build GRCh38
rs906210661 Chr.6: 127969071-127969071 on Build GRCh38
rs544413115 Chr.6: 127969081-127969081 on Build GRCh38
rs924669027 Chr.6: 127968978-127968978 on Build GRCh38
rs977811141 Chr.6: 127968934-127968934 on Build GRCh38
rs368019692 Chr.6: 127969037-127969037 on Build GRCh38
rs1006277557 Chr.6: 127968785-127968790 on Build GRCh38
rs3763275 Chr.6: 127968989-127968989 on Build GRCh38
rs542011196 Chr.6: 127968825-127968825 on Build GRCh38
rs11544475 Chr.6: 127968880-127968880 on Build GRCh38
rs965450232 Chr.6: 127968995-127968995 on Build GRCh38
rs1025301503 Chr.6: 127969105-127969105 on Build GRCh38
rs550179683 Chr.6: 127969119-127969119 on Build GRCh38
rs942791564 Chr.6: 127968832-127968832 on Build GRCh38
rs986001536 Chr.6: 127969132-127969132 on Build GRCh38
rs931776321 Chr.6: 127969050-127969050 on Build GRCh38
rs1050736223 Chr.6: 127968725-127968725 on Build GRCh38
rs182217117 Chr.6: 127968744-127968744 on Build GRCh38
rs909105768 Chr.6: 127969143-127969143 on Build GRCh38
rs11544476 Chr.6: 127969030-127969030 on Build GRCh38
rs1041982427 Chr.6: 127968991-127968991 on Build GRCh38
rs140215737 Chr.6: 127968929-127968929 on Build GRCh38
rs994204169 Chr.6: 127969082-127969082 on Build GRCh38
rs1056387736 Chr.6: 127969031-127969031 on Build GRCh38
rs531347951 Chr.6: 127969038-127969038 on Build GRCh38
rs887714693 Chr.6: 127968758-127968758 on Build GRCh38
rs1024655612 Chr.6: 127969039-127969039 on Build GRCh38
rs936266738 Chr.6: 127969016-127969016 on Build GRCh38
rs562407780 Chr.6: 127969151-127969151 on Build GRCh38

View Full Product Details