Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 133888985-133889511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARID
Entrez Gene ID
100507308
Gene Name
TCF21 antisense RNA inducing promoter demethylation
Gene Aliases
EYA4-AS1
UniGene
Hs.102408
Gene Chromosome Location
Chr.6: 133502252-133889006 on Build GRCh38
Transcripts
NR_109982.1 NM_003206.3 NM_198392.2

Gene Symbol
TCF21
Entrez Gene ID
6943
Gene Name
transcription factor 21
Gene Aliases
POD1, bHLHa23
UniGene
Hs.78061
Gene Chromosome Location
Chr.6: 133889121-133895537 on Build GRCh38
Transcripts
NR_109982.1 NM_003206.3 NM_198392.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073497 c.80C>T p.S27L Chr.6: 133889477-133889477 on Build GRCh38
5884307 c.61G>A p.D21N Chr.6: 133889458-133889458 on Build GRCh38
7393284 c.63C>G p.D21E Chr.6: 133889460-133889460 on Build GRCh38
391041 c.90_91insT p.V32fs*106 Chr.6: 133889487-133889488 on Build GRCh38
4732674 c.19A>G p.S7G Chr.6: 133889416-133889416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759145862 Chr.6: 133889353-133889353 on Build GRCh38
rs756380889 Chr.6: 133889423-133889423 on Build GRCh38
rs137942473 Chr.6: 133889356-133889359 on Build GRCh38
rs771498542 Chr.6: 133889367-133889367 on Build GRCh38
rs778713189 Chr.6: 133889439-133889439 on Build GRCh38
rs180692886 Chr.6: 133889047-133889047 on Build GRCh38
rs921076908 Chr.6: 133889323-133889323 on Build GRCh38
rs760277190 Chr.6: 133889381-133889381 on Build GRCh38
rs931226052 Chr.6: 133889039-133889039 on Build GRCh38
rs398002873 Chr.6: 133889050-133889051 on Build GRCh38
rs561971771 Chr.6: 133889197-133889197 on Build GRCh38
rs906650649 Chr.6: 133889393-133889393 on Build GRCh38
rs750857469 Chr.6: 133889386-133889386 on Build GRCh38
rs564308524 Chr.6: 133889216-133889216 on Build GRCh38
rs965088438 Chr.6: 133889301-133889301 on Build GRCh38
rs766654886 Chr.6: 133889470-133889470 on Build GRCh38
rs3190723 Chr.6: 133889283-133889283 on Build GRCh38
rs763405748 Chr.6: 133889387-133889387 on Build GRCh38
rs912594326 Chr.6: 133889229-133889229 on Build GRCh38
rs570347337 Chr.6: 133889406-133889406 on Build GRCh38
rs755847724 Chr.6: 133889155-133889155 on Build GRCh38
rs1012550424 Chr.6: 133889064-133889064 on Build GRCh38
rs34718376 Chr.6: 133889360-133889361 on Build GRCh38
rs547987017 Chr.6: 133889299-133889299 on Build GRCh38
rs971019868 Chr.6: 133889127-133889127 on Build GRCh38
rs531365701 Chr.6: 133889244-133889244 on Build GRCh38
rs1002340232 Chr.6: 133889191-133889191 on Build GRCh38
rs397730436 Chr.6: 133889049-133889050 on Build GRCh38
rs543509657 Chr.6: 133889227-133889227 on Build GRCh38
rs768488824 Chr.6: 133889461-133889461 on Build GRCh38
rs912277121 Chr.6: 133889257-133889257 on Build GRCh38
rs3068172 Chr.6: 133889377-133889378 on Build GRCh38
rs770702769 Chr.6: 133889438-133889438 on Build GRCh38
rs770899118 Chr.6: 133889351-133889351 on Build GRCh38
rs920152438 Chr.6: 133889293-133889293 on Build GRCh38
rs141634426 Chr.6: 133889324-133889359 on Build GRCh38
rs56412384 Chr.6: 133889462-133889462 on Build GRCh38
rs931260379 Chr.6: 133889329-133889329 on Build GRCh38
rs759978914 Chr.6: 133889480-133889480 on Build GRCh38
rs869047647 Chr.6: 133889048-133889049 on Build GRCh38
rs1023902390 Chr.6: 133889118-133889118 on Build GRCh38
rs755444037 Chr.6: 133889395-133889395 on Build GRCh38
rs113499922 Chr.6: 133889359-133889359 on Build GRCh38
rs1046455217 Chr.6: 133889165-133889165 on Build GRCh38
rs374738237 Chr.6: 133889385-133889385 on Build GRCh38
rs5880197 Chr.6: 133889359-133889360 on Build GRCh38
rs868018344 Chr.6: 133889485-133889485 on Build GRCh38
rs5880195 Chr.6: 133889047-133889048 on Build GRCh38
rs899792577 Chr.6: 133889027-133889027 on Build GRCh38
rs200552219 Chr.6: 133889355-133889355 on Build GRCh38
rs372484305 Chr.6: 133889449-133889449 on Build GRCh38
rs767049462 Chr.6: 133889389-133889389 on Build GRCh38
rs764954176 Chr.6: 133889358-133889359 on Build GRCh38
rs775850686 Chr.6: 133889471-133889471 on Build GRCh38
rs954130618 Chr.6: 133889217-133889217 on Build GRCh38
rs533391163 Chr.6: 133889222-133889222 on Build GRCh38
rs368405662 Chr.6: 133889359-133889359 on Build GRCh38
rs542448654 Chr.6: 133889177-133889177 on Build GRCh38
rs186141377 Chr.6: 133889051-133889051 on Build GRCh38
rs538738060 Chr.6: 133889228-133889228 on Build GRCh38
rs957433577 Chr.6: 133889186-133889186 on Build GRCh38
rs967456656 Chr.6: 133889478-133889478 on Build GRCh38
rs975255114 Chr.6: 133889310-133889310 on Build GRCh38
rs376800209 Chr.6: 133889455-133889455 on Build GRCh38
rs1048895877 Chr.6: 133889055-133889055 on Build GRCh38
rs79935065 Chr.6: 133889137-133889137 on Build GRCh38
rs368271556 Chr.6: 133889412-133889412 on Build GRCh38
rs200648700 Chr.6: 133889376-133889376 on Build GRCh38
rs749150716 Chr.6: 133889430-133889430 on Build GRCh38
rs887103641 Chr.6: 133889131-133889131 on Build GRCh38
rs12209636 Chr.6: 133889417-133889417 on Build GRCh38
rs933280010 Chr.6: 133889116-133889116 on Build GRCh38
rs142486086 Chr.6: 133889424-133889424 on Build GRCh38
rs371010861 Chr.6: 133889351-133889352 on Build GRCh38
rs1037328155 Chr.6: 133889290-133889290 on Build GRCh38
rs77092807 Chr.6: 133889378-133889378 on Build GRCh38
rs1046926164 Chr.6: 133889388-133889388 on Build GRCh38
rs539136648 Chr.6: 133889021-133889021 on Build GRCh38
rs906704671 Chr.6: 133889168-133889168 on Build GRCh38
rs572670111 Chr.6: 133889202-133889202 on Build GRCh38
rs779872002 Chr.6: 133889361-133889362 on Build GRCh38
rs943834292 Chr.6: 133889013-133889013 on Build GRCh38
rs1131141 Chr.6: 133889287-133889287 on Build GRCh38
rs61729591 Chr.6: 133889460-133889460 on Build GRCh38
rs184160585 Chr.6: 133889215-133889215 on Build GRCh38
rs774154714 Chr.6: 133889352-133889352 on Build GRCh38
rs556339026 Chr.6: 133889022-133889022 on Build GRCh38

View Full Product Details