Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138812449-138812950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

SNP Detail

SNP ID Chromosome Location
rs983672158 Chr.6: 138812570-138812570 on Build GRCh38
rs188199425 Chr.6: 138812575-138812575 on Build GRCh38
rs964354544 Chr.6: 138812714-138812714 on Build GRCh38
rs139339151 Chr.6: 138812773-138812773 on Build GRCh38
rs546603965 Chr.6: 138812676-138812676 on Build GRCh38
rs977941432 Chr.6: 138812590-138812590 on Build GRCh38
rs983194536 Chr.6: 138812744-138812744 on Build GRCh38
rs147503903 Chr.6: 138812792-138812792 on Build GRCh38
rs559941555 Chr.6: 138812737-138812737 on Build GRCh38
rs571755068 Chr.6: 138812858-138812858 on Build GRCh38
rs940816494 Chr.6: 138812529-138812529 on Build GRCh38
rs920303662 Chr.6: 138812558-138812558 on Build GRCh38
rs542922232 Chr.6: 138812760-138812760 on Build GRCh38
rs113874269 Chr.6: 138812791-138812791 on Build GRCh38
rs917105780 Chr.6: 138812759-138812759 on Build GRCh38
rs9403003 Chr.6: 138812904-138812904 on Build GRCh38
rs903211798 Chr.6: 138812865-138812865 on Build GRCh38
rs1009759707 Chr.6: 138812624-138812624 on Build GRCh38
rs1009120658 Chr.6: 138812675-138812675 on Build GRCh38
rs776866999 Chr.6: 138812541-138812541 on Build GRCh38
rs1044313301 Chr.6: 138812583-138812583 on Build GRCh38
rs140053131 Chr.6: 138812897-138812897 on Build GRCh38
rs141270426 Chr.6: 138812715-138812719 on Build GRCh38
rs1017235064 Chr.6: 138812524-138812524 on Build GRCh38
rs573594822 Chr.6: 138812586-138812586 on Build GRCh38
rs527552869 Chr.6: 138812739-138812739 on Build GRCh38
rs746866 Chr.6: 138812900-138812900 on Build GRCh38
rs375071333 Chr.6: 138812717-138812721 on Build GRCh38
rs945463598 Chr.6: 138812874-138812874 on Build GRCh38
rs546097588 Chr.6: 138812574-138812574 on Build GRCh38
rs536976673 Chr.6: 138812532-138812532 on Build GRCh38
rs62440909 Chr.6: 138812659-138812659 on Build GRCh38
rs960168084 Chr.6: 138812640-138812640 on Build GRCh38
rs937977936 Chr.6: 138812589-138812589 on Build GRCh38
rs1054176546 Chr.6: 138812814-138812814 on Build GRCh38
rs950507223 Chr.6: 138812743-138812743 on Build GRCh38
rs1053910597 Chr.6: 138812603-138812603 on Build GRCh38
rs765411609 Chr.6: 138812674-138812674 on Build GRCh38
rs902045861 Chr.6: 138812539-138812539 on Build GRCh38
rs1050639628 Chr.6: 138812805-138812805 on Build GRCh38
rs543927489 Chr.6: 138812595-138812595 on Build GRCh38
rs929188964 Chr.6: 138812568-138812568 on Build GRCh38
rs1042426284 Chr.6: 138812896-138812896 on Build GRCh38
rs1027444493 Chr.6: 138812742-138812742 on Build GRCh38
rs1036201857 Chr.6: 138812875-138812875 on Build GRCh38
rs994902625 Chr.6: 138812736-138812736 on Build GRCh38
rs113349682 Chr.6: 138812606-138812606 on Build GRCh38
rs759408511 Chr.6: 138812560-138812560 on Build GRCh38
rs1011872590 Chr.6: 138812846-138812846 on Build GRCh38
rs890591992 Chr.6: 138812665-138812665 on Build GRCh38
rs939778911 Chr.6: 138812781-138812781 on Build GRCh38

View Full Product Details