Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149517270-149517774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL4
Entrez Gene ID
85313
Gene Name
peptidylprolyl isomerase like 4
Gene Aliases
HDCME13P
UniGene
Hs.744238
Gene Chromosome Location
Chr.6: 149504495-149546102 on Build GRCh38
Transcripts
NM_139126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884317 c.1062A>C p.K354N Chr.6: 149517371-149517371 on Build GRCh38
7305043 c.1024G>T p.E342* Chr.6: 149517409-149517409 on Build GRCh38
3018640 c.989A>C p.K330T Chr.6: 149517444-149517444 on Build GRCh38
483519 c.983G>T p.G328V Chr.6: 149517450-149517450 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761220378 Chr.6: 149517494-149517494 on Build GRCh38
rs768795658 Chr.6: 149517445-149517445 on Build GRCh38
rs750960342 Chr.6: 149517319-149517319 on Build GRCh38
rs746319069 Chr.6: 149517751-149517751 on Build GRCh38
rs542219967 Chr.6: 149517666-149517666 on Build GRCh38
rs758109872 Chr.6: 149517330-149517330 on Build GRCh38
rs754405485 Chr.6: 149517322-149517322 on Build GRCh38
rs950693 Chr.6: 149517488-149517488 on Build GRCh38
rs983029335 Chr.6: 149517522-149517522 on Build GRCh38
rs752304787 Chr.6: 149517325-149517325 on Build GRCh38
rs767952044 Chr.6: 149517480-149517480 on Build GRCh38
rs767852020 Chr.6: 149517313-149517313 on Build GRCh38
rs974957425 Chr.6: 149517397-149517397 on Build GRCh38
rs79215975 Chr.6: 149517717-149517717 on Build GRCh38
rs775621486 Chr.6: 149517422-149517422 on Build GRCh38
rs987492208 Chr.6: 149517705-149517705 on Build GRCh38
rs763082000 Chr.6: 149517430-149517430 on Build GRCh38
rs148267012 Chr.6: 149517484-149517484 on Build GRCh38
rs748511958 Chr.6: 149517385-149517385 on Build GRCh38
rs770048647 Chr.6: 149517374-149517374 on Build GRCh38
rs777637073 Chr.6: 149517337-149517337 on Build GRCh38
rs769943472 Chr.6: 149517297-149517297 on Build GRCh38
rs565349126 Chr.6: 149517588-149517588 on Build GRCh38
rs780356731 Chr.6: 149517558-149517558 on Build GRCh38
rs778453370 Chr.6: 149517324-149517324 on Build GRCh38
rs781635605 Chr.6: 149517528-149517529 on Build GRCh38
rs759476084 Chr.6: 149517296-149517296 on Build GRCh38
rs770775526 Chr.6: 149517351-149517351 on Build GRCh38
rs906468876 Chr.6: 149517680-149517680 on Build GRCh38
rs773015237 Chr.6: 149517483-149517484 on Build GRCh38
rs746734566 Chr.6: 149517340-149517340 on Build GRCh38
rs912839062 Chr.6: 149517543-149517543 on Build GRCh38
rs780943934 Chr.6: 149517393-149517393 on Build GRCh38
rs554185424 Chr.6: 149517601-149517601 on Build GRCh38
rs116528026 Chr.6: 149517630-149517630 on Build GRCh38
rs773885731 Chr.6: 149517295-149517305 on Build GRCh38
rs766852344 Chr.6: 149517498-149517498 on Build GRCh38
rs527688682 Chr.6: 149517662-149517662 on Build GRCh38
rs965562268 Chr.6: 149517579-149517579 on Build GRCh38
rs200911337 Chr.6: 149517304-149517304 on Build GRCh38
rs904390619 Chr.6: 149517627-149517627 on Build GRCh38
rs745852125 Chr.6: 149517401-149517401 on Build GRCh38
rs558272578 Chr.6: 149517548-149517548 on Build GRCh38
rs769661736 Chr.6: 149517413-149517413 on Build GRCh38
rs775538330 Chr.6: 149517298-149517298 on Build GRCh38
rs191857829 Chr.6: 149517557-149517557 on Build GRCh38
rs943409328 Chr.6: 149517373-149517373 on Build GRCh38
rs913863867 Chr.6: 149517641-149517641 on Build GRCh38
rs774565547 Chr.6: 149517456-149517456 on Build GRCh38
rs964838866 Chr.6: 149517731-149517731 on Build GRCh38
rs1025952549 Chr.6: 149517368-149517368 on Build GRCh38
rs762113728 Chr.6: 149517469-149517469 on Build GRCh38
rs886683321 Chr.6: 149517528-149517528 on Build GRCh38
rs920733811 Chr.6: 149517421-149517421 on Build GRCh38
rs751913578 Chr.6: 149517306-149517306 on Build GRCh38
rs569374967 Chr.6: 149517300-149517300 on Build GRCh38
rs936720089 Chr.6: 149517613-149517613 on Build GRCh38
rs761428138 Chr.6: 149517452-149517455 on Build GRCh38
rs144110636 Chr.6: 149517468-149517468 on Build GRCh38
rs750791550 Chr.6: 149517485-149517485 on Build GRCh38

View Full Product Details