Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158585217-158585712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM181
Entrez Gene ID
57583
Gene Name
transmembrane protein 181
Gene Aliases
GPR178, KIAA1423
UniGene
Hs.666596
Gene Chromosome Location
Chr.6: 158536432-158635435 on Build GRCh38
Transcripts
NM_020823.1 XM_005267074.3 XM_011535999.1 XM_011536000.1 XM_011536001.2 XM_011536002.2 XM_011536003.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5774164 c.709G>T p.V237F Chr.6: 158585342-158585342 on Build GRCh38
1187209 c.767G>C p.R256P Chr.6: 158585400-158585400 on Build GRCh38
3765246 c.758T>C p.V253A Chr.6: 158585391-158585391 on Build GRCh38
1075438 c.711C>T p.V237V Chr.6: 158585344-158585344 on Build GRCh38
5610859 c.744C>A p.Y248* Chr.6: 158585377-158585377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933392689 Chr.6: 158585405-158585405 on Build GRCh38
rs777463198 Chr.6: 158585654-158585655 on Build GRCh38
rs374976562 Chr.6: 158585473-158585473 on Build GRCh38
rs140836924 Chr.6: 158585301-158585301 on Build GRCh38
rs775157793 Chr.6: 158585358-158585358 on Build GRCh38
rs748850216 Chr.6: 158585451-158585451 on Build GRCh38
rs758860264 Chr.6: 158585414-158585414 on Build GRCh38
rs766451395 Chr.6: 158585318-158585318 on Build GRCh38
rs566609873 Chr.6: 158585397-158585397 on Build GRCh38
rs779673557 Chr.6: 158585446-158585446 on Build GRCh38
rs894395422 Chr.6: 158585420-158585420 on Build GRCh38
rs563237784 Chr.6: 158585690-158585690 on Build GRCh38
rs1037685475 Chr.6: 158585677-158585677 on Build GRCh38
rs745471236 Chr.6: 158585424-158585424 on Build GRCh38
rs369652765 Chr.6: 158585354-158585354 on Build GRCh38
rs1016139041 Chr.6: 158585240-158585240 on Build GRCh38
rs923435754 Chr.6: 158585607-158585607 on Build GRCh38
rs1015765879 Chr.6: 158585517-158585517 on Build GRCh38
rs752954019 Chr.6: 158585518-158585518 on Build GRCh38
rs755176803 Chr.6: 158585348-158585349 on Build GRCh38
rs549155771 Chr.6: 158585627-158585627 on Build GRCh38
rs372779214 Chr.6: 158585307-158585307 on Build GRCh38
rs371922418 Chr.6: 158585366-158585366 on Build GRCh38
rs773550349 Chr.6: 158585378-158585378 on Build GRCh38
rs565772467 Chr.6: 158585312-158585312 on Build GRCh38
rs773451593 Chr.6: 158585553-158585554 on Build GRCh38
rs529287135 Chr.6: 158585287-158585287 on Build GRCh38
rs201315962 Chr.6: 158585263-158585263 on Build GRCh38
rs962486139 Chr.6: 158585522-158585522 on Build GRCh38
rs537311620 Chr.6: 158585401-158585401 on Build GRCh38
rs914795426 Chr.6: 158585349-158585349 on Build GRCh38
rs1012048479 Chr.6: 158585245-158585245 on Build GRCh38
rs146629665 Chr.6: 158585659-158585659 on Build GRCh38
rs765779991 Chr.6: 158585400-158585400 on Build GRCh38
rs767822713 Chr.6: 158585308-158585308 on Build GRCh38
rs779206999 Chr.6: 158585328-158585328 on Build GRCh38
rs201860196 Chr.6: 158585254-158585254 on Build GRCh38
rs748582934 Chr.6: 158585335-158585335 on Build GRCh38
rs780952559 Chr.6: 158585345-158585345 on Build GRCh38
rs754284199 Chr.6: 158585320-158585320 on Build GRCh38
rs755259470 Chr.6: 158585494-158585494 on Build GRCh38
rs550120392 Chr.6: 158585261-158585261 on Build GRCh38
rs1024795723 Chr.6: 158585539-158585539 on Build GRCh38
rs760819959 Chr.6: 158585392-158585392 on Build GRCh38
rs1038032071 Chr.6: 158585529-158585529 on Build GRCh38
rs199898880 Chr.6: 158585367-158585367 on Build GRCh38
rs777878100 Chr.6: 158585531-158585531 on Build GRCh38
rs376062690 Chr.6: 158585371-158585371 on Build GRCh38
rs148505276 Chr.6: 158585606-158585606 on Build GRCh38
rs780484250 Chr.6: 158585536-158585538 on Build GRCh38
rs745361509 Chr.6: 158585346-158585346 on Build GRCh38
rs986801940 Chr.6: 158585545-158585545 on Build GRCh38
rs911909798 Chr.6: 158585306-158585306 on Build GRCh38
rs145714004 Chr.6: 158585399-158585399 on Build GRCh38
rs1022311841 Chr.6: 158585582-158585582 on Build GRCh38
rs202188884 Chr.6: 158585416-158585416 on Build GRCh38
rs767620888 Chr.6: 158585377-158585377 on Build GRCh38
rs142999141 Chr.6: 158585619-158585619 on Build GRCh38
rs750930382 Chr.6: 158585269-158585269 on Build GRCh38
rs768693000 Chr.6: 158585468-158585468 on Build GRCh38
rs759131621 Chr.6: 158585281-158585281 on Build GRCh38
rs753981270 Chr.6: 158585287-158585288 on Build GRCh38
rs781138712 Chr.6: 158585444-158585444 on Build GRCh38
rs768942588 Chr.6: 158585256-158585256 on Build GRCh38
rs942040958 Chr.6: 158585676-158585676 on Build GRCh38
rs41266341 Chr.6: 158585341-158585341 on Build GRCh38
rs777960663 Chr.6: 158585512-158585512 on Build GRCh38
rs977500471 Chr.6: 158585597-158585597 on Build GRCh38
rs944706153 Chr.6: 158585572-158585572 on Build GRCh38
rs911872492 Chr.6: 158585553-158585553 on Build GRCh38
rs1024743977 Chr.6: 158585260-158585260 on Build GRCh38
rs959472250 Chr.6: 158585242-158585242 on Build GRCh38
rs755495893 Chr.6: 158585327-158585327 on Build GRCh38
rs752988045 Chr.6: 158585410-158585410 on Build GRCh38
rs964970319 Chr.6: 158585591-158585591 on Build GRCh38
rs749191902 Chr.6: 158585364-158585364 on Build GRCh38
rs757037627 Chr.6: 158585426-158585429 on Build GRCh38
rs753948509 Chr.6: 158585395-158585395 on Build GRCh38
rs769892233 Chr.6: 158585464-158585465 on Build GRCh38
rs779118688 Chr.6: 158585389-158585390 on Build GRCh38
rs923378636 Chr.6: 158585373-158585373 on Build GRCh38
rs545027105 Chr.6: 158585660-158585660 on Build GRCh38
rs775971237 Chr.6: 158585272-158585272 on Build GRCh38
rs373058809 Chr.6: 158585470-158585470 on Build GRCh38
rs948637929 Chr.6: 158585634-158585634 on Build GRCh38
rs745774124 Chr.6: 158585451-158585465 on Build GRCh38
rs957949261 Chr.6: 158585561-158585561 on Build GRCh38
rs879241415 Chr.6: 158585372-158585372 on Build GRCh38
rs779262272 Chr.6: 158585259-158585259 on Build GRCh38
rs1027846802 Chr.6: 158585527-158585527 on Build GRCh38
rs200928050 Chr.6: 158585253-158585253 on Build GRCh38
rs750449294 Chr.6: 158585310-158585310 on Build GRCh38
rs906830835 Chr.6: 158585666-158585666 on Build GRCh38
rs762993033 Chr.6: 158585288-158585288 on Build GRCh38
rs766396337 Chr.6: 158585394-158585394 on Build GRCh38
rs41266343 Chr.6: 158585439-158585439 on Build GRCh38
rs756198218 Chr.6: 158585526-158585526 on Build GRCh38
rs116646542 Chr.6: 158585495-158585495 on Build GRCh38
rs895035448 Chr.6: 158585530-158585530 on Build GRCh38
rs746753710 Chr.6: 158585416-158585417 on Build GRCh38
rs200954268 Chr.6: 158585283-158585283 on Build GRCh38
rs983504610 Chr.6: 158585628-158585628 on Build GRCh38
rs936126983 Chr.6: 158585615-158585615 on Build GRCh38

View Full Product Details