Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161548733-161549241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741239 c.1041G>T p.Q347H Chr.6: 161548896-161548896 on Build GRCh38
3622662 c.1004C>T p.P335L Chr.6: 161548933-161548933 on Build GRCh38
3622663 c.1004C>T p.P335L Chr.6: 161548933-161548933 on Build GRCh38
6470931 c.1030G>T p.E344* Chr.6: 161548907-161548907 on Build GRCh38
6938490 c.1001G>A p.R334H Chr.6: 161548936-161548936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767703008 Chr.6: 161548974-161548974 on Build GRCh38
rs146599326 Chr.6: 161548876-161548876 on Build GRCh38
rs1018001612 Chr.6: 161548873-161548873 on Build GRCh38
rs201300874 Chr.6: 161548861-161548861 on Build GRCh38
rs777171316 Chr.6: 161548836-161548836 on Build GRCh38
rs546676036 Chr.6: 161548920-161548920 on Build GRCh38
rs117828575 Chr.6: 161549145-161549145 on Build GRCh38
rs59889520 Chr.6: 161549138-161549141 on Build GRCh38
rs759364763 Chr.6: 161548982-161548982 on Build GRCh38
rs111859679 Chr.6: 161549119-161549124 on Build GRCh38
rs753638548 Chr.6: 161549121-161549124 on Build GRCh38
rs509495 Chr.6: 161549199-161549199 on Build GRCh38
rs977337396 Chr.6: 161548795-161548795 on Build GRCh38
rs763652747 Chr.6: 161548996-161548996 on Build GRCh38
rs372184805 Chr.6: 161549144-161549144 on Build GRCh38
rs386707970 Chr.6: 161549136-161549144 on Build GRCh38
rs34321960 Chr.6: 161549183-161549184 on Build GRCh38
rs756440791 Chr.6: 161549026-161549026 on Build GRCh38
rs749768565 Chr.6: 161548949-161548949 on Build GRCh38
rs916696646 Chr.6: 161549102-161549102 on Build GRCh38
rs966998800 Chr.6: 161548774-161548774 on Build GRCh38
rs878854707 Chr.6: 161548866-161548866 on Build GRCh38
rs200409552 Chr.6: 161548843-161548843 on Build GRCh38
rs990915889 Chr.6: 161549099-161549099 on Build GRCh38
rs71694927 Chr.6: 161549119-161549122 on Build GRCh38
rs577233591 Chr.6: 161549186-161549186 on Build GRCh38
rs529808032 Chr.6: 161548909-161548909 on Build GRCh38
rs757914388 Chr.6: 161548813-161548813 on Build GRCh38
rs746315190 Chr.6: 161548839-161548839 on Build GRCh38
rs746215864 Chr.6: 161548936-161548936 on Build GRCh38
rs68042180 Chr.6: 161549136-161549141 on Build GRCh38
rs527410580 Chr.6: 161549070-161549070 on Build GRCh38
rs762023080 Chr.6: 161548893-161548893 on Build GRCh38
rs544514101 Chr.6: 161548792-161548792 on Build GRCh38
rs575638693 Chr.6: 161548766-161548766 on Build GRCh38
rs761973930 Chr.6: 161548965-161548965 on Build GRCh38
rs1056432031 Chr.6: 161549167-161549167 on Build GRCh38
rs776048801 Chr.6: 161548939-161548939 on Build GRCh38
rs111675670 Chr.6: 161549198-161549198 on Build GRCh38
rs935258959 Chr.6: 161549118-161549123 on Build GRCh38
rs765187897 Chr.6: 161548911-161548911 on Build GRCh38
rs538196116 Chr.6: 161549067-161549067 on Build GRCh38
rs752636690 Chr.6: 161548914-161548914 on Build GRCh38
rs555557925 Chr.6: 161548827-161548827 on Build GRCh38
rs774445597 Chr.6: 161548951-161548951 on Build GRCh38
rs998452659 Chr.6: 161549061-161549061 on Build GRCh38
rs78593675 Chr.6: 161549138-161549138 on Build GRCh38
rs1006599682 Chr.6: 161548763-161548763 on Build GRCh38
rs971199923 Chr.6: 161549111-161549111 on Build GRCh38
rs199657839 Chr.6: 161548937-161548937 on Build GRCh38
rs763015574 Chr.6: 161548995-161548995 on Build GRCh38
rs752614495 Chr.6: 161548992-161548992 on Build GRCh38
rs559962331 Chr.6: 161548935-161548935 on Build GRCh38
rs144340740 Chr.6: 161548980-161548980 on Build GRCh38
rs386707971 Chr.6: 161549198-161549199 on Build GRCh38
rs75976935 Chr.6: 161549139-161549139 on Build GRCh38
rs919986324 Chr.6: 161549143-161549143 on Build GRCh38
rs746673460 Chr.6: 161548819-161548819 on Build GRCh38
rs867140687 Chr.6: 161548769-161548769 on Build GRCh38
rs148786996 Chr.6: 161549161-161549161 on Build GRCh38
rs768652293 Chr.6: 161548950-161548950 on Build GRCh38
rs7738794 Chr.6: 161549180-161549180 on Build GRCh38
rs745647746 Chr.6: 161548950-161548951 on Build GRCh38
rs974200767 Chr.6: 161549130-161549130 on Build GRCh38
rs200796582 Chr.6: 161549025-161549025 on Build GRCh38
rs910424061 Chr.6: 161549118-161549118 on Build GRCh38
rs751095098 Chr.6: 161548997-161548997 on Build GRCh38
rs986442757 Chr.6: 161549107-161549107 on Build GRCh38
rs954592636 Chr.6: 161549103-161549103 on Build GRCh38
rs963532958 Chr.6: 161549127-161549127 on Build GRCh38
rs756869892 Chr.6: 161549002-161549002 on Build GRCh38
rs111546299 Chr.6: 161548853-161548853 on Build GRCh38
rs923928527 Chr.6: 161549118-161549119 on Build GRCh38
rs7758433 Chr.6: 161549047-161549047 on Build GRCh38
rs982171995 Chr.6: 161549113-161549113 on Build GRCh38
rs764212924 Chr.6: 161548867-161548867 on Build GRCh38
rs919970556 Chr.6: 161548894-161548894 on Build GRCh38
rs780996466 Chr.6: 161548881-161548881 on Build GRCh38
rs141061596 Chr.6: 161548957-161548957 on Build GRCh38
rs113955887 Chr.6: 161549013-161549013 on Build GRCh38
rs746620234 Chr.6: 161548921-161548921 on Build GRCh38
rs763082278 Chr.6: 161548862-161548862 on Build GRCh38
rs777555733 Chr.6: 161548815-161548815 on Build GRCh38
rs545744886 Chr.6: 161549019-161549023 on Build GRCh38
rs757869529 Chr.6: 161548917-161548917 on Build GRCh38
rs764955994 Chr.6: 161548990-161548990 on Build GRCh38
rs866772592 Chr.6: 161549137-161549141 on Build GRCh38
rs756996581 Chr.6: 161548927-161548927 on Build GRCh38
rs923162349 Chr.6: 161548800-161548800 on Build GRCh38
rs114928122 Chr.6: 161549209-161549209 on Build GRCh38
rs780990333 Chr.6: 161549015-161549015 on Build GRCh38
rs947513188 Chr.6: 161549193-161549193 on Build GRCh38
rs766121894 Chr.6: 161548896-161548896 on Build GRCh38
rs938985620 Chr.6: 161549162-161549162 on Build GRCh38
rs894633561 Chr.6: 161548952-161548952 on Build GRCh38
rs753655327 Chr.6: 161548908-161548908 on Build GRCh38

View Full Product Details