Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20136674-20137179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927668
Entrez Gene ID
101927668
Gene Name
uncharacterized LOC101927668
Gene Aliases
-
UniGene
Hs.715900
Gene Chromosome Location
Chr.7: 19918981-20140426 on Build GRCh38
Transcripts
NR_110114.1 NM_182762.3

Gene Symbol
MACC1
Entrez Gene ID
346389
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
7A5, SH3BP4L
UniGene
Hs.598388
Gene Chromosome Location
Chr.7: 20134655-20217390 on Build GRCh38
Transcripts
NR_110114.1 NM_182762.3

SNP Detail

SNP ID Chromosome Location
rs537791267 Chr.7: 20136837-20136837 on Build GRCh38
rs921983575 Chr.7: 20136819-20136819 on Build GRCh38
rs148185597 Chr.7: 20136855-20136855 on Build GRCh38
rs941867516 Chr.7: 20136805-20136807 on Build GRCh38
rs987946416 Chr.7: 20137065-20137065 on Build GRCh38
rs4719605 Chr.7: 20136754-20136754 on Build GRCh38
rs999696587 Chr.7: 20136961-20136961 on Build GRCh38
rs1033390494 Chr.7: 20136778-20136778 on Build GRCh38
rs954037130 Chr.7: 20136713-20136713 on Build GRCh38
rs1016092769 Chr.7: 20136988-20136988 on Build GRCh38
rs567580691 Chr.7: 20136757-20136757 on Build GRCh38
rs904678575 Chr.7: 20136764-20136764 on Build GRCh38
rs931433321 Chr.7: 20136797-20136797 on Build GRCh38
rs1031308737 Chr.7: 20136964-20136964 on Build GRCh38
rs77696507 Chr.7: 20136822-20136822 on Build GRCh38
rs558310931 Chr.7: 20137138-20137138 on Build GRCh38
rs1013736650 Chr.7: 20136932-20136932 on Build GRCh38
rs879555173 Chr.7: 20136854-20136895 on Build GRCh38
rs946096466 Chr.7: 20136837-20136840 on Build GRCh38
rs567277821 Chr.7: 20136820-20136820 on Build GRCh38
rs895220412 Chr.7: 20136811-20136811 on Build GRCh38
rs201696362 Chr.7: 20136895-20136896 on Build GRCh38
rs913349418 Chr.7: 20137080-20137081 on Build GRCh38
rs904747429 Chr.7: 20137145-20137145 on Build GRCh38
rs950438062 Chr.7: 20136785-20136785 on Build GRCh38
rs1010921803 Chr.7: 20136709-20136709 on Build GRCh38
rs1034530734 Chr.7: 20136912-20136912 on Build GRCh38
rs1019425620 Chr.7: 20137042-20137042 on Build GRCh38
rs58950132 Chr.7: 20137146-20137146 on Build GRCh38
rs987042199 Chr.7: 20137009-20137009 on Build GRCh38
rs953811735 Chr.7: 20137001-20137001 on Build GRCh38
rs950428574 Chr.7: 20136945-20136945 on Build GRCh38
rs974517467 Chr.7: 20137002-20137004 on Build GRCh38
rs957011153 Chr.7: 20136970-20136970 on Build GRCh38
rs1001738190 Chr.7: 20136766-20136766 on Build GRCh38
rs972622349 Chr.7: 20137100-20137100 on Build GRCh38
rs577631188 Chr.7: 20136873-20136873 on Build GRCh38
rs1055644481 Chr.7: 20136803-20136803 on Build GRCh38
rs912515987 Chr.7: 20136772-20136772 on Build GRCh38
rs904815914 Chr.7: 20136856-20136856 on Build GRCh38
rs1028518921 Chr.7: 20136994-20136994 on Build GRCh38
rs1014206371 Chr.7: 20136826-20136826 on Build GRCh38
rs1001669699 Chr.7: 20136870-20136870 on Build GRCh38
rs191710818 Chr.7: 20136710-20136710 on Build GRCh38
rs535453048 Chr.7: 20136918-20136918 on Build GRCh38
rs919897134 Chr.7: 20136796-20136796 on Build GRCh38
rs868561384 Chr.7: 20136903-20136903 on Build GRCh38
rs1051743875 Chr.7: 20136830-20136830 on Build GRCh38
rs1021907193 Chr.7: 20136730-20136730 on Build GRCh38
rs894872044 Chr.7: 20136923-20136925 on Build GRCh38
rs552701918 Chr.7: 20137017-20137019 on Build GRCh38
rs986636651 Chr.7: 20136750-20136750 on Build GRCh38
rs78998751 Chr.7: 20136784-20136784 on Build GRCh38
rs1011222607 Chr.7: 20136981-20136981 on Build GRCh38
rs974548454 Chr.7: 20136814-20136814 on Build GRCh38
rs1024756291 Chr.7: 20136939-20136939 on Build GRCh38
rs961297461 Chr.7: 20137090-20137090 on Build GRCh38
rs939881519 Chr.7: 20136779-20136779 on Build GRCh38
rs371745012 Chr.7: 20137143-20137143 on Build GRCh38
rs891771412 Chr.7: 20136834-20136840 on Build GRCh38

View Full Product Details