Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156310172-156310716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT3
Entrez Gene ID
7203
Gene Name
chaperonin containing TCP1 subunit 3
Gene Aliases
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5
UniGene
Hs.491494
Gene Chromosome Location
Chr.1: 156308961-156338415 on Build GRCh38
Transcripts
NM_001008800.2 NM_005998.4 NR_036564.1 NR_036565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297964 c.1467C>A p.D489E Chr.1: 156310624-156310624 on Build GRCh38
1472749 c.1447G>A p.E483K Chr.1: 156310644-156310644 on Build GRCh38
1983320 c.1415A>T p.Q472L Chr.1: 156310676-156310676 on Build GRCh38
1983319 c.1452G>A p.T484T Chr.1: 156310639-156310639 on Build GRCh38
4527298 c.1453G>A p.G485S Chr.1: 156310638-156310638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774502448 Chr.1: 156310651-156310651 on Build GRCh38
rs879187720 Chr.1: 156310691-156310691 on Build GRCh38
rs562425485 Chr.1: 156310516-156310516 on Build GRCh38
rs141644009 Chr.1: 156310623-156310623 on Build GRCh38
rs11582672 Chr.1: 156310509-156310509 on Build GRCh38
rs185274336 Chr.1: 156310336-156310336 on Build GRCh38
rs1015164094 Chr.1: 156310537-156310537 on Build GRCh38
rs764984004 Chr.1: 156310692-156310692 on Build GRCh38
rs576024215 Chr.1: 156310485-156310485 on Build GRCh38
rs911887771 Chr.1: 156310431-156310431 on Build GRCh38
rs1015214958 Chr.1: 156310265-156310265 on Build GRCh38
rs769723527 Chr.1: 156310548-156310548 on Build GRCh38
rs527340114 Chr.1: 156310568-156310568 on Build GRCh38
rs777799358 Chr.1: 156310640-156310640 on Build GRCh38
rs772287582 Chr.1: 156310660-156310660 on Build GRCh38
rs111507501 Chr.1: 156310532-156310532 on Build GRCh38
rs905225668 Chr.1: 156310218-156310218 on Build GRCh38
rs778815617 Chr.1: 156310515-156310515 on Build GRCh38
rs993947794 Chr.1: 156310387-156310387 on Build GRCh38
rs973618249 Chr.1: 156310300-156310300 on Build GRCh38
rs560092480 Chr.1: 156310550-156310550 on Build GRCh38
rs898081880 Chr.1: 156310538-156310538 on Build GRCh38
rs771132284 Chr.1: 156310647-156310647 on Build GRCh38
rs749536653 Chr.1: 156310512-156310514 on Build GRCh38
rs753082041 Chr.1: 156310576-156310576 on Build GRCh38
rs776110119 Chr.1: 156310662-156310662 on Build GRCh38
rs753581876 Chr.1: 156310609-156310609 on Build GRCh38
rs372385648 Chr.1: 156310326-156310326 on Build GRCh38
rs899254645 Chr.1: 156310368-156310368 on Build GRCh38
rs148953041 Chr.1: 156310487-156310487 on Build GRCh38
rs149766764 Chr.1: 156310262-156310262 on Build GRCh38
rs771324645 Chr.1: 156310565-156310565 on Build GRCh38
rs927246248 Chr.1: 156310549-156310549 on Build GRCh38
rs145728345 Chr.1: 156310345-156310345 on Build GRCh38
rs948450807 Chr.1: 156310506-156310506 on Build GRCh38
rs115667984 Chr.1: 156310533-156310533 on Build GRCh38
rs376398280 Chr.1: 156310639-156310639 on Build GRCh38
rs757921171 Chr.1: 156310516-156310517 on Build GRCh38
rs749579352 Chr.1: 156310646-156310646 on Build GRCh38
rs1047828522 Chr.1: 156310629-156310629 on Build GRCh38
rs564643782 Chr.1: 156310443-156310443 on Build GRCh38
rs772032511 Chr.1: 156310523-156310523 on Build GRCh38
rs575241696 Chr.1: 156310366-156310366 on Build GRCh38
rs745866924 Chr.1: 156310520-156310520 on Build GRCh38
rs67432426 Chr.1: 156310516-156310519 on Build GRCh38
rs536043676 Chr.1: 156310393-156310393 on Build GRCh38
rs746250638 Chr.1: 156310653-156310653 on Build GRCh38
rs201084244 Chr.1: 156310505-156310505 on Build GRCh38
rs532186433 Chr.1: 156310348-156310348 on Build GRCh38
rs750030740 Chr.1: 156310516-156310516 on Build GRCh38
rs763139918 Chr.1: 156310563-156310563 on Build GRCh38
rs867370775 Chr.1: 156310672-156310672 on Build GRCh38
rs779474289 Chr.1: 156310517-156310520 on Build GRCh38
rs545988558 Chr.1: 156310425-156310425 on Build GRCh38
rs72414608 Chr.1: 156310506-156310509 on Build GRCh38
rs76353961 Chr.1: 156310663-156310663 on Build GRCh38
rs113676483 Chr.1: 156310539-156310539 on Build GRCh38
rs761348385 Chr.1: 156310681-156310681 on Build GRCh38
rs1023970737 Chr.1: 156310224-156310224 on Build GRCh38
rs765182056 Chr.1: 156310617-156310617 on Build GRCh38
rs780070541 Chr.1: 156310625-156310625 on Build GRCh38
rs760023454 Chr.1: 156310570-156310570 on Build GRCh38
rs539960593 Chr.1: 156310426-156310426 on Build GRCh38
rs977293873 Chr.1: 156310459-156310459 on Build GRCh38
rs957302689 Chr.1: 156310685-156310685 on Build GRCh38
rs199530951 Chr.1: 156310619-156310619 on Build GRCh38
rs768086456 Chr.1: 156310573-156310573 on Build GRCh38
rs955819038 Chr.1: 156310503-156310503 on Build GRCh38
rs962496400 Chr.1: 156310283-156310283 on Build GRCh38
rs965310292 Chr.1: 156310483-156310483 on Build GRCh38
rs763672947 Chr.1: 156310590-156310590 on Build GRCh38
rs944732618 Chr.1: 156310432-156310432 on Build GRCh38
rs554091650 Chr.1: 156310418-156310418 on Build GRCh38
rs760251758 Chr.1: 156310582-156310582 on Build GRCh38
rs748223228 Chr.1: 156310404-156310404 on Build GRCh38
rs763760969 Chr.1: 156310676-156310676 on Build GRCh38
rs1044276973 Chr.1: 156310208-156310208 on Build GRCh38
rs778572747 Chr.1: 156310513-156310520 on Build GRCh38
rs138435772 Chr.1: 156310615-156310615 on Build GRCh38
rs750256353 Chr.1: 156310618-156310618 on Build GRCh38
rs572453567 Chr.1: 156310419-156310419 on Build GRCh38
rs906908018 Chr.1: 156310239-156310239 on Build GRCh38
rs369168014 Chr.1: 156310679-156310679 on Build GRCh38
rs746873180 Chr.1: 156310631-156310631 on Build GRCh38
rs772112744 Chr.1: 156310516-156310518 on Build GRCh38

View Full Product Details