Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156325016-156325561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT3
Entrez Gene ID
7203
Gene Name
chaperonin containing TCP1 subunit 3
Gene Aliases
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5
UniGene
Hs.491494
Gene Chromosome Location
Chr.1: 156308961-156338415 on Build GRCh38
Transcripts
NM_001008800.2 NM_005998.4 NR_036564.1 NR_036565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3862966 c.336C>T p.F112F Chr.1: 156325058-156325058 on Build GRCh38
897200 c.358A>G p.T120A Chr.1: 156325036-156325036 on Build GRCh38
3788823 c.354C>G p.H118Q Chr.1: 156325040-156325040 on Build GRCh38
3984317 c.327T>C p.A109A Chr.1: 156325067-156325067 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1021615647 Chr.1: 156325154-156325154 on Build GRCh38
rs745978534 Chr.1: 156325083-156325083 on Build GRCh38
rs764221181 Chr.1: 156325120-156325120 on Build GRCh38
rs138140920 Chr.1: 156325076-156325076 on Build GRCh38
rs543061678 Chr.1: 156325277-156325277 on Build GRCh38
rs968693054 Chr.1: 156325152-156325156 on Build GRCh38
rs572134994 Chr.1: 156325092-156325092 on Build GRCh38
rs11548199 Chr.1: 156325042-156325042 on Build GRCh38
rs760973174 Chr.1: 156325113-156325114 on Build GRCh38
rs181637252 Chr.1: 156325252-156325252 on Build GRCh38
rs1030523436 Chr.1: 156325057-156325057 on Build GRCh38
rs757750753 Chr.1: 156325250-156325250 on Build GRCh38
rs529565341 Chr.1: 156325429-156325429 on Build GRCh38
rs951172692 Chr.1: 156325258-156325258 on Build GRCh38
rs781121153 Chr.1: 156325074-156325074 on Build GRCh38
rs776988098 Chr.1: 156325111-156325111 on Build GRCh38
rs1048078486 Chr.1: 156325386-156325386 on Build GRCh38
rs948967304 Chr.1: 156325330-156325330 on Build GRCh38
rs541193594 Chr.1: 156325467-156325467 on Build GRCh38
rs112977402 Chr.1: 156325483-156325483 on Build GRCh38
rs1034240962 Chr.1: 156325185-156325185 on Build GRCh38
rs909565356 Chr.1: 156325396-156325396 on Build GRCh38
rs756520982 Chr.1: 156325455-156325455 on Build GRCh38
rs866303436 Chr.1: 156325058-156325058 on Build GRCh38
rs754852082 Chr.1: 156325051-156325051 on Build GRCh38
rs756123714 Chr.1: 156325079-156325079 on Build GRCh38
rs374403389 Chr.1: 156325140-156325140 on Build GRCh38
rs968816333 Chr.1: 156325180-156325180 on Build GRCh38
rs1001475701 Chr.1: 156325529-156325529 on Build GRCh38
rs562362639 Chr.1: 156325424-156325424 on Build GRCh38
rs1011224936 Chr.1: 156325507-156325507 on Build GRCh38
rs1022009445 Chr.1: 156325141-156325141 on Build GRCh38
rs977515513 Chr.1: 156325244-156325244 on Build GRCh38
rs1039418199 Chr.1: 156325352-156325352 on Build GRCh38
rs909517405 Chr.1: 156325303-156325303 on Build GRCh38
rs145655992 Chr.1: 156325082-156325082 on Build GRCh38
rs904542781 Chr.1: 156325520-156325520 on Build GRCh38
rs959974262 Chr.1: 156325233-156325233 on Build GRCh38
rs1036746555 Chr.1: 156325449-156325449 on Build GRCh38
rs751141406 Chr.1: 156325040-156325040 on Build GRCh38
rs776028346 Chr.1: 156325102-156325103 on Build GRCh38
rs1050021765 Chr.1: 156325492-156325492 on Build GRCh38
rs1052356526 Chr.1: 156325400-156325400 on Build GRCh38
rs750711830 Chr.1: 156325436-156325436 on Build GRCh38
rs367585473 Chr.1: 156325133-156325133 on Build GRCh38
rs565712232 Chr.1: 156325495-156325495 on Build GRCh38
rs575983236 Chr.1: 156325271-156325271 on Build GRCh38
rs1043163746 Chr.1: 156325497-156325497 on Build GRCh38
rs545897067 Chr.1: 156325170-156325170 on Build GRCh38
rs368599989 Chr.1: 156325241-156325241 on Build GRCh38
rs747188687 Chr.1: 156325107-156325107 on Build GRCh38
rs1022189745 Chr.1: 156325514-156325514 on Build GRCh38
rs547305241 Chr.1: 156325329-156325329 on Build GRCh38
rs891872484 Chr.1: 156325093-156325093 on Build GRCh38
rs752089213 Chr.1: 156325136-156325136 on Build GRCh38
rs933875104 Chr.1: 156325472-156325472 on Build GRCh38
rs769079494 Chr.1: 156325108-156325108 on Build GRCh38

View Full Product Details