Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27142966-27143470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_019102.3

Gene Symbol
HOXA3
Entrez Gene ID
3200
Gene Name
homeobox A3
Gene Aliases
HOX1, HOX1E
UniGene
Hs.659337
Gene Chromosome Location
Chr.7: 27106190-27152581 on Build GRCh38
Transcripts
NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_019102.3

Gene Symbol
HOXA5
Entrez Gene ID
3202
Gene Name
homeobox A5
Gene Aliases
HOX1, HOX1.3, HOX1C
UniGene
Hs.655218
Gene Chromosome Location
Chr.7: 27141052-27143668 on Build GRCh38
Transcripts
NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_019102.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192029 c.279G>T p.T93T Chr.7: 27143329-27143329 on Build GRCh38
302386 c.349G>T p.G117C Chr.7: 27143259-27143259 on Build GRCh38
452918 c.492C>A p.S164R Chr.7: 27143116-27143116 on Build GRCh38
7088932 c.498G>T p.P166P Chr.7: 27143110-27143110 on Build GRCh38
5982492 c.366A>G p.L122L Chr.7: 27143242-27143242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768853015 Chr.7: 27143125-27143125 on Build GRCh38
rs138895978 Chr.7: 27143440-27143440 on Build GRCh38
rs776457173 Chr.7: 27143238-27143238 on Build GRCh38
rs756462084 Chr.7: 27143284-27143284 on Build GRCh38
rs760237815 Chr.7: 27143086-27143086 on Build GRCh38
rs778118565 Chr.7: 27143442-27143442 on Build GRCh38
rs760180381 Chr.7: 27143413-27143413 on Build GRCh38
rs749344683 Chr.7: 27143191-27143191 on Build GRCh38
rs764768499 Chr.7: 27143257-27143257 on Build GRCh38
rs779199840 Chr.7: 27143302-27143302 on Build GRCh38
rs776140545 Chr.7: 27143153-27143153 on Build GRCh38
rs747387275 Chr.7: 27142995-27142995 on Build GRCh38
rs546604133 Chr.7: 27142992-27142992 on Build GRCh38
rs778055930 Chr.7: 27143077-27143077 on Build GRCh38
rs202017218 Chr.7: 27143128-27143128 on Build GRCh38
rs781576714 Chr.7: 27143230-27143238 on Build GRCh38
rs762450812 Chr.7: 27143380-27143380 on Build GRCh38
rs750737620 Chr.7: 27143278-27143278 on Build GRCh38
rs910381369 Chr.7: 27142988-27142988 on Build GRCh38
rs899055964 Chr.7: 27143010-27143010 on Build GRCh38
rs762654452 Chr.7: 27143272-27143272 on Build GRCh38
rs770679132 Chr.7: 27143348-27143348 on Build GRCh38
rs778171938 Chr.7: 27143332-27143332 on Build GRCh38
rs200748640 Chr.7: 27143329-27143329 on Build GRCh38
rs773252225 Chr.7: 27143131-27143131 on Build GRCh38
rs374569929 Chr.7: 27143431-27143431 on Build GRCh38
rs763754207 Chr.7: 27143277-27143277 on Build GRCh38
rs150111660 Chr.7: 27143099-27143099 on Build GRCh38
rs920471143 Chr.7: 27143411-27143411 on Build GRCh38
rs1055240029 Chr.7: 27143313-27143313 on Build GRCh38
rs746793307 Chr.7: 27143233-27143233 on Build GRCh38
rs745901959 Chr.7: 27143012-27143012 on Build GRCh38
rs201542016 Chr.7: 27143271-27143271 on Build GRCh38
rs759000941 Chr.7: 27143154-27143154 on Build GRCh38
rs368436023 Chr.7: 27143293-27143293 on Build GRCh38
rs952745648 Chr.7: 27143365-27143365 on Build GRCh38
rs952573601 Chr.7: 27143040-27143040 on Build GRCh38
rs771987984 Chr.7: 27143226-27143226 on Build GRCh38
rs746769658 Chr.7: 27143335-27143335 on Build GRCh38
rs1027345422 Chr.7: 27143035-27143035 on Build GRCh38
rs373725292 Chr.7: 27143319-27143319 on Build GRCh38
rs766695975 Chr.7: 27143400-27143400 on Build GRCh38
rs769277822 Chr.7: 27143377-27143377 on Build GRCh38
rs755051776 Chr.7: 27143300-27143300 on Build GRCh38
rs370767951 Chr.7: 27143110-27143110 on Build GRCh38
rs1035739395 Chr.7: 27143090-27143090 on Build GRCh38
rs965206722 Chr.7: 27143424-27143424 on Build GRCh38
rs774919421 Chr.7: 27143378-27143378 on Build GRCh38
rs767519501 Chr.7: 27143058-27143058 on Build GRCh38
rs983024121 Chr.7: 27143108-27143108 on Build GRCh38
rs774133906 Chr.7: 27143026-27143026 on Build GRCh38
rs773901396 Chr.7: 27143388-27143388 on Build GRCh38
rs377343707 Chr.7: 27143111-27143111 on Build GRCh38
rs995285363 Chr.7: 27143362-27143362 on Build GRCh38
rs200385351 Chr.7: 27143013-27143013 on Build GRCh38
rs754368141 Chr.7: 27143410-27143410 on Build GRCh38
rs751882772 Chr.7: 27143092-27143092 on Build GRCh38
rs770861655 Chr.7: 27143237-27143237 on Build GRCh38
rs748418865 Chr.7: 27143219-27143219 on Build GRCh38
rs73290304 Chr.7: 27143014-27143014 on Build GRCh38
rs759065939 Chr.7: 27143252-27143252 on Build GRCh38
rs781688061 Chr.7: 27142999-27142999 on Build GRCh38
rs1047218 Chr.7: 27143383-27143383 on Build GRCh38
rs757614432 Chr.7: 27143196-27143198 on Build GRCh38
rs200819149 Chr.7: 27143414-27143414 on Build GRCh38
rs761081495 Chr.7: 27143397-27143397 on Build GRCh38
rs370086397 Chr.7: 27143443-27143443 on Build GRCh38
rs765793664 Chr.7: 27143152-27143152 on Build GRCh38
rs1021811483 Chr.7: 27143114-27143114 on Build GRCh38
rs772039301 Chr.7: 27143130-27143130 on Build GRCh38
rs768261566 Chr.7: 27143382-27143382 on Build GRCh38
rs765953050 Chr.7: 27143087-27143087 on Build GRCh38
rs756590217 Chr.7: 27143180-27143180 on Build GRCh38
rs781649860 Chr.7: 27143100-27143100 on Build GRCh38
rs746416761 Chr.7: 27143105-27143105 on Build GRCh38
rs768326607 Chr.7: 27143019-27143019 on Build GRCh38
rs755032149 Chr.7: 27143196-27143196 on Build GRCh38
rs760731020 Chr.7: 27143150-27143150 on Build GRCh38
rs930510004 Chr.7: 27143129-27143130 on Build GRCh38
rs35281041 Chr.7: 27143209-27143210 on Build GRCh38
rs780424975 Chr.7: 27143181-27143181 on Build GRCh38
rs774741194 Chr.7: 27143126-27143126 on Build GRCh38
rs1042959456 Chr.7: 27143282-27143282 on Build GRCh38
rs750405781 Chr.7: 27143074-27143074 on Build GRCh38
rs965111568 Chr.7: 27143116-27143116 on Build GRCh38
rs757590045 Chr.7: 27143170-27143170 on Build GRCh38
rs535686511 Chr.7: 27143088-27143088 on Build GRCh38
rs761701779 Chr.7: 27143029-27143029 on Build GRCh38
rs376356041 Chr.7: 27143422-27143422 on Build GRCh38
rs767938696 Chr.7: 27143173-27143173 on Build GRCh38
rs773199075 Chr.7: 27143229-27143229 on Build GRCh38
rs748367476 Chr.7: 27143309-27143309 on Build GRCh38
rs750919367 Chr.7: 27143179-27143179 on Build GRCh38
rs758573284 Chr.7: 27143428-27143428 on Build GRCh38
rs776598451 Chr.7: 27143364-27143364 on Build GRCh38
rs373032286 Chr.7: 27143065-27143065 on Build GRCh38
rs764896847 Chr.7: 27143160-27143160 on Build GRCh38
rs752398373 Chr.7: 27143161-27143161 on Build GRCh38
rs779252915 Chr.7: 27143197-27143197 on Build GRCh38
rs1054035547 Chr.7: 27143209-27143209 on Build GRCh38
rs771361387 Chr.7: 27142996-27142996 on Build GRCh38
rs752758628 Chr.7: 27143425-27143425 on Build GRCh38

View Full Product Details