Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27163149-27163660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA10-HOXA9
Entrez Gene ID
100534589
Gene Name
HOXA10-HOXA9 readthrough
Gene Aliases
-
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27180261 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

Gene Symbol
HOXA9
Entrez Gene ID
3205
Gene Name
homeobox A9
Gene Aliases
ABD-B, HOX1, HOX1.7, HOX1G
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27165530 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142958 c.792C>A p.I264I Chr.7: 27163630-27163630 on Build GRCh38
6486108 c.802C>A p.R268R Chr.7: 27163620-27163620 on Build GRCh38
1673075 c.802C>T p.R268* Chr.7: 27163620-27163620 on Build GRCh38
1088876 c.770G>T p.R257L Chr.7: 27163652-27163652 on Build GRCh38
3832582 c.792C>T p.I264I Chr.7: 27163630-27163630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17500987 Chr.7: 27163566-27163566 on Build GRCh38
rs34248544 Chr.7: 27163427-27163428 on Build GRCh38
rs1051807454 Chr.7: 27163521-27163521 on Build GRCh38
rs575670067 Chr.7: 27163599-27163599 on Build GRCh38
rs750730778 Chr.7: 27163611-27163611 on Build GRCh38
rs564237365 Chr.7: 27163568-27163568 on Build GRCh38
rs893680138 Chr.7: 27163347-27163347 on Build GRCh38
rs17472140 Chr.7: 27163576-27163576 on Build GRCh38
rs576948299 Chr.7: 27163381-27163381 on Build GRCh38
rs915075553 Chr.7: 27163512-27163512 on Build GRCh38
rs938241459 Chr.7: 27163540-27163540 on Build GRCh38
rs953563449 Chr.7: 27163391-27163391 on Build GRCh38
rs566327028 Chr.7: 27163305-27163305 on Build GRCh38
rs545859037 Chr.7: 27163389-27163389 on Build GRCh38
rs559732995 Chr.7: 27163489-27163489 on Build GRCh38
rs964007244 Chr.7: 27163440-27163440 on Build GRCh38
rs963892339 Chr.7: 27163457-27163457 on Build GRCh38
rs1032584712 Chr.7: 27163488-27163488 on Build GRCh38
rs752234662 Chr.7: 27163475-27163475 on Build GRCh38
rs145028851 Chr.7: 27163181-27163181 on Build GRCh38
rs189587233 Chr.7: 27163530-27163530 on Build GRCh38
rs757393231 Chr.7: 27163600-27163600 on Build GRCh38
rs879460651 Chr.7: 27163455-27163455 on Build GRCh38
rs996761524 Chr.7: 27163482-27163482 on Build GRCh38
rs373195636 Chr.7: 27163596-27163596 on Build GRCh38
rs972102728 Chr.7: 27163255-27163255 on Build GRCh38
rs185084360 Chr.7: 27163374-27163374 on Build GRCh38
rs200749905 Chr.7: 27163346-27163347 on Build GRCh38
rs896723340 Chr.7: 27163352-27163352 on Build GRCh38
rs866292228 Chr.7: 27163445-27163445 on Build GRCh38
rs751197378 Chr.7: 27163602-27163602 on Build GRCh38
rs1007650059 Chr.7: 27163392-27163392 on Build GRCh38
rs771692391 Chr.7: 27163556-27163556 on Build GRCh38
rs762038103 Chr.7: 27163249-27163249 on Build GRCh38
rs778960235 Chr.7: 27163636-27163636 on Build GRCh38
rs981876744 Chr.7: 27163496-27163496 on Build GRCh38
rs1037987085 Chr.7: 27163544-27163544 on Build GRCh38
rs377402962 Chr.7: 27163630-27163630 on Build GRCh38
rs546588205 Chr.7: 27163301-27163301 on Build GRCh38
rs903203776 Chr.7: 27163517-27163517 on Build GRCh38
rs539510908 Chr.7: 27163238-27163238 on Build GRCh38
rs765701054 Chr.7: 27163302-27163302 on Build GRCh38
rs554957950 Chr.7: 27163354-27163354 on Build GRCh38
rs763304806 Chr.7: 27163591-27163591 on Build GRCh38
rs765846663 Chr.7: 27163305-27163306 on Build GRCh38
rs918929213 Chr.7: 27163270-27163274 on Build GRCh38
rs573134752 Chr.7: 27163506-27163506 on Build GRCh38
rs553750216 Chr.7: 27163368-27163368 on Build GRCh38
rs552713334 Chr.7: 27163498-27163498 on Build GRCh38
rs537259388 Chr.7: 27163372-27163372 on Build GRCh38
rs970476251 Chr.7: 27163513-27163513 on Build GRCh38
rs957923556 Chr.7: 27163492-27163492 on Build GRCh38
rs542147750 Chr.7: 27163510-27163510 on Build GRCh38
rs1008824847 Chr.7: 27163406-27163406 on Build GRCh38
rs749137808 Chr.7: 27163628-27163628 on Build GRCh38
rs925146417 Chr.7: 27163188-27163188 on Build GRCh38
rs776708316 Chr.7: 27163240-27163240 on Build GRCh38
rs779925283 Chr.7: 27163621-27163621 on Build GRCh38
rs7810502 Chr.7: 27163520-27163520 on Build GRCh38
rs112752252 Chr.7: 27163216-27163216 on Build GRCh38
rs913626050 Chr.7: 27163507-27163507 on Build GRCh38
rs535353485 Chr.7: 27163353-27163353 on Build GRCh38
rs761410366 Chr.7: 27163553-27163553 on Build GRCh38
rs779788025 Chr.7: 27163327-27163327 on Build GRCh38
rs1047830842 Chr.7: 27163398-27163398 on Build GRCh38
rs879155081 Chr.7: 27163178-27163178 on Build GRCh38
rs767844804 Chr.7: 27163609-27163609 on Build GRCh38
rs1017569381 Chr.7: 27163438-27163438 on Build GRCh38
rs375518739 Chr.7: 27163177-27163177 on Build GRCh38
rs890555446 Chr.7: 27163403-27163403 on Build GRCh38
rs756418004 Chr.7: 27163620-27163620 on Build GRCh38
rs41316444 Chr.7: 27163360-27163360 on Build GRCh38
rs750779480 Chr.7: 27163320-27163320 on Build GRCh38
rs988275681 Chr.7: 27163494-27163494 on Build GRCh38
rs755864938 Chr.7: 27163308-27163308 on Build GRCh38
rs368820871 Chr.7: 27163179-27163179 on Build GRCh38
rs772891366 Chr.7: 27163561-27163561 on Build GRCh38

View Full Product Details