Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43625999-43626511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
UniGene
Hs.268887
Gene Chromosome Location
Chr.7: 43608452-43729541 on Build GRCh38
Transcripts
NR_135580.1 NR_135581.1 NR_135582.1 NR_135583.1 XM_005249807.4 XM_006715752.2 XM_017012414.1 XM_017012415.1 XM_017012416.1 XM_017012417.1 XM_017012418.1 XM_017012419.1 XM_017012421.1 XM_017012422.1 XM_017012424.1 XM_017012430.1 XM_017012431.1 XM_017012432.1 NM_004760.2 XM_017012792.1

Gene Symbol
STK17A
Entrez Gene ID
9263
Gene Name
serine/threonine kinase 17a
Gene Aliases
DRAK1
UniGene
Hs.709489
Gene Chromosome Location
Chr.7: 43583093-43627379 on Build GRCh38
Transcripts
NR_135580.1 NR_135581.1 NR_135582.1 NR_135583.1 XM_005249807.4 XM_006715752.2 XM_017012414.1 XM_017012415.1 XM_017012416.1 XM_017012417.1 XM_017012418.1 XM_017012419.1 XM_017012421.1 XM_017012422.1 XM_017012424.1 XM_017012430.1 XM_017012431.1 XM_017012432.1 NM_004760.2 XM_017012792.1

SNP Detail

SNP ID Chromosome Location
rs762609574 Chr.7: 43626383-43626383 on Build GRCh38
rs542812173 Chr.7: 43626145-43626154 on Build GRCh38
rs1047348771 Chr.7: 43626334-43626334 on Build GRCh38
rs561516695 Chr.7: 43626035-43626035 on Build GRCh38
rs770964191 Chr.7: 43626052-43626052 on Build GRCh38
rs1047012430 Chr.7: 43626241-43626241 on Build GRCh38
rs370929471 Chr.7: 43626121-43626121 on Build GRCh38
rs2108411 Chr.7: 43626209-43626209 on Build GRCh38
rs534021297 Chr.7: 43626269-43626269 on Build GRCh38
rs796727703 Chr.7: 43626208-43626209 on Build GRCh38
rs977332162 Chr.7: 43626202-43626202 on Build GRCh38
rs1009193359 Chr.7: 43626388-43626388 on Build GRCh38
rs1029918801 Chr.7: 43626474-43626475 on Build GRCh38
rs994827526 Chr.7: 43626331-43626331 on Build GRCh38
rs953398710 Chr.7: 43626057-43626057 on Build GRCh38
rs139973881 Chr.7: 43626058-43626058 on Build GRCh38
rs149863507 Chr.7: 43626289-43626289 on Build GRCh38
rs894260808 Chr.7: 43626230-43626230 on Build GRCh38
rs1005536802 Chr.7: 43626384-43626384 on Build GRCh38
rs533314876 Chr.7: 43626149-43626149 on Build GRCh38
rs1041401013 Chr.7: 43626306-43626306 on Build GRCh38
rs1018873527 Chr.7: 43626415-43626415 on Build GRCh38
rs748205616 Chr.7: 43626045-43626045 on Build GRCh38
rs945713312 Chr.7: 43626278-43626278 on Build GRCh38
rs972101483 Chr.7: 43626154-43626154 on Build GRCh38
rs1056727705 Chr.7: 43626256-43626256 on Build GRCh38
rs768217860 Chr.7: 43626407-43626407 on Build GRCh38
rs781329949 Chr.7: 43626191-43626191 on Build GRCh38
rs961993257 Chr.7: 43626457-43626457 on Build GRCh38
rs904266435 Chr.7: 43626319-43626319 on Build GRCh38
rs35427606 Chr.7: 43626029-43626030 on Build GRCh38
rs189679831 Chr.7: 43626098-43626098 on Build GRCh38
rs1028853867 Chr.7: 43626043-43626043 on Build GRCh38
rs569469470 Chr.7: 43626117-43626117 on Build GRCh38
rs962044777 Chr.7: 43626141-43626141 on Build GRCh38
rs939572327 Chr.7: 43626220-43626220 on Build GRCh38
rs566585659 Chr.7: 43626245-43626245 on Build GRCh38
rs1050661292 Chr.7: 43626370-43626370 on Build GRCh38
rs930747483 Chr.7: 43626190-43626190 on Build GRCh38
rs906795945 Chr.7: 43626027-43626027 on Build GRCh38
rs1018211583 Chr.7: 43626386-43626386 on Build GRCh38
rs920673339 Chr.7: 43626183-43626183 on Build GRCh38
rs999971401 Chr.7: 43626335-43626335 on Build GRCh38
rs952672741 Chr.7: 43626061-43626061 on Build GRCh38
rs964082752 Chr.7: 43626142-43626142 on Build GRCh38
rs545569187 Chr.7: 43626197-43626197 on Build GRCh38
rs935694335 Chr.7: 43626218-43626218 on Build GRCh38
rs967199011 Chr.7: 43626484-43626484 on Build GRCh38
rs756266378 Chr.7: 43626087-43626087 on Build GRCh38
rs527783926 Chr.7: 43626268-43626268 on Build GRCh38
rs777312714 Chr.7: 43626406-43626406 on Build GRCh38

View Full Product Details