Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44228710-44229212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2B
Entrez Gene ID
816
Gene Name
calcium/calmodulin dependent protein kinase II beta
Gene Aliases
CAM2, CAMK2, CAMKB
UniGene
Hs.351887
Gene Chromosome Location
Chr.7: 44217150-44325994 on Build GRCh38
Transcripts
NM_001220.4 NM_001293170.1 NM_172078.2 NM_172079.2 NM_172080.2 NM_172081.2 NM_172082.2 NM_172083.2 NM_172084.2 XM_005249862.3 XM_005249864.3 XM_006715781.2 XM_006715784.2 XM_011515547.1 XM_011515549.1 XM_011515550.1 XM_011515551.1 XM_011515552.1 XM_011515553.1 XM_011515554.1 XM_011515555.1 XM_011515556.1 XM_011515557.1 XM_011515558.2 XM_011515559.1 XM_017012660.1 XM_017012661.1 XM_017012662.1 XM_017012663.1 XM_017012664.1 XM_017012665.1 XM_017012666.1 XM_017012667.1 XM_017012668.1 XM_017012669.1 XM_017012670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4385926 c.1425_1426insC p.C477fs*19 Chr.7: 44228838-44228839 on Build GRCh38
6177791 c.1418G>C p.G473A Chr.7: 44228846-44228846 on Build GRCh38
4424051 c.1424_1425insC p.P476fs*20 Chr.7: 44228839-44228840 on Build GRCh38
20487 c.1466C>T p.P489L Chr.7: 44228798-44228798 on Build GRCh38
3747718 c.1419G>C p.G473G Chr.7: 44228845-44228845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574003384 Chr.7: 44228801-44228801 on Build GRCh38
rs542942171 Chr.7: 44228889-44228889 on Build GRCh38
rs540113994 Chr.7: 44228972-44228972 on Build GRCh38
rs374476287 Chr.7: 44228865-44228865 on Build GRCh38
rs202187533 Chr.7: 44228849-44228849 on Build GRCh38
rs766378378 Chr.7: 44228960-44228960 on Build GRCh38
rs752514706 Chr.7: 44228807-44228807 on Build GRCh38
rs988580900 Chr.7: 44228956-44228956 on Build GRCh38
rs968074701 Chr.7: 44229136-44229136 on Build GRCh38
rs779136769 Chr.7: 44228830-44228830 on Build GRCh38
rs761693546 Chr.7: 44228995-44228995 on Build GRCh38
rs548434704 Chr.7: 44229087-44229087 on Build GRCh38
rs191414856 Chr.7: 44228734-44228734 on Build GRCh38
rs745379322 Chr.7: 44228925-44228925 on Build GRCh38
rs981949694 Chr.7: 44228856-44228856 on Build GRCh38
rs769180402 Chr.7: 44228784-44228784 on Build GRCh38
rs748610422 Chr.7: 44228860-44228860 on Build GRCh38
rs992767689 Chr.7: 44229173-44229173 on Build GRCh38
rs770263670 Chr.7: 44228914-44228914 on Build GRCh38
rs751148662 Chr.7: 44228802-44228802 on Build GRCh38
rs536589861 Chr.7: 44228797-44228797 on Build GRCh38
rs763809488 Chr.7: 44228872-44228872 on Build GRCh38
rs1036054095 Chr.7: 44229066-44229066 on Build GRCh38
rs747503458 Chr.7: 44228971-44228971 on Build GRCh38
rs537871427 Chr.7: 44228840-44228840 on Build GRCh38
rs897556184 Chr.7: 44229067-44229067 on Build GRCh38
rs746099954 Chr.7: 44228771-44228771 on Build GRCh38
rs772012531 Chr.7: 44228737-44228737 on Build GRCh38
rs572392865 Chr.7: 44228884-44228884 on Build GRCh38
rs886264940 Chr.7: 44228785-44228785 on Build GRCh38
rs778113158 Chr.7: 44229074-44229074 on Build GRCh38
rs373942433 Chr.7: 44228777-44228777 on Build GRCh38
rs763908351 Chr.7: 44228945-44228945 on Build GRCh38
rs751569852 Chr.7: 44229026-44229026 on Build GRCh38
rs772956306 Chr.7: 44228987-44228989 on Build GRCh38
rs898182161 Chr.7: 44229143-44229143 on Build GRCh38
rs866750196 Chr.7: 44228780-44228780 on Build GRCh38
rs759522806 Chr.7: 44229179-44229179 on Build GRCh38
rs556331685 Chr.7: 44229106-44229106 on Build GRCh38
rs377236573 Chr.7: 44228952-44228952 on Build GRCh38
rs201007866 Chr.7: 44228866-44228866 on Build GRCh38
rs749355162 Chr.7: 44228761-44228761 on Build GRCh38
rs779461512 Chr.7: 44228978-44228978 on Build GRCh38
rs896767653 Chr.7: 44229031-44229031 on Build GRCh38
rs547604701 Chr.7: 44229081-44229081 on Build GRCh38
rs760503107 Chr.7: 44228869-44228869 on Build GRCh38
rs760917459 Chr.7: 44229192-44229192 on Build GRCh38
rs775071445 Chr.7: 44228868-44228868 on Build GRCh38
rs956061727 Chr.7: 44229078-44229078 on Build GRCh38
rs770742580 Chr.7: 44229117-44229117 on Build GRCh38
rs761683356 Chr.7: 44228826-44228851 on Build GRCh38
rs766436734 Chr.7: 44229022-44229022 on Build GRCh38
rs757353069 Chr.7: 44228825-44228825 on Build GRCh38
rs780227298 Chr.7: 44229051-44229051 on Build GRCh38
rs759120479 Chr.7: 44228986-44228986 on Build GRCh38
rs938188600 Chr.7: 44229154-44229154 on Build GRCh38
rs776461764 Chr.7: 44228987-44228987 on Build GRCh38
rs188718464 Chr.7: 44229191-44229191 on Build GRCh38
rs755307162 Chr.7: 44228895-44228895 on Build GRCh38
rs778195212 Chr.7: 44228946-44228946 on Build GRCh38
rs778237962 Chr.7: 44228975-44228975 on Build GRCh38
rs549820182 Chr.7: 44228733-44228733 on Build GRCh38
rs949735810 Chr.7: 44229033-44229033 on Build GRCh38
rs775148650 Chr.7: 44228931-44228931 on Build GRCh38
rs1039244014 Chr.7: 44228908-44228908 on Build GRCh38
rs577708900 Chr.7: 44229116-44229116 on Build GRCh38
rs369308225 Chr.7: 44228947-44228947 on Build GRCh38
rs56377718 Chr.7: 44228818-44228818 on Build GRCh38
rs569865942 Chr.7: 44229088-44229088 on Build GRCh38
rs777621741 Chr.7: 44228759-44228759 on Build GRCh38
rs753741079 Chr.7: 44228874-44228874 on Build GRCh38
rs951136458 Chr.7: 44228804-44228804 on Build GRCh38
rs370501551 Chr.7: 44228955-44228955 on Build GRCh38
rs776026885 Chr.7: 44228779-44228779 on Build GRCh38
rs752976781 Chr.7: 44229061-44229061 on Build GRCh38
rs917791 Chr.7: 44229186-44229186 on Build GRCh38
rs765355723 Chr.7: 44228878-44228878 on Build GRCh38
rs760263712 Chr.7: 44229076-44229077 on Build GRCh38
rs547615227 Chr.7: 44228745-44228745 on Build GRCh38
rs777076470 Chr.7: 44228790-44228790 on Build GRCh38
rs556584792 Chr.7: 44228845-44228845 on Build GRCh38
rs764797365 Chr.7: 44229023-44229023 on Build GRCh38
rs758496859 Chr.7: 44228958-44228958 on Build GRCh38
rs773166695 Chr.7: 44229093-44229093 on Build GRCh38
rs555460132 Chr.7: 44228798-44228798 on Build GRCh38
rs141438454 Chr.7: 44228773-44228779 on Build GRCh38
rs767630911 Chr.7: 44229057-44229057 on Build GRCh38
rs781445599 Chr.7: 44228900-44228900 on Build GRCh38
rs773750991 Chr.7: 44228924-44228924 on Build GRCh38
rs773219593 Chr.7: 44229004-44229004 on Build GRCh38
rs759702082 Chr.7: 44229044-44229044 on Build GRCh38
rs753776898 Chr.7: 44228824-44228824 on Build GRCh38
rs767253246 Chr.7: 44228806-44228806 on Build GRCh38
rs755903024 Chr.7: 44228812-44228812 on Build GRCh38
rs920500162 Chr.7: 44228787-44228787 on Build GRCh38
rs769728374 Chr.7: 44228996-44228996 on Build GRCh38
rs142232269 Chr.7: 44228940-44228940 on Build GRCh38
rs771544071 Chr.7: 44228977-44228977 on Build GRCh38
rs757288207 Chr.7: 44228876-44228876 on Build GRCh38
rs565916639 Chr.7: 44229083-44229083 on Build GRCh38
rs367916709 Chr.7: 44229012-44229012 on Build GRCh38
rs752538643 Chr.7: 44228749-44228749 on Build GRCh38
rs748605908 Chr.7: 44228905-44228905 on Build GRCh38
rs1029836986 Chr.7: 44228941-44228941 on Build GRCh38
rs773875653 Chr.7: 44228864-44228864 on Build GRCh38
rs934903000 Chr.7: 44228758-44228758 on Build GRCh38
rs200217668 Chr.7: 44228848-44228848 on Build GRCh38
rs532292183 Chr.7: 44228976-44228976 on Build GRCh38
rs538798039 Chr.7: 44228744-44228744 on Build GRCh38
rs759218951 Chr.7: 44228805-44228805 on Build GRCh38
rs1018029951 Chr.7: 44228783-44228783 on Build GRCh38
rs779263051 Chr.7: 44228770-44228770 on Build GRCh38
rs772443083 Chr.7: 44228844-44228844 on Build GRCh38
rs755179163 Chr.7: 44228970-44228970 on Build GRCh38
rs747212234 Chr.7: 44228890-44228890 on Build GRCh38
rs770983360 Chr.7: 44228763-44228763 on Build GRCh38
rs565975789 Chr.7: 44228782-44228782 on Build GRCh38
rs886894723 Chr.7: 44229112-44229112 on Build GRCh38
rs893430420 Chr.7: 44228894-44228894 on Build GRCh38
rs770390072 Chr.7: 44228861-44228861 on Build GRCh38
rs756099627 Chr.7: 44228751-44228751 on Build GRCh38
rs751768978 Chr.7: 44228961-44228961 on Build GRCh38
rs994603233 Chr.7: 44229073-44229073 on Build GRCh38
rs867514255 Chr.7: 44228808-44228808 on Build GRCh38
rs986514625 Chr.7: 44228793-44228793 on Build GRCh38
rs895488325 Chr.7: 44228875-44228875 on Build GRCh38
rs1012754991 Chr.7: 44229146-44229146 on Build GRCh38
rs750505899 Chr.7: 44228882-44228882 on Build GRCh38
rs756411421 Chr.7: 44229072-44229072 on Build GRCh38
rs746620577 Chr.7: 44228979-44228979 on Build GRCh38
rs768167103 Chr.7: 44228980-44228980 on Build GRCh38
rs754302669 Chr.7: 44229077-44229077 on Build GRCh38
rs538196537 Chr.7: 44229100-44229100 on Build GRCh38
rs142386444 Chr.7: 44228926-44228926 on Build GRCh38
rs774415488 Chr.7: 44229142-44229142 on Build GRCh38
rs183951594 Chr.7: 44229084-44229084 on Build GRCh38
rs767499732 Chr.7: 44228958-44228961 on Build GRCh38
rs990589125 Chr.7: 44229079-44229079 on Build GRCh38
rs374808974 Chr.7: 44228917-44228917 on Build GRCh38

View Full Product Details