Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55794147-55794648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT14
Entrez Gene ID
346288
Gene Name
septin 14
Gene Aliases
-
UniGene
Hs.453629
Gene Chromosome Location
Chr.7: 55793544-55862789 on Build GRCh38
Transcripts
NM_207366.2

SNP Detail

SNP ID Chromosome Location
rs375928195 Chr.7: 55794235-55794235 on Build GRCh38
rs1018427384 Chr.7: 55794393-55794393 on Build GRCh38
rs922235524 Chr.7: 55794438-55794438 on Build GRCh38
rs140175842 Chr.7: 55794622-55794622 on Build GRCh38
rs535133187 Chr.7: 55794490-55794490 on Build GRCh38
rs1001475609 Chr.7: 55794305-55794305 on Build GRCh38
rs942213224 Chr.7: 55794212-55794212 on Build GRCh38
rs1035676791 Chr.7: 55794309-55794309 on Build GRCh38
rs556310334 Chr.7: 55794301-55794301 on Build GRCh38
rs1010078321 Chr.7: 55794363-55794363 on Build GRCh38
rs1038187935 Chr.7: 55794261-55794261 on Build GRCh38
rs978653207 Chr.7: 55794381-55794381 on Build GRCh38
rs937402255 Chr.7: 55794549-55794549 on Build GRCh38
rs963812944 Chr.7: 55794412-55794412 on Build GRCh38
rs886574377 Chr.7: 55794201-55794201 on Build GRCh38
rs1037900615 Chr.7: 55794543-55794543 on Build GRCh38
rs1020207681 Chr.7: 55794370-55794370 on Build GRCh38
rs537912807 Chr.7: 55794193-55794193 on Build GRCh38
rs568183831 Chr.7: 55794423-55794423 on Build GRCh38
rs984072738 Chr.7: 55794480-55794480 on Build GRCh38
rs758396921 Chr.7: 55794559-55794560 on Build GRCh38
rs929638289 Chr.7: 55794459-55794459 on Build GRCh38
rs796862188 Chr.7: 55794609-55794609 on Build GRCh38
rs895786631 Chr.7: 55794619-55794619 on Build GRCh38
rs1047989908 Chr.7: 55794170-55794170 on Build GRCh38
rs968955308 Chr.7: 55794372-55794372 on Build GRCh38
rs559909949 Chr.7: 55794501-55794501 on Build GRCh38
rs756430927 Chr.7: 55794168-55794168 on Build GRCh38
rs200366169 Chr.7: 55794549-55794550 on Build GRCh38
rs777920821 Chr.7: 55794481-55794481 on Build GRCh38
rs895838984 Chr.7: 55794330-55794330 on Build GRCh38
rs796609462 Chr.7: 55794547-55794548 on Build GRCh38

View Full Product Details