Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56081801-56082301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKG1
Entrez Gene ID
5260
Gene Name
phosphorylase kinase catalytic subunit gamma 1
Gene Aliases
PHKG
UniGene
Hs.730821
Gene Chromosome Location
Chr.7: 56079927-56093450 on Build GRCh38
Transcripts
NM_001258459.1 NM_001258460.1 NM_006213.4 NR_047689.1 XM_005271772.4 XM_017012324.1 XM_017012325.1 XM_017012326.1 XM_017012327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110392 c.675C>A p.A225A Chr.7: 56082010-56082010 on Build GRCh38
3762792 c.555C>T p.C185C Chr.7: 56082246-56082246 on Build GRCh38
345982 c.720G>T p.R240S Chr.7: 56081965-56081965 on Build GRCh38
6745385 c.674delC p.P225fs*8 Chr.7: 56082223-56082223 on Build GRCh38
3639860 c.614C>T p.P205L Chr.7: 56082187-56082187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116607814 Chr.7: 56082155-56082155 on Build GRCh38
rs547904081 Chr.7: 56081932-56081932 on Build GRCh38
rs141344297 Chr.7: 56082033-56082033 on Build GRCh38
rs757583640 Chr.7: 56081957-56081957 on Build GRCh38
rs1035982932 Chr.7: 56081867-56081867 on Build GRCh38
rs777335793 Chr.7: 56082070-56082070 on Build GRCh38
rs753255332 Chr.7: 56081891-56081891 on Build GRCh38
rs778380422 Chr.7: 56081955-56081955 on Build GRCh38
rs142859782 Chr.7: 56081941-56081941 on Build GRCh38
rs572958289 Chr.7: 56081843-56081843 on Build GRCh38
rs751734916 Chr.7: 56081874-56081874 on Build GRCh38
rs762358255 Chr.7: 56082094-56082094 on Build GRCh38
rs746142464 Chr.7: 56081954-56081954 on Build GRCh38
rs749618495 Chr.7: 56081902-56081902 on Build GRCh38
rs757769249 Chr.7: 56082046-56082046 on Build GRCh38
rs375996951 Chr.7: 56082199-56082199 on Build GRCh38
rs369986453 Chr.7: 56081856-56081856 on Build GRCh38
rs769760173 Chr.7: 56082271-56082271 on Build GRCh38
rs372681025 Chr.7: 56082212-56082212 on Build GRCh38
rs752345457 Chr.7: 56082150-56082150 on Build GRCh38
rs375438215 Chr.7: 56082043-56082043 on Build GRCh38
rs201288093 Chr.7: 56082084-56082084 on Build GRCh38
rs544129411 Chr.7: 56082180-56082180 on Build GRCh38
rs372823387 Chr.7: 56081958-56081958 on Build GRCh38
rs775883923 Chr.7: 56082034-56082034 on Build GRCh38
rs369323877 Chr.7: 56082179-56082179 on Build GRCh38
rs773165951 Chr.7: 56081928-56081928 on Build GRCh38
rs374223622 Chr.7: 56082019-56082019 on Build GRCh38
rs781191589 Chr.7: 56081888-56081888 on Build GRCh38
rs749221074 Chr.7: 56082071-56082071 on Build GRCh38
rs755611668 Chr.7: 56082069-56082069 on Build GRCh38
rs774364515 Chr.7: 56082078-56082078 on Build GRCh38
rs779686431 Chr.7: 56081909-56081909 on Build GRCh38
rs17172508 Chr.7: 56082154-56082154 on Build GRCh38
rs780756936 Chr.7: 56082063-56082063 on Build GRCh38
rs1049205825 Chr.7: 56082248-56082248 on Build GRCh38
rs758449839 Chr.7: 56082238-56082238 on Build GRCh38
rs148542905 Chr.7: 56081912-56081912 on Build GRCh38
rs538193071 Chr.7: 56082039-56082039 on Build GRCh38
rs776188215 Chr.7: 56081914-56081914 on Build GRCh38
rs145703798 Chr.7: 56081911-56081911 on Build GRCh38
rs375001435 Chr.7: 56081927-56081927 on Build GRCh38
rs749246204 Chr.7: 56082010-56082010 on Build GRCh38
rs375990959 Chr.7: 56082009-56082009 on Build GRCh38
rs147654051 Chr.7: 56082221-56082221 on Build GRCh38
rs748460463 Chr.7: 56082263-56082263 on Build GRCh38
rs770826108 Chr.7: 56082076-56082076 on Build GRCh38
rs771381553 Chr.7: 56082280-56082280 on Build GRCh38
rs763432195 Chr.7: 56082279-56082279 on Build GRCh38
rs940268562 Chr.7: 56082038-56082038 on Build GRCh38
rs774056941 Chr.7: 56082020-56082020 on Build GRCh38
rs373707742 Chr.7: 56082081-56082081 on Build GRCh38
rs541582462 Chr.7: 56082239-56082239 on Build GRCh38
rs766789055 Chr.7: 56082134-56082134 on Build GRCh38
rs201139713 Chr.7: 56082168-56082168 on Build GRCh38
rs755031337 Chr.7: 56081876-56081876 on Build GRCh38
rs546618556 Chr.7: 56081832-56081832 on Build GRCh38
rs376787464 Chr.7: 56082184-56082184 on Build GRCh38
rs370265951 Chr.7: 56082057-56082057 on Build GRCh38
rs750705770 Chr.7: 56082049-56082049 on Build GRCh38
rs764754022 Chr.7: 56082134-56082134 on Build GRCh38
rs768578172 Chr.7: 56082251-56082251 on Build GRCh38
rs200728838 Chr.7: 56081896-56081896 on Build GRCh38
rs377598630 Chr.7: 56081987-56081987 on Build GRCh38
rs774424794 Chr.7: 56081908-56081913 on Build GRCh38
rs750946296 Chr.7: 56082120-56082120 on Build GRCh38
rs761035392 Chr.7: 56082087-56082087 on Build GRCh38
rs940215502 Chr.7: 56082183-56082183 on Build GRCh38
rs907065301 Chr.7: 56082085-56082085 on Build GRCh38
rs750381091 Chr.7: 56082227-56082227 on Build GRCh38
rs760588653 Chr.7: 56081922-56081922 on Build GRCh38
rs1024192978 Chr.7: 56081919-56081919 on Build GRCh38
rs551840444 Chr.7: 56082002-56082002 on Build GRCh38
rs771975915 Chr.7: 56082160-56082160 on Build GRCh38
rs192764931 Chr.7: 56082226-56082226 on Build GRCh38
rs998756099 Chr.7: 56082122-56082122 on Build GRCh38
rs754396196 Chr.7: 56081956-56081956 on Build GRCh38
rs765623978 Chr.7: 56082116-56082116 on Build GRCh38
rs754624575 Chr.7: 56081991-56081991 on Build GRCh38
rs56053511 Chr.7: 56081997-56081997 on Build GRCh38
rs762760644 Chr.7: 56081862-56081862 on Build GRCh38
rs921426676 Chr.7: 56081833-56081833 on Build GRCh38
rs373763601 Chr.7: 56082245-56082245 on Build GRCh38
rs781216239 Chr.7: 56082254-56082254 on Build GRCh38
rs752241091 Chr.7: 56082222-56082223 on Build GRCh38
rs761718565 Chr.7: 56081926-56081926 on Build GRCh38
rs759747023 Chr.7: 56081933-56081933 on Build GRCh38
rs375244738 Chr.7: 56082173-56082173 on Build GRCh38
rs957627212 Chr.7: 56082007-56082007 on Build GRCh38
rs776647595 Chr.7: 56082033-56082035 on Build GRCh38
rs768054720 Chr.7: 56081842-56081842 on Build GRCh38
rs751972729 Chr.7: 56082210-56082210 on Build GRCh38
rs771992500 Chr.7: 56082082-56082082 on Build GRCh38
rs759347162 Chr.7: 56082034-56082039 on Build GRCh38
rs146971620 Chr.7: 56082186-56082186 on Build GRCh38
rs777245675 Chr.7: 56082152-56082152 on Build GRCh38
rs377310121 Chr.7: 56082029-56082029 on Build GRCh38
rs149784104 Chr.7: 56081904-56081904 on Build GRCh38
rs115227938 Chr.7: 56082275-56082275 on Build GRCh38
rs763610501 Chr.7: 56082213-56082213 on Build GRCh38
rs765010787 Chr.7: 56081852-56081852 on Build GRCh38
rs868769445 Chr.7: 56082202-56082202 on Build GRCh38
rs202235154 Chr.7: 56081935-56081935 on Build GRCh38
rs745827411 Chr.7: 56082158-56082158 on Build GRCh38
rs13238521 Chr.7: 56082246-56082246 on Build GRCh38
rs746667766 Chr.7: 56081990-56081990 on Build GRCh38
rs544486646 Chr.7: 56081834-56081834 on Build GRCh38
rs529818565 Chr.7: 56081929-56081929 on Build GRCh38
rs780847047 Chr.7: 56082000-56082000 on Build GRCh38
rs770568626 Chr.7: 56082031-56082032 on Build GRCh38
rs370632474 Chr.7: 56082001-56082001 on Build GRCh38
rs756281200 Chr.7: 56081953-56081953 on Build GRCh38
rs766584104 Chr.7: 56081865-56081865 on Build GRCh38
rs747814066 Chr.7: 56082066-56082066 on Build GRCh38
rs766703359 Chr.7: 56082204-56082204 on Build GRCh38
rs73126331 Chr.7: 56082203-56082203 on Build GRCh38
rs755807991 Chr.7: 56082151-56082151 on Build GRCh38
rs759785327 Chr.7: 56082026-56082026 on Build GRCh38
rs765602938 Chr.7: 56082047-56082047 on Build GRCh38
rs760767064 Chr.7: 56082035-56082035 on Build GRCh38
rs373557654 Chr.7: 56082182-56082182 on Build GRCh38
rs777211779 Chr.7: 56082093-56082093 on Build GRCh38
rs147684619 Chr.7: 56081952-56081952 on Build GRCh38
rs200245343 Chr.7: 56082124-56082124 on Build GRCh38
rs367729730 Chr.7: 56082056-56082056 on Build GRCh38
rs372050246 Chr.7: 56081844-56081844 on Build GRCh38

View Full Product Details