Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74249661-74250204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC2
Entrez Gene ID
5982
Gene Name
replication factor C subunit 2
Gene Aliases
RFC40
UniGene
Hs.647062
Gene Chromosome Location
Chr.7: 74231502-74254458 on Build GRCh38
Transcripts
NM_001278791.1 NM_001278792.1 NM_001278793.1 NM_002914.4 NM_181471.2 XM_006716080.2 XM_017012492.1 XM_017012493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5956105 c.225+1delG p.? Chr.7: 74249738-74249738 on Build GRCh38
53525 c.198A>T p.E66D Chr.7: 74249766-74249766 on Build GRCh38
5588806 c.219C>T p.I73I Chr.7: 74249745-74249745 on Build GRCh38
5005878 c.225+6T>C p.? Chr.7: 74249733-74249733 on Build GRCh38
6178279 c.208C>G p.P70A Chr.7: 74249756-74249756 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs555040751 Chr.7: 74249888-74249888 on Build GRCh38
rs1048108084 Chr.7: 74249740-74249740 on Build GRCh38
rs888135381 Chr.7: 74249980-74249980 on Build GRCh38
rs377631238 Chr.7: 74250162-74250162 on Build GRCh38
rs540757412 Chr.7: 74250060-74250060 on Build GRCh38
rs782541693 Chr.7: 74249756-74249756 on Build GRCh38
rs552015440 Chr.7: 74249800-74249800 on Build GRCh38
rs1003862450 Chr.7: 74249788-74249788 on Build GRCh38
rs781988734 Chr.7: 74249718-74249718 on Build GRCh38
rs3135662 Chr.7: 74249897-74249897 on Build GRCh38
rs371145382 Chr.7: 74249844-74249844 on Build GRCh38
rs899459889 Chr.7: 74249702-74249702 on Build GRCh38
rs541044289 Chr.7: 74249917-74249917 on Build GRCh38
rs144214608 Chr.7: 74249784-74249784 on Build GRCh38
rs1003946918 Chr.7: 74249774-74249774 on Build GRCh38
rs113913982 Chr.7: 74250156-74250156 on Build GRCh38
rs386714687 Chr.7: 74250156-74250157 on Build GRCh38
rs782819905 Chr.7: 74249861-74249861 on Build GRCh38
rs781850817 Chr.7: 74249866-74249866 on Build GRCh38
rs560335015 Chr.7: 74250046-74250046 on Build GRCh38
rs58743010 Chr.7: 74250156-74250156 on Build GRCh38
rs966733284 Chr.7: 74250125-74250125 on Build GRCh38
rs781961081 Chr.7: 74249807-74249807 on Build GRCh38
rs573252152 Chr.7: 74250052-74250052 on Build GRCh38
rs782134891 Chr.7: 74249720-74249720 on Build GRCh38
rs529546213 Chr.7: 74249760-74249760 on Build GRCh38
rs782256983 Chr.7: 74249780-74249780 on Build GRCh38
rs562058083 Chr.7: 74250091-74250091 on Build GRCh38
rs932280675 Chr.7: 74249955-74249955 on Build GRCh38
rs530205853 Chr.7: 74250092-74250092 on Build GRCh38
rs782459114 Chr.7: 74249876-74249876 on Build GRCh38
rs782504934 Chr.7: 74249846-74249847 on Build GRCh38
rs782735953 Chr.7: 74249723-74249723 on Build GRCh38
rs201725860 Chr.7: 74249739-74249739 on Build GRCh38
rs373799762 Chr.7: 74249685-74249685 on Build GRCh38
rs1033993447 Chr.7: 74250077-74250077 on Build GRCh38
rs888209316 Chr.7: 74249758-74249758 on Build GRCh38
rs73705358 Chr.7: 74249793-74249793 on Build GRCh38
rs577519255 Chr.7: 74249954-74249954 on Build GRCh38
rs781940707 Chr.7: 74249823-74249823 on Build GRCh38
rs139813753 Chr.7: 74249892-74249892 on Build GRCh38
rs565149707 Chr.7: 74249817-74249817 on Build GRCh38
rs782639470 Chr.7: 74249779-74249779 on Build GRCh38
rs782716165 Chr.7: 74249742-74249742 on Build GRCh38
rs555974308 Chr.7: 74249979-74249980 on Build GRCh38
rs1003382736 Chr.7: 74250068-74250068 on Build GRCh38
rs782353167 Chr.7: 74249691-74249691 on Build GRCh38
rs568243742 Chr.7: 74249938-74249938 on Build GRCh38
rs782692037 Chr.7: 74250067-74250067 on Build GRCh38
rs781845584 Chr.7: 74249734-74249734 on Build GRCh38
rs781891419 Chr.7: 74249745-74249745 on Build GRCh38
rs201908314 Chr.7: 74249819-74249819 on Build GRCh38
rs781912578 Chr.7: 74249699-74249699 on Build GRCh38
rs544764582 Chr.7: 74250000-74250000 on Build GRCh38
rs191122686 Chr.7: 74249877-74249877 on Build GRCh38
rs782184213 Chr.7: 74250074-74250074 on Build GRCh38
rs374444469 Chr.7: 74249708-74249708 on Build GRCh38
rs978135008 Chr.7: 74250169-74250169 on Build GRCh38
rs782321180 Chr.7: 74249787-74249787 on Build GRCh38
rs782721098 Chr.7: 74249879-74249879 on Build GRCh38
rs186354392 Chr.7: 74250102-74250102 on Build GRCh38
rs1022300787 Chr.7: 74250121-74250121 on Build GRCh38
rs779794345 Chr.7: 74250109-74250109 on Build GRCh38
rs959333286 Chr.7: 74250088-74250088 on Build GRCh38
rs781997707 Chr.7: 74249688-74249688 on Build GRCh38
rs181573649 Chr.7: 74249933-74249933 on Build GRCh38
rs3135661 Chr.7: 74250136-74250136 on Build GRCh38
rs1048035821 Chr.7: 74249968-74249968 on Build GRCh38
rs782392522 Chr.7: 74249806-74249806 on Build GRCh38
rs782214195 Chr.7: 74249690-74249690 on Build GRCh38

View Full Product Details