Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74252440-74252985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC2
Entrez Gene ID
5982
Gene Name
replication factor C subunit 2
Gene Aliases
RFC40
UniGene
Hs.647062
Gene Chromosome Location
Chr.7: 74231502-74254458 on Build GRCh38
Transcripts
NM_001278791.1 NM_001278792.1 NM_001278793.1 NM_002914.4 NM_181471.2 XM_006716080.2 XM_017012492.1 XM_017012493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4721138 c.169G>A p.V57M Chr.7: 74252443-74252443 on Build GRCh38
6765228 c.153C>T p.V51V Chr.7: 74252459-74252459 on Build GRCh38
6283681 c.163G>A p.D55N Chr.7: 74252449-74252449 on Build GRCh38
7189988 c.114-1G>A p.? Chr.7: 74252499-74252499 on Build GRCh38
4796199 c.129G>A p.R43R Chr.7: 74252483-74252483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942004726 Chr.7: 74252789-74252789 on Build GRCh38
rs782396570 Chr.7: 74252493-74252493 on Build GRCh38
rs1030091543 Chr.7: 74252949-74252949 on Build GRCh38
rs3135659 Chr.7: 74252827-74252827 on Build GRCh38
rs782491148 Chr.7: 74252625-74252625 on Build GRCh38
rs199941958 Chr.7: 74252542-74252542 on Build GRCh38
rs1805393 Chr.7: 74252505-74252505 on Build GRCh38
rs781885929 Chr.7: 74252527-74252527 on Build GRCh38
rs569655818 Chr.7: 74252605-74252605 on Build GRCh38
rs563894833 Chr.7: 74252911-74252911 on Build GRCh38
rs1057478877 Chr.7: 74252584-74252584 on Build GRCh38
rs782059392 Chr.7: 74252492-74252492 on Build GRCh38
rs142748428 Chr.7: 74252647-74252647 on Build GRCh38
rs1021892345 Chr.7: 74252630-74252630 on Build GRCh38
rs182531887 Chr.7: 74252843-74252843 on Build GRCh38
rs782639412 Chr.7: 74252664-74252664 on Build GRCh38
rs558775029 Chr.7: 74252667-74252667 on Build GRCh38
rs892172232 Chr.7: 74252910-74252910 on Build GRCh38
rs897916211 Chr.7: 74252525-74252525 on Build GRCh38
rs938807153 Chr.7: 74252575-74252575 on Build GRCh38
rs781848136 Chr.7: 74252515-74252515 on Build GRCh38
rs189242666 Chr.7: 74252673-74252673 on Build GRCh38
rs782736816 Chr.7: 74252513-74252513 on Build GRCh38
rs894680763 Chr.7: 74252612-74252612 on Build GRCh38
rs781865519 Chr.7: 74252677-74252677 on Build GRCh38
rs956647348 Chr.7: 74252704-74252704 on Build GRCh38
rs953274076 Chr.7: 74252743-74252743 on Build GRCh38
rs571759445 Chr.7: 74252462-74252462 on Build GRCh38
rs1030728857 Chr.7: 74252695-74252695 on Build GRCh38
rs782638188 Chr.7: 74252536-74252536 on Build GRCh38
rs948868188 Chr.7: 74252851-74252851 on Build GRCh38
rs529627874 Chr.7: 74252796-74252796 on Build GRCh38
rs77326053 Chr.7: 74252523-74252523 on Build GRCh38
rs3135658 Chr.7: 74252840-74252840 on Build GRCh38
rs919287070 Chr.7: 74252805-74252805 on Build GRCh38
rs201288132 Chr.7: 74252537-74252537 on Build GRCh38
rs782029395 Chr.7: 74252497-74252497 on Build GRCh38
rs536622278 Chr.7: 74252764-74252764 on Build GRCh38
rs985978562 Chr.7: 74252760-74252760 on Build GRCh38
rs904789815 Chr.7: 74252893-74252893 on Build GRCh38
rs782446532 Chr.7: 74252546-74252546 on Build GRCh38
rs782673476 Chr.7: 74252863-74252863 on Build GRCh38
rs782546206 Chr.7: 74252832-74252832 on Build GRCh38
rs554797179 Chr.7: 74252560-74252560 on Build GRCh38
rs782736496 Chr.7: 74252524-74252524 on Build GRCh38
rs1052514170 Chr.7: 74252900-74252900 on Build GRCh38
rs3135657 Chr.7: 74252898-74252898 on Build GRCh38
rs782468306 Chr.7: 74252538-74252538 on Build GRCh38
rs781918654 Chr.7: 74252486-74252486 on Build GRCh38
rs1030578468 Chr.7: 74252727-74252727 on Build GRCh38
rs6964029 Chr.7: 74252461-74252461 on Build GRCh38
rs559416320 Chr.7: 74252784-74252784 on Build GRCh38
rs1013167023 Chr.7: 74252627-74252627 on Build GRCh38
rs543895433 Chr.7: 74252706-74252706 on Build GRCh38
rs782566268 Chr.7: 74252531-74252531 on Build GRCh38
rs782610861 Chr.7: 74252539-74252539 on Build GRCh38

View Full Product Details